Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 139005858-139006357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR23A
Entrez Gene ID
729627
Gene Name
proline rich 23A
Gene Aliases
-
UniGene
Hs.121314
Gene Chromosome Location
Chr.3: 139003962-139006268 on Build GRCh38
Transcripts
NM_001134659.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4489859 c.353C>T p.S118L Chr.3: 139005916-139005916 on Build GRCh38
1419465 c.159C>T p.A53A Chr.3: 139006110-139006110 on Build GRCh38
5565961 c.119G>A p.R40H Chr.3: 139006150-139006150 on Build GRCh38
326253 c.367G>C p.E123Q Chr.3: 139005902-139005902 on Build GRCh38
5600295 c.84C>G p.R28R Chr.3: 139006185-139006185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371641266 Chr.3: 139005957-139005957 on Build GRCh38
rs1055956525 Chr.3: 139006024-139006024 on Build GRCh38
rs185399823 Chr.3: 139006230-139006230 on Build GRCh38
rs780521253 Chr.3: 139005932-139005932 on Build GRCh38
rs370540186 Chr.3: 139005887-139005887 on Build GRCh38
rs946103031 Chr.3: 139006259-139006259 on Build GRCh38
rs961773532 Chr.3: 139006153-139006153 on Build GRCh38
rs541068087 Chr.3: 139005908-139005908 on Build GRCh38
rs758708891 Chr.3: 139005967-139005967 on Build GRCh38
rs768041335 Chr.3: 139006116-139006116 on Build GRCh38
rs766723245 Chr.3: 139006105-139006105 on Build GRCh38
rs780420071 Chr.3: 139005973-139005973 on Build GRCh38
rs368963461 Chr.3: 139005878-139005878 on Build GRCh38
rs752467767 Chr.3: 139005886-139005886 on Build GRCh38
rs773998997 Chr.3: 139005880-139005880 on Build GRCh38
rs753723162 Chr.3: 139005956-139005956 on Build GRCh38
rs772966586 Chr.3: 139006261-139006261 on Build GRCh38
rs1014426679 Chr.3: 139006310-139006310 on Build GRCh38
rs748823379 Chr.3: 139005941-139005941 on Build GRCh38
rs751784358 Chr.3: 139006106-139006106 on Build GRCh38
rs781770668 Chr.3: 139005991-139005991 on Build GRCh38
rs770432344 Chr.3: 139005942-139005942 on Build GRCh38
rs759894359 Chr.3: 139006284-139006284 on Build GRCh38
rs769354906 Chr.3: 139005935-139005935 on Build GRCh38
rs565622055 Chr.3: 139006178-139006178 on Build GRCh38
rs1055987791 Chr.3: 139006298-139006299 on Build GRCh38
rs368434952 Chr.3: 139005882-139005882 on Build GRCh38
rs763116906 Chr.3: 139006267-139006267 on Build GRCh38
rs770178041 Chr.3: 139005998-139005998 on Build GRCh38
rs757851980 Chr.3: 139006185-139006185 on Build GRCh38
rs750451446 Chr.3: 139005964-139005964 on Build GRCh38
rs567538155 Chr.3: 139006258-139006258 on Build GRCh38
rs778543109 Chr.3: 139006000-139006000 on Build GRCh38
rs761862407 Chr.3: 139006072-139006072 on Build GRCh38
rs994451823 Chr.3: 139006166-139006166 on Build GRCh38
rs751977957 Chr.3: 139005983-139005983 on Build GRCh38
rs762874670 Chr.3: 139006103-139006103 on Build GRCh38
rs768279615 Chr.3: 139006215-139006215 on Build GRCh38
rs11924188 Chr.3: 139005915-139005915 on Build GRCh38
rs781737657 Chr.3: 139005936-139005936 on Build GRCh38
rs774229446 Chr.3: 139006058-139006066 on Build GRCh38
rs755218514 Chr.3: 139006114-139006114 on Build GRCh38
rs759281278 Chr.3: 139005945-139005945 on Build GRCh38
rs752990068 Chr.3: 139006145-139006145 on Build GRCh38
rs757174685 Chr.3: 139006304-139006304 on Build GRCh38
rs543169978 Chr.3: 139005944-139005944 on Build GRCh38
rs1027554939 Chr.3: 139006175-139006175 on Build GRCh38
rs147495715 Chr.3: 139005884-139005884 on Build GRCh38
rs756711109 Chr.3: 139006169-139006169 on Build GRCh38
rs113724862 Chr.3: 139006320-139006320 on Build GRCh38
rs764729832 Chr.3: 139006224-139006224 on Build GRCh38
rs79499878 Chr.3: 139006314-139006314 on Build GRCh38
rs764028500 Chr.3: 139005951-139005951 on Build GRCh38
rs965852813 Chr.3: 139006242-139006242 on Build GRCh38
rs758521785 Chr.3: 139005927-139005927 on Build GRCh38
rs753109490 Chr.3: 139006305-139006305 on Build GRCh38
rs745361540 Chr.3: 139006008-139006008 on Build GRCh38
rs745392892 Chr.3: 139006180-139006180 on Build GRCh38
rs760483799 Chr.3: 139006044-139006044 on Build GRCh38
rs114330403 Chr.3: 139006016-139006016 on Build GRCh38
rs1001733491 Chr.3: 139006022-139006022 on Build GRCh38
rs746706978 Chr.3: 139006212-139006212 on Build GRCh38
rs765076882 Chr.3: 139006075-139006075 on Build GRCh38
rs747896331 Chr.3: 139006254-139006254 on Build GRCh38
rs572000331 Chr.3: 139005893-139005893 on Build GRCh38
rs753639101 Chr.3: 139006233-139006233 on Build GRCh38
rs545578674 Chr.3: 139006052-139006052 on Build GRCh38
rs139955662 Chr.3: 139006271-139006271 on Build GRCh38
rs779710022 Chr.3: 139006200-139006200 on Build GRCh38
rs775062437 Chr.3: 139006021-139006021 on Build GRCh38
rs750583037 Chr.3: 139006077-139006077 on Build GRCh38
rs749225015 Chr.3: 139005913-139005915 on Build GRCh38
rs201305464 Chr.3: 139005911-139005911 on Build GRCh38
rs764011784 Chr.3: 139005902-139005902 on Build GRCh38
rs547801428 Chr.3: 139006257-139006257 on Build GRCh38
rs767847107 Chr.3: 139006293-139006293 on Build GRCh38
rs895575376 Chr.3: 139006041-139006041 on Build GRCh38
rs765400623 Chr.3: 139005963-139005963 on Build GRCh38
rs530075937 Chr.3: 139006270-139006270 on Build GRCh38
rs770666982 Chr.3: 139006005-139006006 on Build GRCh38
rs747365941 Chr.3: 139005934-139005934 on Build GRCh38
rs755337408 Chr.3: 139005987-139005987 on Build GRCh38
rs768402548 Chr.3: 139006048-139006048 on Build GRCh38
rs779029059 Chr.3: 139005923-139005923 on Build GRCh38
rs748703025 Chr.3: 139005997-139005997 on Build GRCh38
rs750621808 Chr.3: 139005924-139005924 on Build GRCh38
rs775326572 Chr.3: 139005948-139005948 on Build GRCh38

View Full Product Details