Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 146076678-146077182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLOD2
Entrez Gene ID
5352
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 2
Gene Aliases
BRKS2, LH2, TLH
UniGene
Hs.477866
Gene Chromosome Location
Chr.3: 146069436-146161495 on Build GRCh38
Transcripts
NM_000935.2 NM_182943.2 XM_005247535.3 XM_017006625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1221222 c.1589G>T p.R530I Chr.3: 146076870-146076870 on Build GRCh38
1039560 c.1625A>G p.Y542C Chr.3: 146076834-146076834 on Build GRCh38
7198502 c.1613C>T p.S538F Chr.3: 146076846-146076846 on Build GRCh38
445747 c.1569A>T p.V523V Chr.3: 146076890-146076890 on Build GRCh38
3001911 c.1568T>C p.V523A Chr.3: 146076891-146076891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768461690 Chr.3: 146076845-146076845 on Build GRCh38
rs939955513 Chr.3: 146076728-146076728 on Build GRCh38
rs1007498499 Chr.3: 146076808-146076808 on Build GRCh38
rs144881583 Chr.3: 146076842-146076842 on Build GRCh38
rs891593198 Chr.3: 146077108-146077108 on Build GRCh38
rs201821424 Chr.3: 146076846-146076846 on Build GRCh38
rs773230412 Chr.3: 146076865-146076865 on Build GRCh38
rs771612190 Chr.3: 146076887-146076887 on Build GRCh38
rs142623177 Chr.3: 146076990-146076993 on Build GRCh38
rs1050050268 Chr.3: 146077078-146077078 on Build GRCh38
rs768925888 Chr.3: 146076989-146076990 on Build GRCh38
rs781476978 Chr.3: 146076736-146076739 on Build GRCh38
rs757927611 Chr.3: 146076935-146076935 on Build GRCh38
rs1051872709 Chr.3: 146076765-146076765 on Build GRCh38
rs543580697 Chr.3: 146076755-146076755 on Build GRCh38
rs983520532 Chr.3: 146076933-146076933 on Build GRCh38
rs549890582 Chr.3: 146076982-146076982 on Build GRCh38
rs748332300 Chr.3: 146076851-146076851 on Build GRCh38
rs191742453 Chr.3: 146076977-146076977 on Build GRCh38
rs999033370 Chr.3: 146077142-146077142 on Build GRCh38
rs183525329 Chr.3: 146077103-146077103 on Build GRCh38
rs754709890 Chr.3: 146076947-146076947 on Build GRCh38
rs552693350 Chr.3: 146076938-146076938 on Build GRCh38
rs148051196 Chr.3: 146076801-146076801 on Build GRCh38
rs1044305094 Chr.3: 146076710-146076712 on Build GRCh38
rs376954849 Chr.3: 146076824-146076824 on Build GRCh38
rs115199093 Chr.3: 146076820-146076820 on Build GRCh38
rs964918415 Chr.3: 146076889-146076889 on Build GRCh38
rs950005358 Chr.3: 146076959-146076959 on Build GRCh38
rs375572526 Chr.3: 146076911-146076911 on Build GRCh38
rs778350525 Chr.3: 146076813-146076813 on Build GRCh38
rs972653622 Chr.3: 146076940-146076940 on Build GRCh38
rs141850775 Chr.3: 146076812-146076812 on Build GRCh38
rs901009727 Chr.3: 146076747-146076747 on Build GRCh38
rs150609339 Chr.3: 146076815-146076815 on Build GRCh38
rs755799423 Chr.3: 146076784-146076784 on Build GRCh38
rs759935455 Chr.3: 146076897-146076897 on Build GRCh38
rs202113559 Chr.3: 146076993-146076993 on Build GRCh38
rs763766109 Chr.3: 146076799-146076799 on Build GRCh38
rs531816185 Chr.3: 146076978-146076978 on Build GRCh38
rs889195385 Chr.3: 146077099-146077099 on Build GRCh38
rs535422814 Chr.3: 146077004-146077004 on Build GRCh38
rs377273732 Chr.3: 146076987-146076987 on Build GRCh38
rs748214459 Chr.3: 146076753-146076755 on Build GRCh38
rs950950545 Chr.3: 146076926-146076926 on Build GRCh38
rs909589117 Chr.3: 146076924-146076924 on Build GRCh38
rs943493700 Chr.3: 146077104-146077104 on Build GRCh38
rs1032220027 Chr.3: 146076831-146076831 on Build GRCh38
rs565918046 Chr.3: 146077038-146077038 on Build GRCh38
rs1054784968 Chr.3: 146077061-146077061 on Build GRCh38
rs1036863740 Chr.3: 146076738-146076738 on Build GRCh38
rs904538356 Chr.3: 146077136-146077136 on Build GRCh38
rs1050842173 Chr.3: 146077135-146077135 on Build GRCh38
rs201260094 Chr.3: 146076991-146076991 on Build GRCh38
rs779791611 Chr.3: 146076828-146076828 on Build GRCh38
rs935901449 Chr.3: 146077005-146077005 on Build GRCh38
rs751503992 Chr.3: 146076939-146076939 on Build GRCh38
rs568235662 Chr.3: 146076994-146076994 on Build GRCh38
rs867916692 Chr.3: 146076876-146076876 on Build GRCh38
rs372618583 Chr.3: 146076985-146076985 on Build GRCh38
rs766838820 Chr.3: 146076737-146076737 on Build GRCh38
rs761443520 Chr.3: 146076914-146076914 on Build GRCh38
rs775017069 Chr.3: 146076894-146076894 on Build GRCh38
rs891663945 Chr.3: 146076806-146076806 on Build GRCh38
rs749987466 Chr.3: 146076922-146076922 on Build GRCh38
rs375780156 Chr.3: 146076899-146076899 on Build GRCh38
rs1054290688 Chr.3: 146076759-146076759 on Build GRCh38
rs35274473 Chr.3: 146076991-146076994 on Build GRCh38
rs763622304 Chr.3: 146076734-146076734 on Build GRCh38
rs867435085 Chr.3: 146076896-146076896 on Build GRCh38
rs547395378 Chr.3: 146077030-146077030 on Build GRCh38
rs1027818795 Chr.3: 146076912-146076912 on Build GRCh38
rs368297159 Chr.3: 146076984-146076984 on Build GRCh38
rs369619513 Chr.3: 146076921-146076921 on Build GRCh38
rs763520586 Chr.3: 146076868-146076868 on Build GRCh38
rs557922072 Chr.3: 146077120-146077120 on Build GRCh38
rs933860401 Chr.3: 146077074-146077074 on Build GRCh38

View Full Product Details