Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149324783-149325330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TM4SF18
Entrez Gene ID
116441
Gene Name
transmembrane 4 L six family member 18
Gene Aliases
L6D
UniGene
Hs.22026
Gene Chromosome Location
Chr.3: 149318498-149333761 on Build GRCh38
Transcripts
NM_001184723.1 NM_138786.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5824145 c.354G>C p.G118G Chr.3: 149324936-149324936 on Build GRCh38
6863266 c.321C>T p.C107C Chr.3: 149324969-149324969 on Build GRCh38
4591337 c.365G>T p.R122L Chr.3: 149324925-149324925 on Build GRCh38
72996 c.334G>A p.A112T Chr.3: 149324956-149324956 on Build GRCh38
3588583 c.297C>T p.L99L Chr.3: 149324993-149324993 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906084273 Chr.3: 149325089-149325089 on Build GRCh38
rs966580989 Chr.3: 149325136-149325136 on Build GRCh38
rs905884935 Chr.3: 149325300-149325300 on Build GRCh38
rs761030192 Chr.3: 149324909-149324909 on Build GRCh38
rs375164046 Chr.3: 149325108-149325108 on Build GRCh38
rs771098900 Chr.3: 149325043-149325044 on Build GRCh38
rs867624346 Chr.3: 149324841-149324841 on Build GRCh38
rs754150803 Chr.3: 149324959-149324959 on Build GRCh38
rs772461069 Chr.3: 149324976-149324976 on Build GRCh38
rs777428789 Chr.3: 149324956-149324956 on Build GRCh38
rs116569142 Chr.3: 149324809-149324809 on Build GRCh38
rs960264122 Chr.3: 149325125-149325125 on Build GRCh38
rs564387142 Chr.3: 149325164-149325164 on Build GRCh38
rs898695062 Chr.3: 149325045-149325045 on Build GRCh38
rs373451340 Chr.3: 149325036-149325036 on Build GRCh38
rs1043044693 Chr.3: 149325041-149325041 on Build GRCh38
rs760247601 Chr.3: 149324808-149324808 on Build GRCh38
rs145414265 Chr.3: 149324881-149324881 on Build GRCh38
rs749405254 Chr.3: 149324920-149324920 on Build GRCh38
rs780876271 Chr.3: 149324867-149324867 on Build GRCh38
rs1033684481 Chr.3: 149325071-149325071 on Build GRCh38
rs769129238 Chr.3: 149324993-149324993 on Build GRCh38
rs150033600 Chr.3: 149325214-149325214 on Build GRCh38
rs959757184 Chr.3: 149324816-149324816 on Build GRCh38
rs887331235 Chr.3: 149325006-149325006 on Build GRCh38
rs770531463 Chr.3: 149325031-149325031 on Build GRCh38
rs757843505 Chr.3: 149324832-149324832 on Build GRCh38
rs765923820 Chr.3: 149324916-149324916 on Build GRCh38
rs992090407 Chr.3: 149325114-149325114 on Build GRCh38
rs994304170 Chr.3: 149325138-149325138 on Build GRCh38
rs759501091 Chr.3: 149324921-149324921 on Build GRCh38
rs369012204 Chr.3: 149324997-149324997 on Build GRCh38
rs530503158 Chr.3: 149325262-149325262 on Build GRCh38
rs193070674 Chr.3: 149324913-149324913 on Build GRCh38
rs952475879 Chr.3: 149325148-149325148 on Build GRCh38
rs374999686 Chr.3: 149324964-149324964 on Build GRCh38
rs111772436 Chr.3: 149324911-149324911 on Build GRCh38
rs937580725 Chr.3: 149325305-149325305 on Build GRCh38
rs777995316 Chr.3: 149324882-149324882 on Build GRCh38
rs766927558 Chr.3: 149325062-149325062 on Build GRCh38
rs764806795 Chr.3: 149324910-149324910 on Build GRCh38
rs776086170 Chr.3: 149324926-149324926 on Build GRCh38
rs574152896 Chr.3: 149325029-149325030 on Build GRCh38
rs543940053 Chr.3: 149325294-149325294 on Build GRCh38
rs755998813 Chr.3: 149324930-149324930 on Build GRCh38
rs974985024 Chr.3: 149325180-149325180 on Build GRCh38
rs559660859 Chr.3: 149325154-149325154 on Build GRCh38
rs754891450 Chr.3: 149324852-149324852 on Build GRCh38
rs976681043 Chr.3: 149325181-149325181 on Build GRCh38
rs796082834 Chr.3: 149325160-149325160 on Build GRCh38
rs1047418051 Chr.3: 149325206-149325206 on Build GRCh38
rs773855560 Chr.3: 149325034-149325034 on Build GRCh38
rs1002350749 Chr.3: 149325291-149325291 on Build GRCh38
rs61094734 Chr.3: 149324849-149324849 on Build GRCh38
rs760501676 Chr.3: 149325072-149325072 on Build GRCh38
rs745899433 Chr.3: 149324966-149324966 on Build GRCh38
rs16861780 Chr.3: 149325140-149325140 on Build GRCh38
rs989291767 Chr.3: 149325230-149325230 on Build GRCh38
rs910894640 Chr.3: 149325155-149325155 on Build GRCh38
rs777960709 Chr.3: 149324860-149324860 on Build GRCh38
rs780551984 Chr.3: 149324992-149324992 on Build GRCh38
rs771203500 Chr.3: 149324889-149324889 on Build GRCh38
rs762617469 Chr.3: 149325013-149325013 on Build GRCh38
rs746425743 Chr.3: 149324897-149324897 on Build GRCh38
rs898743788 Chr.3: 149325282-149325282 on Build GRCh38
rs774540427 Chr.3: 149324895-149324895 on Build GRCh38
rs767426141 Chr.3: 149324924-149324924 on Build GRCh38
rs151260047 Chr.3: 149324925-149324925 on Build GRCh38
rs775748630 Chr.3: 149324902-149324902 on Build GRCh38
rs2291602 Chr.3: 149325123-149325123 on Build GRCh38
rs796152785 Chr.3: 149325198-149325198 on Build GRCh38
rs142278288 Chr.3: 149324919-149324919 on Build GRCh38
rs747939117 Chr.3: 149324880-149324880 on Build GRCh38
rs749509419 Chr.3: 149324885-149324885 on Build GRCh38
rs372677223 Chr.3: 149324954-149324954 on Build GRCh38
rs1004499938 Chr.3: 149325035-149325035 on Build GRCh38
rs201633894 Chr.3: 149324848-149324848 on Build GRCh38
rs555886702 Chr.3: 149324963-149324963 on Build GRCh38
rs554987306 Chr.3: 149324934-149324934 on Build GRCh38
rs963106729 Chr.3: 149324826-149324826 on Build GRCh38
rs960795154 Chr.3: 149325080-149325080 on Build GRCh38
rs140666552 Chr.3: 149324915-149324915 on Build GRCh38
rs752749605 Chr.3: 149325063-149325063 on Build GRCh38
rs751022866 Chr.3: 149324917-149324917 on Build GRCh38
rs74981119 Chr.3: 149324900-149324900 on Build GRCh38
rs1001758788 Chr.3: 149325104-149325104 on Build GRCh38
rs73168811 Chr.3: 149325258-149325258 on Build GRCh38
rs765365251 Chr.3: 149325213-149325213 on Build GRCh38
rs913796736 Chr.3: 149324901-149324901 on Build GRCh38

View Full Product Details