Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 158354568-158355096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSRC1
Entrez Gene ID
51319
Gene Name
arginine and serine rich coiled-coil 1
Gene Aliases
BM-011, SFRS21, SRrp53
UniGene
Hs.591283
Gene Chromosome Location
Chr.3: 158110052-158544835 on Build GRCh38
Transcripts
NM_001271834.1 NM_001271838.1 NM_016625.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6163687 c.561A>G p.L187L Chr.3: 158354886-158354886 on Build GRCh38
7388289 c.536C>G p.P179R Chr.3: 158354861-158354861 on Build GRCh38
6562888 c.574G>C p.E192Q Chr.3: 158354899-158354899 on Build GRCh38
6279401 c.532-9C>T p.? Chr.3: 158354848-158354848 on Build GRCh38
6529103 c.579T>C p.A193A Chr.3: 158354904-158354904 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs566894425 Chr.3: 158355003-158355003 on Build GRCh38
rs964169685 Chr.3: 158354712-158354712 on Build GRCh38
rs746510843 Chr.3: 158354855-158354855 on Build GRCh38
rs202050721 Chr.3: 158354997-158354997 on Build GRCh38
rs779425478 Chr.3: 158354973-158354973 on Build GRCh38
rs750216356 Chr.3: 158354844-158354844 on Build GRCh38
rs760910684 Chr.3: 158354933-158354933 on Build GRCh38
rs765032522 Chr.3: 158354835-158354835 on Build GRCh38
rs1052686065 Chr.3: 158355047-158355047 on Build GRCh38
rs760068831 Chr.3: 158354833-158354834 on Build GRCh38
rs778747258 Chr.3: 158354768-158354768 on Build GRCh38
rs991408613 Chr.3: 158354648-158354648 on Build GRCh38
rs758619121 Chr.3: 158354799-158354799 on Build GRCh38
rs751541023 Chr.3: 158354949-158354949 on Build GRCh38
rs5853823 Chr.3: 158354988-158354988 on Build GRCh38
rs978072937 Chr.3: 158354620-158354620 on Build GRCh38
rs1048836773 Chr.3: 158354662-158354662 on Build GRCh38
rs542982459 Chr.3: 158354630-158354630 on Build GRCh38
rs930399435 Chr.3: 158354660-158354660 on Build GRCh38
rs754550509 Chr.3: 158354856-158354856 on Build GRCh38
rs35682267 Chr.3: 158354963-158354963 on Build GRCh38
rs775734620 Chr.3: 158354929-158354929 on Build GRCh38
rs750175086 Chr.3: 158354939-158354939 on Build GRCh38
rs991959556 Chr.3: 158354957-158354957 on Build GRCh38
rs201732321 Chr.3: 158354848-158354848 on Build GRCh38
rs1040001026 Chr.3: 158354697-158354697 on Build GRCh38
rs767752552 Chr.3: 158354921-158354921 on Build GRCh38
rs779979967 Chr.3: 158354850-158354850 on Build GRCh38
rs534212567 Chr.3: 158355068-158355068 on Build GRCh38
rs753006165 Chr.3: 158354823-158354823 on Build GRCh38
rs771571979 Chr.3: 158354814-158354815 on Build GRCh38
rs192696074 Chr.3: 158354834-158354834 on Build GRCh38
rs76781544 Chr.3: 158354996-158354998 on Build GRCh38
rs771778634 Chr.3: 158354966-158354966 on Build GRCh38
rs928229256 Chr.3: 158354624-158354624 on Build GRCh38
rs764228441 Chr.3: 158354938-158354938 on Build GRCh38
rs936357979 Chr.3: 158355044-158355044 on Build GRCh38
rs1050686772 Chr.3: 158354796-158354796 on Build GRCh38
rs771452059 Chr.3: 158354879-158354879 on Build GRCh38
rs754464122 Chr.3: 158354950-158354950 on Build GRCh38
rs764152717 Chr.3: 158354835-158354835 on Build GRCh38
rs749623927 Chr.3: 158354869-158354869 on Build GRCh38
rs991049465 Chr.3: 158354976-158354976 on Build GRCh38
rs901777546 Chr.3: 158354707-158354707 on Build GRCh38
rs924961712 Chr.3: 158355020-158355020 on Build GRCh38
rs775111953 Chr.3: 158354833-158354833 on Build GRCh38
rs397689690 Chr.3: 158354998-158354998 on Build GRCh38
rs996081407 Chr.3: 158354750-158354750 on Build GRCh38
rs916721282 Chr.3: 158354659-158354660 on Build GRCh38
rs773589303 Chr.3: 158354866-158354866 on Build GRCh38
rs528316757 Chr.3: 158354800-158354800 on Build GRCh38
rs892632594 Chr.3: 158354601-158354601 on Build GRCh38
rs201295997 Chr.3: 158354988-158354988 on Build GRCh38
rs373008107 Chr.3: 158354811-158354811 on Build GRCh38
rs750195075 Chr.3: 158354848-158354848 on Build GRCh38
rs556640103 Chr.3: 158354998-158354998 on Build GRCh38
rs1019672480 Chr.3: 158354699-158354699 on Build GRCh38
rs138991341 Chr.3: 158354918-158354918 on Build GRCh38
rs368294453 Chr.3: 158354834-158354835 on Build GRCh38
rs2698321 Chr.3: 158354655-158354655 on Build GRCh38
rs1034997244 Chr.3: 158354922-158354922 on Build GRCh38
rs1008441064 Chr.3: 158354687-158354687 on Build GRCh38
rs561057096 Chr.3: 158354639-158354639 on Build GRCh38
rs75924897 Chr.3: 158354668-158354668 on Build GRCh38
rs371406316 Chr.3: 158354863-158354863 on Build GRCh38
rs907623317 Chr.3: 158354663-158354663 on Build GRCh38
rs1026969121 Chr.3: 158354791-158354791 on Build GRCh38
rs775062006 Chr.3: 158354905-158354905 on Build GRCh38
rs940392110 Chr.3: 158354681-158354683 on Build GRCh38
rs766300428 Chr.3: 158354947-158354947 on Build GRCh38
rs764275680 Chr.3: 158354870-158354870 on Build GRCh38
rs762716559 Chr.3: 158354941-158354941 on Build GRCh38
rs769358485 Chr.3: 158354865-158354865 on Build GRCh38
rs761779538 Chr.3: 158354835-158354836 on Build GRCh38
rs917626445 Chr.3: 158354992-158354992 on Build GRCh38

View Full Product Details