Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 158696973-158697474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RARRES1
Entrez Gene ID
5918
Gene Name
retinoic acid receptor responder 1
Gene Aliases
LXNL, PERG-1, TIG1
UniGene
Hs.131269
Gene Chromosome Location
Chr.3: 158697103-158732944 on Build GRCh38
Transcripts
NM_206963.1 XM_005247686.4

SNP Detail

SNP ID Chromosome Location
rs988825844 Chr.3: 158697301-158697301 on Build GRCh38
rs910957423 Chr.3: 158697328-158697328 on Build GRCh38
rs989246331 Chr.3: 158697186-158697186 on Build GRCh38
rs577314682 Chr.3: 158697212-158697212 on Build GRCh38
rs1008531920 Chr.3: 158697193-158697193 on Build GRCh38
rs576098417 Chr.3: 158697364-158697364 on Build GRCh38
rs554629521 Chr.3: 158697315-158697315 on Build GRCh38
rs949338317 Chr.3: 158697305-158697305 on Build GRCh38
rs146450744 Chr.3: 158697433-158697433 on Build GRCh38
rs994558877 Chr.3: 158697059-158697059 on Build GRCh38
rs914170482 Chr.3: 158697303-158697303 on Build GRCh38
rs1025960987 Chr.3: 158697168-158697168 on Build GRCh38
rs750580647 Chr.3: 158697277-158697277 on Build GRCh38
rs993860901 Chr.3: 158697101-158697101 on Build GRCh38
rs10276 Chr.3: 158697377-158697377 on Build GRCh38
rs954556331 Chr.3: 158697188-158697188 on Build GRCh38
rs781099133 Chr.3: 158697023-158697024 on Build GRCh38
rs1049418151 Chr.3: 158697237-158697237 on Build GRCh38
rs966431379 Chr.3: 158697203-158697203 on Build GRCh38
rs1037536620 Chr.3: 158697407-158697407 on Build GRCh38
rs536377098 Chr.3: 158697244-158697244 on Build GRCh38
rs757561874 Chr.3: 158697119-158697119 on Build GRCh38
rs564894642 Chr.3: 158697406-158697406 on Build GRCh38
rs958314316 Chr.3: 158697249-158697249 on Build GRCh38
rs982009893 Chr.3: 158697087-158697087 on Build GRCh38
rs899074505 Chr.3: 158697100-158697100 on Build GRCh38
rs75905420 Chr.3: 158697374-158697374 on Build GRCh38
rs922252994 Chr.3: 158697218-158697218 on Build GRCh38
rs534576007 Chr.3: 158697085-158697085 on Build GRCh38
rs1046059858 Chr.3: 158697435-158697435 on Build GRCh38
rs553173835 Chr.3: 158697086-158697086 on Build GRCh38
rs11718269 Chr.3: 158697207-158697207 on Build GRCh38
rs897182097 Chr.3: 158697007-158697007 on Build GRCh38
rs914974476 Chr.3: 158697197-158697197 on Build GRCh38
rs1032958560 Chr.3: 158697246-158697249 on Build GRCh38
rs1033518254 Chr.3: 158697091-158697091 on Build GRCh38
rs140860553 Chr.3: 158697445-158697445 on Build GRCh38
rs933648476 Chr.3: 158697221-158697221 on Build GRCh38
rs78456125 Chr.3: 158697235-158697235 on Build GRCh38
rs141453852 Chr.3: 158697365-158697365 on Build GRCh38

View Full Product Details