Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 165830585-165831089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCHE
Entrez Gene ID
590
Gene Name
butyrylcholinesterase
Gene Aliases
CHE1, CHE2, E1
UniGene
Hs.420483
Gene Chromosome Location
Chr.3: 165772904-165837467 on Build GRCh38
Transcripts
XM_017006967.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1040859 c.92A>G p.D31G Chr.3: 165830942-165830942 on Build GRCh38
729275 c.83C>A p.T28N Chr.3: 165830951-165830951 on Build GRCh38
4665368 c.319T>C p.S107P Chr.3: 165830715-165830715 on Build GRCh38
115767 c.327G>C p.M109I Chr.3: 165830707-165830707 on Build GRCh38
5686694 c.258C>A p.A86A Chr.3: 165830776-165830776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764097445 Chr.3: 165830706-165830706 on Build GRCh38
rs534940179 Chr.3: 165830702-165830702 on Build GRCh38
rs756863360 Chr.3: 165830877-165830877 on Build GRCh38
rs758046853 Chr.3: 165831020-165831020 on Build GRCh38
rs377250579 Chr.3: 165830873-165830873 on Build GRCh38
rs553041362 Chr.3: 165830645-165830645 on Build GRCh38
rs929258510 Chr.3: 165830993-165830993 on Build GRCh38
rs970265625 Chr.3: 165830630-165830630 on Build GRCh38
rs775897605 Chr.3: 165830691-165830691 on Build GRCh38
rs751406038 Chr.3: 165830712-165830712 on Build GRCh38
rs766168551 Chr.3: 165830616-165830616 on Build GRCh38
rs777330640 Chr.3: 165830828-165830828 on Build GRCh38
rs370612464 Chr.3: 165830736-165830736 on Build GRCh38
rs748658883 Chr.3: 165831052-165831052 on Build GRCh38
rs886058156 Chr.3: 165830848-165830848 on Build GRCh38
rs764846974 Chr.3: 165831015-165831015 on Build GRCh38
rs1057517439 Chr.3: 165830924-165830924 on Build GRCh38
rs748950254 Chr.3: 165830663-165830663 on Build GRCh38
rs148170012 Chr.3: 165830841-165830841 on Build GRCh38
rs137883811 Chr.3: 165830679-165830679 on Build GRCh38
rs764588882 Chr.3: 165830632-165830633 on Build GRCh38
rs755391002 Chr.3: 165831033-165831033 on Build GRCh38
rs767219925 Chr.3: 165830859-165830859 on Build GRCh38
rs765432162 Chr.3: 165830709-165830709 on Build GRCh38
rs979653503 Chr.3: 165830727-165830727 on Build GRCh38
rs531492019 Chr.3: 165830672-165830672 on Build GRCh38
rs1035513618 Chr.3: 165830687-165830687 on Build GRCh38
rs1800581 Chr.3: 165831049-165831049 on Build GRCh38
rs750309713 Chr.3: 165830934-165830934 on Build GRCh38
rs754968149 Chr.3: 165830628-165830628 on Build GRCh38
rs568396107 Chr.3: 165830898-165830898 on Build GRCh38
rs374649054 Chr.3: 165830788-165830788 on Build GRCh38
rs149045543 Chr.3: 165830707-165830707 on Build GRCh38
rs867455032 Chr.3: 165830810-165830810 on Build GRCh38
rs767311284 Chr.3: 165830882-165830882 on Build GRCh38
rs755600722 Chr.3: 165830653-165830653 on Build GRCh38
rs114355070 Chr.3: 165830870-165830870 on Build GRCh38
rs529294559 Chr.3: 165831034-165831034 on Build GRCh38
rs1044666393 Chr.3: 165830861-165830861 on Build GRCh38
rs56309853 Chr.3: 165830879-165830879 on Build GRCh38
rs754304373 Chr.3: 165830695-165830695 on Build GRCh38
rs777278160 Chr.3: 165830657-165830657 on Build GRCh38
rs1050901976 Chr.3: 165830760-165830760 on Build GRCh38
rs116097205 Chr.3: 165830852-165830852 on Build GRCh38
rs771168157 Chr.3: 165830969-165830969 on Build GRCh38
rs374939607 Chr.3: 165831013-165831013 on Build GRCh38
rs753381521 Chr.3: 165830804-165830804 on Build GRCh38
rs990121358 Chr.3: 165830739-165830739 on Build GRCh38
rs761197925 Chr.3: 165830700-165830700 on Build GRCh38
rs368402967 Chr.3: 165830875-165830875 on Build GRCh38
rs762091295 Chr.3: 165830708-165830708 on Build GRCh38
rs745482371 Chr.3: 165830825-165830825 on Build GRCh38
rs773207113 Chr.3: 165830963-165830963 on Build GRCh38
rs116047990 Chr.3: 165830915-165830915 on Build GRCh38
rs763759719 Chr.3: 165831009-165831009 on Build GRCh38
rs776735567 Chr.3: 165831010-165831010 on Build GRCh38
rs988127270 Chr.3: 165830692-165830692 on Build GRCh38
rs775196097 Chr.3: 165830981-165830981 on Build GRCh38
rs1057516784 Chr.3: 165830827-165830828 on Build GRCh38
rs149985624 Chr.3: 165831025-165831025 on Build GRCh38
rs776750503 Chr.3: 165830838-165830838 on Build GRCh38
rs374198965 Chr.3: 165830688-165830688 on Build GRCh38
rs765565615 Chr.3: 165830893-165830894 on Build GRCh38
rs780411054 Chr.3: 165830826-165830826 on Build GRCh38
rs763101056 Chr.3: 165830858-165830858 on Build GRCh38
rs781642185 Chr.3: 165830921-165830921 on Build GRCh38
rs746209182 Chr.3: 165830682-165830682 on Build GRCh38
rs3732880 Chr.3: 165830652-165830652 on Build GRCh38
rs967441914 Chr.3: 165830744-165830744 on Build GRCh38
rs531738678 Chr.3: 165830868-165830868 on Build GRCh38
rs759357575 Chr.3: 165830719-165830719 on Build GRCh38
rs886058157 Chr.3: 165831044-165831047 on Build GRCh38
rs1799807 Chr.3: 165830741-165830741 on Build GRCh38
rs769824118 Chr.3: 165830851-165830851 on Build GRCh38
rs757016659 Chr.3: 165830821-165830821 on Build GRCh38
rs780960236 Chr.3: 165830631-165830631 on Build GRCh38
rs757414059 Chr.3: 165830611-165830611 on Build GRCh38
rs762189020 Chr.3: 165830885-165830885 on Build GRCh38
rs769092899 Chr.3: 165830920-165830920 on Build GRCh38
rs769202966 Chr.3: 165830834-165830834 on Build GRCh38
rs114063013 Chr.3: 165830967-165830967 on Build GRCh38
rs55950599 Chr.3: 165831018-165831018 on Build GRCh38
rs150673481 Chr.3: 165830878-165830878 on Build GRCh38
rs769875412 Chr.3: 165830955-165830955 on Build GRCh38
rs939791364 Chr.3: 165831050-165831050 on Build GRCh38
rs760484389 Chr.3: 165830988-165830988 on Build GRCh38
rs540274817 Chr.3: 165831056-165831056 on Build GRCh38
rs367844208 Chr.3: 165830823-165830823 on Build GRCh38
rs201226143 Chr.3: 165830905-165830905 on Build GRCh38
rs377659208 Chr.3: 165831048-165831048 on Build GRCh38
rs200035175 Chr.3: 165830720-165830720 on Build GRCh38
rs372942772 Chr.3: 165830997-165830997 on Build GRCh38
rs751352739 Chr.3: 165830619-165830619 on Build GRCh38
rs748604050 Chr.3: 165830936-165830936 on Build GRCh38
rs751910853 Chr.3: 165831032-165831032 on Build GRCh38

View Full Product Details