Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 168027649-168028147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLIM4
Entrez Gene ID
27333
Gene Name
golgi integral membrane protein 4
Gene Aliases
GIMPC, GOLPH4, GPP130, P138
UniGene
Hs.143600
Gene Chromosome Location
Chr.3: 168008672-168095925 on Build GRCh38
Transcripts
NM_001308155.1 NM_014498.4 XM_005247364.2 XM_005247365.2 XM_017006183.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4406353 c.1554A>C p.G518G Chr.3: 168027797-168027797 on Build GRCh38
5449817 c.1618G>A p.A540T Chr.3: 168027733-168027733 on Build GRCh38
3589825 c.1559C>T p.P520L Chr.3: 168027792-168027792 on Build GRCh38
6402816 c.1547C>T p.A516V Chr.3: 168027804-168027804 on Build GRCh38
3589824 c.1611A>G p.E537E Chr.3: 168027740-168027740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747568376 Chr.3: 168027817-168027817 on Build GRCh38
rs770587551 Chr.3: 168027714-168027714 on Build GRCh38
rs374064981 Chr.3: 168027749-168027749 on Build GRCh38
rs199949272 Chr.3: 168027673-168027673 on Build GRCh38
rs769254713 Chr.3: 168027821-168027821 on Build GRCh38
rs753662382 Chr.3: 168027873-168027873 on Build GRCh38
rs968629006 Chr.3: 168027671-168027672 on Build GRCh38
rs975488183 Chr.3: 168027899-168027899 on Build GRCh38
rs916464604 Chr.3: 168028066-168028066 on Build GRCh38
rs748941813 Chr.3: 168027706-168027706 on Build GRCh38
rs772593489 Chr.3: 168027797-168027797 on Build GRCh38
rs776143740 Chr.3: 168027727-168027727 on Build GRCh38
rs149807288 Chr.3: 168027756-168027756 on Build GRCh38
rs947838966 Chr.3: 168027909-168027909 on Build GRCh38
rs995972331 Chr.3: 168028074-168028074 on Build GRCh38
rs182400073 Chr.3: 168027683-168027683 on Build GRCh38
rs754809155 Chr.3: 168027758-168027758 on Build GRCh38
rs566131140 Chr.3: 168027920-168027920 on Build GRCh38
rs965494427 Chr.3: 168027851-168027851 on Build GRCh38
rs764008073 Chr.3: 168027869-168027869 on Build GRCh38
rs774646515 Chr.3: 168027715-168027715 on Build GRCh38
rs143218887 Chr.3: 168027674-168027674 on Build GRCh38
rs1008814485 Chr.3: 168028031-168028031 on Build GRCh38
rs779073453 Chr.3: 168027989-168027989 on Build GRCh38
rs9861339 Chr.3: 168027915-168027915 on Build GRCh38
rs999835185 Chr.3: 168028016-168028016 on Build GRCh38
rs948505310 Chr.3: 168027969-168027970 on Build GRCh38
rs531602124 Chr.3: 168027726-168027726 on Build GRCh38
rs770733349 Chr.3: 168027791-168027792 on Build GRCh38
rs73878613 Chr.3: 168027842-168027842 on Build GRCh38
rs928686717 Chr.3: 168027874-168027874 on Build GRCh38
rs773515688 Chr.3: 168027835-168027835 on Build GRCh38
rs375332371 Chr.3: 168027963-168027963 on Build GRCh38
rs756793507 Chr.3: 168027885-168027885 on Build GRCh38
rs767349340 Chr.3: 168027840-168027840 on Build GRCh38
rs1057489328 Chr.3: 168027931-168027931 on Build GRCh38
rs759160961 Chr.3: 168027825-168027826 on Build GRCh38
rs991499290 Chr.3: 168028052-168028052 on Build GRCh38
rs1056417831 Chr.3: 168027890-168027890 on Build GRCh38
rs774110873 Chr.3: 168027799-168027801 on Build GRCh38
rs763338154 Chr.3: 168027704-168027704 on Build GRCh38
rs898207431 Chr.3: 168027917-168027917 on Build GRCh38
rs368944710 Chr.3: 168027891-168027891 on Build GRCh38
rs375314963 Chr.3: 168027791-168027791 on Build GRCh38
rs1025318132 Chr.3: 168027730-168027730 on Build GRCh38
rs748015175 Chr.3: 168027698-168027698 on Build GRCh38
rs549787801 Chr.3: 168027987-168027987 on Build GRCh38
rs372302207 Chr.3: 168027780-168027780 on Build GRCh38
rs746232358 Chr.3: 168027721-168027721 on Build GRCh38
rs371687959 Chr.3: 168027829-168027829 on Build GRCh38
rs141572176 Chr.3: 168028045-168028045 on Build GRCh38
rs568175935 Chr.3: 168027991-168027991 on Build GRCh38
rs73878614 Chr.3: 168027933-168027933 on Build GRCh38
rs570336135 Chr.3: 168027936-168027936 on Build GRCh38
rs756114882 Chr.3: 168027864-168027864 on Build GRCh38
rs374445265 Chr.3: 168027772-168027772 on Build GRCh38
rs947962542 Chr.3: 168028071-168028071 on Build GRCh38
rs748666127 Chr.3: 168027999-168027999 on Build GRCh38
rs537855658 Chr.3: 168027969-168027969 on Build GRCh38
rs368126514 Chr.3: 168027814-168027814 on Build GRCh38
rs777219528 Chr.3: 168027705-168027705 on Build GRCh38
rs557655172 Chr.3: 168028105-168028105 on Build GRCh38
rs978644277 Chr.3: 168027671-168027671 on Build GRCh38
rs923645866 Chr.3: 168027696-168027708 on Build GRCh38
rs370597666 Chr.3: 168027691-168027691 on Build GRCh38
rs916402485 Chr.3: 168027904-168027904 on Build GRCh38
rs772699821 Chr.3: 168028034-168028034 on Build GRCh38
rs764286955 Chr.3: 168027748-168027748 on Build GRCh38
rs111793973 Chr.3: 168027928-168027928 on Build GRCh38
rs370455114 Chr.3: 168027757-168027757 on Build GRCh38
rs200864087 Chr.3: 168027763-168027763 on Build GRCh38
rs76360762 Chr.3: 168028010-168028010 on Build GRCh38
rs967490857 Chr.3: 168027844-168027844 on Build GRCh38
rs539574793 Chr.3: 168028048-168028048 on Build GRCh38
rs767116701 Chr.3: 168027848-168027848 on Build GRCh38
rs534465111 Chr.3: 168027771-168027771 on Build GRCh38
rs776640778 Chr.3: 168027822-168027822 on Build GRCh38
rs1028429455 Chr.3: 168028086-168028086 on Build GRCh38
rs777309117 Chr.3: 168027775-168027775 on Build GRCh38
rs769328905 Chr.3: 168027699-168027699 on Build GRCh38
rs891733651 Chr.3: 168028029-168028029 on Build GRCh38
rs1012600398 Chr.3: 168027820-168027820 on Build GRCh38
rs767261323 Chr.3: 168027770-168027770 on Build GRCh38
rs977789927 Chr.3: 168027848-168027848 on Build GRCh38
rs890821531 Chr.3: 168027944-168027944 on Build GRCh38
rs1011660483 Chr.3: 168027994-168027994 on Build GRCh38
rs148304086 Chr.3: 168028023-168028023 on Build GRCh38
rs939261810 Chr.3: 168027886-168027886 on Build GRCh38
rs964549584 Chr.3: 168028046-168028046 on Build GRCh38
rs955807016 Chr.3: 168027779-168027779 on Build GRCh38
rs921316364 Chr.3: 168027901-168027904 on Build GRCh38
rs375951408 Chr.3: 168027781-168027781 on Build GRCh38
rs761219523 Chr.3: 168027728-168027728 on Build GRCh38
rs780290502 Chr.3: 168027834-168027834 on Build GRCh38

View Full Product Details