Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 182944119-182944647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCUN1D1
Entrez Gene ID
54165
Gene Name
defective in cullin neddylation 1 domain containing 1
Gene Aliases
DCNL1, DCUN1L1, RP42, SCCRO, SCRO, Tes3
UniGene
Hs.592789
Gene Chromosome Location
Chr.3: 182938158-182986364 on Build GRCh38
Transcripts
NM_001308101.1 NM_020640.3 XM_011512912.1 XM_011512914.1 XM_011512915.2 XM_011512916.2 XM_017006633.1 XM_017006634.1 XM_017006635.1

SNP Detail

SNP ID Chromosome Location
rs113844950 Chr.3: 182944480-182944480 on Build GRCh38
rs529537626 Chr.3: 182944604-182944604 on Build GRCh38
rs774404167 Chr.3: 182944465-182944465 on Build GRCh38
rs540369547 Chr.3: 182944495-182944495 on Build GRCh38
rs767446903 Chr.3: 182944496-182944496 on Build GRCh38
rs564111808 Chr.3: 182944402-182944402 on Build GRCh38
rs766916278 Chr.3: 182944262-182944262 on Build GRCh38
rs945707837 Chr.3: 182944148-182944148 on Build GRCh38
rs1036551128 Chr.3: 182944583-182944583 on Build GRCh38
rs773655512 Chr.3: 182944144-182944144 on Build GRCh38
rs1057949 Chr.3: 182944164-182944164 on Build GRCh38
rs183858153 Chr.3: 182944341-182944341 on Build GRCh38
rs7623786 Chr.3: 182944536-182944536 on Build GRCh38
rs764030129 Chr.3: 182944617-182944617 on Build GRCh38
rs541374767 Chr.3: 182944303-182944303 on Build GRCh38
rs143843351 Chr.3: 182944475-182944475 on Build GRCh38
rs532370144 Chr.3: 182944558-182944558 on Build GRCh38
rs981621364 Chr.3: 182944158-182944158 on Build GRCh38
rs771473591 Chr.3: 182944618-182944618 on Build GRCh38
rs369606848 Chr.3: 182944345-182944345 on Build GRCh38
rs370347804 Chr.3: 182944580-182944580 on Build GRCh38
rs200252908 Chr.3: 182944560-182944560 on Build GRCh38
rs752849631 Chr.3: 182944513-182944513 on Build GRCh38

View Full Product Details