Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 191304256-191304641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTS2B
Entrez Gene ID
257313
Gene Name
urotensin 2B
Gene Aliases
U2B, URP, UTS2D
UniGene
Hs.518492
Gene Chromosome Location
Chr.3: 191267155-191330536 on Build GRCh38
Transcripts
NM_198152.3 XM_011512631.1 XM_017006091.1

SNP Detail

SNP ID Chromosome Location
rs941336654 Chr.3: 191304320-191304320 on Build GRCh38
rs535650837 Chr.3: 191304484-191304484 on Build GRCh38
rs920577374 Chr.3: 191304460-191304460 on Build GRCh38
rs907972133 Chr.3: 191304422-191304422 on Build GRCh38
rs570042971 Chr.3: 191304406-191304406 on Build GRCh38
rs929164060 Chr.3: 191304289-191304289 on Build GRCh38
rs968980569 Chr.3: 191304352-191304352 on Build GRCh38
rs931934573 Chr.3: 191304465-191304465 on Build GRCh38
rs973405194 Chr.3: 191304438-191304438 on Build GRCh38
rs768889745 Chr.3: 191304571-191304571 on Build GRCh38
rs923229462 Chr.3: 191304589-191304589 on Build GRCh38
rs547109429 Chr.3: 191304489-191304489 on Build GRCh38
rs1000499811 Chr.3: 191304513-191304513 on Build GRCh38
rs758337115 Chr.3: 191304419-191304419 on Build GRCh38
rs73888536 Chr.3: 191304335-191304335 on Build GRCh38
rs911772741 Chr.3: 191304500-191304500 on Build GRCh38
rs569585804 Chr.3: 191304559-191304559 on Build GRCh38
rs769039226 Chr.3: 191304309-191304309 on Build GRCh38
rs1047138691 Chr.3: 191304304-191304304 on Build GRCh38
rs753449377 Chr.3: 191304340-191304340 on Build GRCh38
rs144939082 Chr.3: 191304554-191304554 on Build GRCh38
rs944554237 Chr.3: 191304501-191304501 on Build GRCh38
rs923092264 Chr.3: 191304281-191304281 on Build GRCh38
rs778620526 Chr.3: 191304381-191304381 on Build GRCh38
rs775888274 Chr.3: 191304526-191304526 on Build GRCh38
rs1010639562 Chr.3: 191304533-191304533 on Build GRCh38
rs499212 Chr.3: 191304504-191304504 on Build GRCh38
rs896638942 Chr.3: 191304423-191304423 on Build GRCh38
rs950543588 Chr.3: 191304593-191304593 on Build GRCh38
rs905419828 Chr.3: 191304363-191304363 on Build GRCh38
rs879557800 Chr.3: 191304482-191304482 on Build GRCh38
rs762699764 Chr.3: 191304415-191304415 on Build GRCh38
rs149917011 Chr.3: 191304507-191304507 on Build GRCh38
rs564100590 Chr.3: 191304369-191304369 on Build GRCh38
rs975950929 Chr.3: 191304572-191304572 on Build GRCh38
rs769932811 Chr.3: 191304525-191304525 on Build GRCh38
rs983696708 Chr.3: 191304594-191304594 on Build GRCh38
rs1002469801 Chr.3: 191304402-191304402 on Build GRCh38
rs533151098 Chr.3: 191304394-191304394 on Build GRCh38
rs777658102 Chr.3: 191304397-191304400 on Build GRCh38

View Full Product Details