Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 193307240-193307749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A5
Entrez Gene ID
344905
Gene Name
ATPase 13A5
Gene Aliases
-
UniGene
Hs.658032
Gene Chromosome Location
Chr.3: 193274789-193378824 on Build GRCh38
Transcripts
NM_198505.2 XM_011512770.1 XM_017006304.1 XM_017006305.1 NR_046758.1

Gene Symbol
ATP13A5-AS1
Entrez Gene ID
100874218
Gene Name
ATP13A5 antisense RNA 1
Gene Aliases
-
UniGene
Hs.407490
Gene Chromosome Location
Chr.3: 193307244-193314521 on Build GRCh38
Transcripts
NM_198505.2 XM_011512770.1 XM_017006304.1 XM_017006305.1 NR_046758.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1042612 c.2552G>A p.G851E Chr.3: 193307343-193307343 on Build GRCh38
3591387 c.2551G>A p.G851R Chr.3: 193307344-193307344 on Build GRCh38
5937073 c.2546G>A p.G849E Chr.3: 193307349-193307349 on Build GRCh38
5502823 c.2568G>A p.G856G Chr.3: 193307327-193307327 on Build GRCh38
159266 c.2558A>C p.N853T Chr.3: 193307337-193307337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs796952725 Chr.3: 193307654-193307654 on Build GRCh38
rs959338847 Chr.3: 193307475-193307475 on Build GRCh38
rs959389306 Chr.3: 193307697-193307697 on Build GRCh38
rs146601980 Chr.3: 193307552-193307552 on Build GRCh38
rs1056250181 Chr.3: 193307579-193307579 on Build GRCh38
rs747090170 Chr.3: 193307286-193307286 on Build GRCh38
rs938099129 Chr.3: 193307264-193307264 on Build GRCh38
rs181762374 Chr.3: 193307297-193307297 on Build GRCh38
rs939060685 Chr.3: 193307562-193307562 on Build GRCh38
rs915308407 Chr.3: 193307529-193307529 on Build GRCh38
rs557795744 Chr.3: 193307672-193307672 on Build GRCh38
rs550759788 Chr.3: 193307492-193307492 on Build GRCh38
rs755151644 Chr.3: 193307353-193307353 on Build GRCh38
rs537805451 Chr.3: 193307660-193307660 on Build GRCh38
rs900131305 Chr.3: 193307332-193307332 on Build GRCh38
rs773881035 Chr.3: 193307710-193307710 on Build GRCh38
rs773797922 Chr.3: 193307305-193307305 on Build GRCh38
rs549038689 Chr.3: 193307612-193307612 on Build GRCh38
rs756632105 Chr.3: 193307372-193307372 on Build GRCh38
rs113308293 Chr.3: 193307574-193307574 on Build GRCh38
rs770561926 Chr.3: 193307381-193307381 on Build GRCh38
rs1003308758 Chr.3: 193307265-193307265 on Build GRCh38
rs981341791 Chr.3: 193307451-193307451 on Build GRCh38
rs369457079 Chr.3: 193307319-193307319 on Build GRCh38
rs1012444602 Chr.3: 193307609-193307609 on Build GRCh38
rs118180922 Chr.3: 193307611-193307611 on Build GRCh38
rs564000420 Chr.3: 193307544-193307544 on Build GRCh38
rs1040627027 Chr.3: 193307598-193307598 on Build GRCh38
rs144058014 Chr.3: 193307273-193307273 on Build GRCh38
rs748135570 Chr.3: 193307384-193307386 on Build GRCh38
rs199531644 Chr.3: 193307358-193307358 on Build GRCh38
rs192341903 Chr.3: 193307658-193307658 on Build GRCh38
rs186998372 Chr.3: 193307481-193307481 on Build GRCh38
rs781517297 Chr.3: 193307362-193307362 on Build GRCh38
rs766721308 Chr.3: 193307336-193307336 on Build GRCh38
rs953939626 Chr.3: 193307450-193307450 on Build GRCh38
rs771662907 Chr.3: 193307407-193307407 on Build GRCh38
rs763933171 Chr.3: 193307326-193307326 on Build GRCh38
rs781751213 Chr.3: 193307293-193307293 on Build GRCh38
rs569009559 Chr.3: 193307547-193307547 on Build GRCh38
rs113530800 Chr.3: 193307329-193307329 on Build GRCh38
rs966489052 Chr.3: 193307366-193307366 on Build GRCh38
rs887514405 Chr.3: 193307633-193307633 on Build GRCh38
rs748736136 Chr.3: 193307371-193307371 on Build GRCh38
rs190608939 Chr.3: 193307269-193307269 on Build GRCh38
rs781534573 Chr.3: 193307373-193307373 on Build GRCh38
rs200704876 Chr.3: 193307301-193307301 on Build GRCh38
rs1042007700 Chr.3: 193307547-193307547 on Build GRCh38
rs550577641 Chr.3: 193307539-193307539 on Build GRCh38
rs1004601445 Chr.3: 193307636-193307636 on Build GRCh38
rs375398862 Chr.3: 193307315-193307315 on Build GRCh38
rs759145875 Chr.3: 193307313-193307313 on Build GRCh38
rs745342887 Chr.3: 193307392-193307392 on Build GRCh38
rs770231992 Chr.3: 193307300-193307300 on Build GRCh38
rs923639423 Chr.3: 193307553-193307553 on Build GRCh38
rs562069828 Chr.3: 193307387-193307387 on Build GRCh38
rs117392683 Chr.3: 193307546-193307546 on Build GRCh38
rs770578449 Chr.3: 193307304-193307304 on Build GRCh38
rs770129744 Chr.3: 193307657-193307657 on Build GRCh38
rs769339988 Chr.3: 193307452-193307453 on Build GRCh38
rs1035001479 Chr.3: 193307464-193307464 on Build GRCh38
rs61419346 Chr.3: 193307593-193307593 on Build GRCh38
rs985332688 Chr.3: 193307477-193307477 on Build GRCh38
rs141755386 Chr.3: 193307406-193307406 on Build GRCh38
rs749671038 Chr.3: 193307374-193307374 on Build GRCh38
rs748887779 Chr.3: 193307302-193307302 on Build GRCh38
rs375251485 Chr.3: 193307270-193307270 on Build GRCh38
rs915184279 Chr.3: 193307540-193307540 on Build GRCh38
rs758626416 Chr.3: 193307335-193307335 on Build GRCh38
rs546378308 Chr.3: 193307288-193307288 on Build GRCh38
rs751575510 Chr.3: 193307349-193307349 on Build GRCh38
rs998002171 Chr.3: 193307719-193307719 on Build GRCh38
rs753717942 Chr.3: 193307331-193307331 on Build GRCh38
rs978659467 Chr.3: 193307559-193307559 on Build GRCh38
rs775539503 Chr.3: 193307316-193307316 on Build GRCh38
rs971728047 Chr.3: 193307394-193307394 on Build GRCh38

View Full Product Details