Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 194358222-194358757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC15
Entrez Gene ID
131578
Gene Name
leucine rich repeat containing 15
Gene Aliases
LIB
UniGene
Hs.288467
Gene Chromosome Location
Chr.3: 194355247-194369743 on Build GRCh38
Transcripts
NM_001135057.2 NM_130830.4

SNP Detail

SNP ID Chromosome Location
rs1010635363 Chr.3: 194358455-194358455 on Build GRCh38
rs114196561 Chr.3: 194358675-194358675 on Build GRCh38
rs140406317 Chr.3: 194358657-194358657 on Build GRCh38
rs1034891069 Chr.3: 194358589-194358589 on Build GRCh38
rs567688469 Chr.3: 194358631-194358631 on Build GRCh38
rs758124237 Chr.3: 194358629-194358629 on Build GRCh38
rs796543265 Chr.3: 194358717-194358717 on Build GRCh38
rs534789387 Chr.3: 194358642-194358642 on Build GRCh38
rs1039540214 Chr.3: 194358538-194358538 on Build GRCh38
rs958854005 Chr.3: 194358607-194358607 on Build GRCh38
rs759704257 Chr.3: 194358517-194358517 on Build GRCh38
rs899679464 Chr.3: 194358404-194358404 on Build GRCh38
rs899586720 Chr.3: 194358295-194358295 on Build GRCh38
rs995622342 Chr.3: 194358322-194358322 on Build GRCh38
rs530835269 Chr.3: 194358476-194358476 on Build GRCh38
rs914644830 Chr.3: 194358444-194358444 on Build GRCh38
rs754560258 Chr.3: 194358354-194358354 on Build GRCh38
rs34984317 Chr.3: 194358412-194358413 on Build GRCh38
rs375546125 Chr.3: 194358389-194358389 on Build GRCh38
rs76922026 Chr.3: 194358692-194358692 on Build GRCh38
rs1015226412 Chr.3: 194358374-194358374 on Build GRCh38
rs569923350 Chr.3: 194358453-194358453 on Build GRCh38
rs747632590 Chr.3: 194358492-194358492 on Build GRCh38
rs1048198249 Chr.3: 194358597-194358597 on Build GRCh38
rs192133140 Chr.3: 194358587-194358587 on Build GRCh38
rs943923025 Chr.3: 194358281-194358281 on Build GRCh38
rs974279089 Chr.3: 194358630-194358630 on Build GRCh38
rs776414240 Chr.3: 194358363-194358363 on Build GRCh38
rs148617501 Chr.3: 194358668-194358668 on Build GRCh38
rs912513198 Chr.3: 194358502-194358502 on Build GRCh38
rs191524183 Chr.3: 194358389-194358389 on Build GRCh38
rs866534176 Chr.3: 194358432-194358432 on Build GRCh38
rs187446762 Chr.3: 194358384-194358384 on Build GRCh38
rs972048833 Chr.3: 194358381-194358381 on Build GRCh38
rs4974532 Chr.3: 194358353-194358353 on Build GRCh38
rs367910670 Chr.3: 194358279-194358279 on Build GRCh38
rs187262842 Chr.3: 194358445-194358445 on Build GRCh38
rs1026665869 Chr.3: 194358323-194358323 on Build GRCh38
rs899639534 Chr.3: 194358547-194358547 on Build GRCh38
rs547101531 Chr.3: 194358511-194358511 on Build GRCh38
rs1003143597 Chr.3: 194358580-194358580 on Build GRCh38
rs929628211 Chr.3: 194358351-194358351 on Build GRCh38
rs959155931 Chr.3: 194358682-194358682 on Build GRCh38
rs936504648 Chr.3: 194358723-194358723 on Build GRCh38
rs1020310679 Chr.3: 194358477-194358477 on Build GRCh38
rs561746690 Chr.3: 194358290-194358290 on Build GRCh38
rs1003915217 Chr.3: 194358620-194358620 on Build GRCh38
rs780026911 Chr.3: 194358332-194358332 on Build GRCh38
rs982826363 Chr.3: 194358656-194358656 on Build GRCh38
rs144927193 Chr.3: 194358429-194358429 on Build GRCh38
rs182561300 Chr.3: 194358436-194358436 on Build GRCh38
rs973819946 Chr.3: 194358289-194358289 on Build GRCh38
rs73081774 Chr.3: 194358709-194358709 on Build GRCh38
rs375512264 Chr.3: 194358575-194358575 on Build GRCh38
rs6772860 Chr.3: 194358258-194358258 on Build GRCh38

View Full Product Details