Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2947944-2948443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOP14
Entrez Gene ID
8602
Gene Name
NOP14 nucleolar protein
Gene Aliases
C4orf9, NOL14, RES4-25, RES425, UTP2
UniGene
Hs.627133
Gene Chromosome Location
Chr.4: 2937936-2963506 on Build GRCh38
Transcripts
NM_001291978.1 NM_001291979.1 NM_003703.2 NR_015453.2

Gene Symbol
NOP14-AS1
Entrez Gene ID
317648
Gene Name
NOP14 antisense RNA 1
Gene Aliases
C4orf10, RES4-24
UniGene
Hs.398178
Gene Chromosome Location
Chr.4: 2935546-2951078 on Build GRCh38
Transcripts
NM_001291978.1 NM_001291979.1 NM_003703.2 NR_015453.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6275143 c.1283C>A p.A428D Chr.4: 2948408-2948408 on Build GRCh38
4332177 c.1328C>T p.S443L Chr.4: 2948363-2948363 on Build GRCh38
5663095 c.1370G>A p.C457Y Chr.4: 2948321-2948321 on Build GRCh38
6275142 c.1283C>A p.A428D Chr.4: 2948408-2948408 on Build GRCh38
5663096 c.1370G>A p.C457Y Chr.4: 2948321-2948321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758025573 Chr.4: 2948271-2948271 on Build GRCh38
rs761618287 Chr.4: 2948117-2948117 on Build GRCh38
rs983802201 Chr.4: 2948201-2948201 on Build GRCh38
rs965761923 Chr.4: 2948160-2948160 on Build GRCh38
rs767752450 Chr.4: 2948240-2948240 on Build GRCh38
rs771475388 Chr.4: 2948318-2948318 on Build GRCh38
rs199774193 Chr.4: 2948251-2948251 on Build GRCh38
rs368314980 Chr.4: 2947967-2947987 on Build GRCh38
rs558275718 Chr.4: 2948004-2948004 on Build GRCh38
rs889548680 Chr.4: 2947998-2947998 on Build GRCh38
rs538689502 Chr.4: 2947989-2947989 on Build GRCh38
rs1039222710 Chr.4: 2948040-2948040 on Build GRCh38
rs1035550592 Chr.4: 2948152-2948152 on Build GRCh38
rs1011532876 Chr.4: 2948113-2948113 on Build GRCh38
rs142817070 Chr.4: 2948360-2948360 on Build GRCh38
rs971746640 Chr.4: 2948186-2948186 on Build GRCh38
rs545860744 Chr.4: 2948157-2948157 on Build GRCh38
rs145463176 Chr.4: 2948252-2948252 on Build GRCh38
rs552221383 Chr.4: 2948401-2948401 on Build GRCh38
rs148438590 Chr.4: 2948363-2948363 on Build GRCh38
rs201732207 Chr.4: 2948289-2948289 on Build GRCh38
rs7665452 Chr.4: 2948151-2948151 on Build GRCh38
rs1012222750 Chr.4: 2948098-2948098 on Build GRCh38
rs765012892 Chr.4: 2948259-2948259 on Build GRCh38
rs907205674 Chr.4: 2947996-2947996 on Build GRCh38
rs760634574 Chr.4: 2948245-2948245 on Build GRCh38
rs771135640 Chr.4: 2948372-2948374 on Build GRCh38
rs1021544588 Chr.4: 2948115-2948115 on Build GRCh38
rs189481728 Chr.4: 2948274-2948274 on Build GRCh38
rs1036175771 Chr.4: 2948408-2948408 on Build GRCh38
rs568837010 Chr.4: 2948409-2948409 on Build GRCh38
rs878985599 Chr.4: 2948330-2948330 on Build GRCh38
rs751087827 Chr.4: 2948394-2948394 on Build GRCh38
rs910868293 Chr.4: 2948211-2948211 on Build GRCh38
rs142506334 Chr.4: 2948399-2948399 on Build GRCh38
rs765068089 Chr.4: 2948395-2948395 on Build GRCh38
rs537888687 Chr.4: 2948412-2948412 on Build GRCh38
rs774653227 Chr.4: 2948405-2948405 on Build GRCh38
rs1048053397 Chr.4: 2947993-2947993 on Build GRCh38
rs566615320 Chr.4: 2947988-2947988 on Build GRCh38
rs776617803 Chr.4: 2948392-2948392 on Build GRCh38
rs774831123 Chr.4: 2948340-2948340 on Build GRCh38
rs997808347 Chr.4: 2948101-2948101 on Build GRCh38
rs754387402 Chr.4: 2948296-2948296 on Build GRCh38
rs749804670 Chr.4: 2948314-2948314 on Build GRCh38
rs901991937 Chr.4: 2948092-2948092 on Build GRCh38
rs368143908 Chr.4: 2948006-2948006 on Build GRCh38
rs367669294 Chr.4: 2948288-2948288 on Build GRCh38
rs746129234 Chr.4: 2948355-2948355 on Build GRCh38
rs185021659 Chr.4: 2948266-2948266 on Build GRCh38
rs562766777 Chr.4: 2948188-2948188 on Build GRCh38
rs76809068 Chr.4: 2948109-2948109 on Build GRCh38
rs931184247 Chr.4: 2948196-2948196 on Build GRCh38
rs918510217 Chr.4: 2948193-2948193 on Build GRCh38
rs746549117 Chr.4: 2948294-2948294 on Build GRCh38
rs376992004 Chr.4: 2948070-2948070 on Build GRCh38
rs201713664 Chr.4: 2948312-2948312 on Build GRCh38
rs918285546 Chr.4: 2947972-2947972 on Build GRCh38
rs764017567 Chr.4: 2948373-2948373 on Build GRCh38
rs889921121 Chr.4: 2948137-2948137 on Build GRCh38
rs759815101 Chr.4: 2948228-2948228 on Build GRCh38
rs537771384 Chr.4: 2948031-2948031 on Build GRCh38
rs757050834 Chr.4: 2948254-2948254 on Build GRCh38
rs370776197 Chr.4: 2948321-2948321 on Build GRCh38
rs139298661 Chr.4: 2948110-2948110 on Build GRCh38
rs1021596641 Chr.4: 2948352-2948352 on Build GRCh38
rs1037337667 Chr.4: 2947964-2947964 on Build GRCh38
rs867165981 Chr.4: 2948194-2948194 on Build GRCh38
rs559869902 Chr.4: 2948111-2948111 on Build GRCh38
rs144484316 Chr.4: 2948362-2948362 on Build GRCh38
rs531762783 Chr.4: 2948192-2948192 on Build GRCh38
rs893430464 Chr.4: 2948057-2948057 on Build GRCh38
rs1007326429 Chr.4: 2948145-2948145 on Build GRCh38

View Full Product Details