Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 3512131-3512637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRPAP1
Entrez Gene ID
4043
Gene Name
LDL receptor related protein associated protein 1
Gene Aliases
A2MRAP, A2RAP, HBP44, MRAP, MYP23, RAP, alpha-2-MRAP
UniGene
Hs.40966
Gene Chromosome Location
Chr.4: 3503597-3532497 on Build GRCh38
Transcripts
NM_002337.3 NR_110005.1

SNP Detail

SNP ID Chromosome Location
rs940898278 Chr.4: 3512334-3512334 on Build GRCh38
rs1040899265 Chr.4: 3512583-3512583 on Build GRCh38
rs556921435 Chr.4: 3512499-3512499 on Build GRCh38
rs1037527829 Chr.4: 3512345-3512345 on Build GRCh38
rs377047499 Chr.4: 3512515-3512515 on Build GRCh38
rs193226384 Chr.4: 3512160-3512160 on Build GRCh38
rs541657745 Chr.4: 3512207-3512207 on Build GRCh38
rs940963594 Chr.4: 3512517-3512517 on Build GRCh38
rs532747792 Chr.4: 3512273-3512273 on Build GRCh38
rs1020976127 Chr.4: 3512427-3512427 on Build GRCh38
rs1049352969 Chr.4: 3512547-3512547 on Build GRCh38
rs886273189 Chr.4: 3512155-3512155 on Build GRCh38
rs889535033 Chr.4: 3512579-3512579 on Build GRCh38
rs572107328 Chr.4: 3512542-3512542 on Build GRCh38
rs564121822 Chr.4: 3512264-3512264 on Build GRCh38
rs540935815 Chr.4: 3512226-3512226 on Build GRCh38
rs571617889 Chr.4: 3512469-3512469 on Build GRCh38
rs777941414 Chr.4: 3512474-3512474 on Build GRCh38
rs975904385 Chr.4: 3512498-3512498 on Build GRCh38
rs2077130 Chr.4: 3512502-3512502 on Build GRCh38
rs886244443 Chr.4: 3512331-3512331 on Build GRCh38
rs895462079 Chr.4: 3512596-3512596 on Build GRCh38
rs1032298733 Chr.4: 3512604-3512604 on Build GRCh38
rs116622990 Chr.4: 3512406-3512406 on Build GRCh38
rs978908901 Chr.4: 3512457-3512457 on Build GRCh38
rs754367437 Chr.4: 3512186-3512186 on Build GRCh38
rs185183975 Chr.4: 3512216-3512216 on Build GRCh38
rs967554168 Chr.4: 3512259-3512259 on Build GRCh38
rs1019564418 Chr.4: 3512253-3512253 on Build GRCh38
rs111558413 Chr.4: 3512174-3512174 on Build GRCh38
rs1794438 Chr.4: 3512510-3512510 on Build GRCh38
rs933212583 Chr.4: 3512323-3512323 on Build GRCh38
rs879534774 Chr.4: 3512428-3512428 on Build GRCh38
rs537087732 Chr.4: 3512487-3512487 on Build GRCh38
rs999349135 Chr.4: 3512599-3512599 on Build GRCh38
rs889526822 Chr.4: 3512403-3512403 on Build GRCh38
rs537704710 Chr.4: 3512177-3512177 on Build GRCh38
rs191853111 Chr.4: 3512500-3512500 on Build GRCh38
rs756569034 Chr.4: 3512439-3512439 on Build GRCh38
rs1014317156 Chr.4: 3512605-3512605 on Build GRCh38
rs1008371632 Chr.4: 3512580-3512580 on Build GRCh38
rs1001143990 Chr.4: 3512191-3512191 on Build GRCh38
rs730748 Chr.4: 3512359-3512359 on Build GRCh38
rs898136248 Chr.4: 3512353-3512353 on Build GRCh38
rs990933709 Chr.4: 3512274-3512274 on Build GRCh38
rs372804654 Chr.4: 3512175-3512175 on Build GRCh38
rs966997621 Chr.4: 3512257-3512257 on Build GRCh38
rs1036716169 Chr.4: 3512340-3512340 on Build GRCh38
rs949517389 Chr.4: 3512154-3512154 on Build GRCh38
rs1003917047 Chr.4: 3512173-3512173 on Build GRCh38
rs937060209 Chr.4: 3512568-3512568 on Build GRCh38
rs549625234 Chr.4: 3512372-3512372 on Build GRCh38
rs112750320 Chr.4: 3512209-3512209 on Build GRCh38
rs577923452 Chr.4: 3512158-3512158 on Build GRCh38
rs916699688 Chr.4: 3512497-3512497 on Build GRCh38
rs774549711 Chr.4: 3512538-3512538 on Build GRCh38
rs1019827607 Chr.4: 3512419-3512419 on Build GRCh38
rs1028069940 Chr.4: 3512379-3512379 on Build GRCh38
rs978120665 Chr.4: 3512261-3512261 on Build GRCh38
rs574695480 Chr.4: 3512432-3512432 on Build GRCh38
rs755587800 Chr.4: 3512210-3512210 on Build GRCh38
rs899008227 Chr.4: 3512564-3512564 on Build GRCh38
rs573787518 Chr.4: 3512187-3512187 on Build GRCh38
rs9995685 Chr.4: 3512589-3512589 on Build GRCh38
rs886699249 Chr.4: 3512329-3512329 on Build GRCh38
rs10024301 Chr.4: 3512433-3512433 on Build GRCh38
rs916648676 Chr.4: 3512308-3512308 on Build GRCh38
rs770989331 Chr.4: 3512523-3512523 on Build GRCh38
rs942788413 Chr.4: 3512311-3512311 on Build GRCh38
rs188505547 Chr.4: 3512321-3512321 on Build GRCh38
rs913367340 Chr.4: 3512246-3512246 on Build GRCh38
rs920844630 Chr.4: 3512536-3512536 on Build GRCh38
rs62272702 Chr.4: 3512322-3512322 on Build GRCh38
rs886687251 Chr.4: 3512161-3512161 on Build GRCh38

View Full Product Details