Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 117144132-117144676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VTCN1
Entrez Gene ID
79679
Gene Name
V-set domain containing T cell activation inhibitor 1
Gene Aliases
B7-H4, B7H4, B7S1, B7X, B7h.5, PRO1291, VCTN1
UniGene
Hs.546434
Gene Chromosome Location
Chr.1: 117143587-117210992 on Build GRCh38
Transcripts
NM_001253849.1 NM_001253850.1 NM_024626.3 NR_045603.1 NR_045604.1 XM_011542143.2 XM_017002335.1

SNP Detail

SNP ID Chromosome Location
rs549145866 Chr.1: 117144449-117144449 on Build GRCh38
rs554352072 Chr.1: 117144563-117144563 on Build GRCh38
rs972669183 Chr.1: 117144169-117144169 on Build GRCh38
rs1001277215 Chr.1: 117144381-117144381 on Build GRCh38
rs993986697 Chr.1: 117144619-117144619 on Build GRCh38
rs949692788 Chr.1: 117144369-117144369 on Build GRCh38
rs534002230 Chr.1: 117144518-117144518 on Build GRCh38
rs1018271134 Chr.1: 117144452-117144452 on Build GRCh38
rs560939379 Chr.1: 117144283-117144283 on Build GRCh38
rs758251859 Chr.1: 117144385-117144385 on Build GRCh38
rs112326285 Chr.1: 117144415-117144415 on Build GRCh38
rs780853657 Chr.1: 117144465-117144465 on Build GRCh38
rs551576201 Chr.1: 117144373-117144373 on Build GRCh38
rs574315224 Chr.1: 117144431-117144431 on Build GRCh38
rs1937956 Chr.1: 117144503-117144503 on Build GRCh38
rs187290138 Chr.1: 117144536-117144536 on Build GRCh38
rs149359126 Chr.1: 117144476-117144476 on Build GRCh38
rs562838452 Chr.1: 117144339-117144339 on Build GRCh38
rs925360901 Chr.1: 117144499-117144499 on Build GRCh38
rs986806798 Chr.1: 117144221-117144221 on Build GRCh38
rs539968124 Chr.1: 117144567-117144570 on Build GRCh38
rs765497987 Chr.1: 117144332-117144332 on Build GRCh38
rs115299292 Chr.1: 117144216-117144216 on Build GRCh38
rs1038085055 Chr.1: 117144602-117144602 on Build GRCh38
rs566399036 Chr.1: 117144403-117144403 on Build GRCh38
rs6664759 Chr.1: 117144292-117144292 on Build GRCh38
rs775359199 Chr.1: 117144326-117144326 on Build GRCh38
rs557660675 Chr.1: 117144230-117144242 on Build GRCh38
rs572347461 Chr.1: 117144589-117144589 on Build GRCh38
rs911282858 Chr.1: 117144226-117144226 on Build GRCh38
rs754814613 Chr.1: 117144451-117144451 on Build GRCh38
rs979536302 Chr.1: 117144474-117144474 on Build GRCh38
rs1025533610 Chr.1: 117144417-117144417 on Build GRCh38
rs755674279 Chr.1: 117144507-117144507 on Build GRCh38
rs1046951761 Chr.1: 117144627-117144627 on Build GRCh38
rs986778632 Chr.1: 117144450-117144450 on Build GRCh38
rs898246761 Chr.1: 117144603-117144603 on Build GRCh38
rs896747700 Chr.1: 117144368-117144368 on Build GRCh38
rs539444165 Chr.1: 117144372-117144372 on Build GRCh38
rs542571336 Chr.1: 117144261-117144261 on Build GRCh38
rs35182629 Chr.1: 117144392-117144393 on Build GRCh38
rs779672852 Chr.1: 117144411-117144411 on Build GRCh38
rs536346413 Chr.1: 117144637-117144637 on Build GRCh38
rs905559515 Chr.1: 117144378-117144379 on Build GRCh38
rs750587799 Chr.1: 117144338-117144338 on Build GRCh38
rs571927563 Chr.1: 117144535-117144535 on Build GRCh38
rs544082174 Chr.1: 117144302-117144302 on Build GRCh38
rs764920900 Chr.1: 117144581-117144583 on Build GRCh38
rs556545310 Chr.1: 117144514-117144514 on Build GRCh38
rs6428679 Chr.1: 117144205-117144205 on Build GRCh38

View Full Product Details