Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 118923332-118923857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX15
Entrez Gene ID
6913
Gene Name
T-box 15
Gene Aliases
TBX14
UniGene
Hs.146196
Gene Chromosome Location
Chr.1: 118883043-118989556 on Build GRCh38
Transcripts
NM_152380.2 XM_005271161.3 XM_005271162.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3849227 c.831C>A p.F277L Chr.1: 118923466-118923466 on Build GRCh38
3472153 c.497T>A p.F166Y Chr.1: 118923482-118923482 on Build GRCh38
4900585 c.784C>T p.P262S Chr.1: 118923513-118923513 on Build GRCh38
4900584 c.466C>T p.P156S Chr.1: 118923513-118923513 on Build GRCh38
3472155 c.435C>T p.F145F Chr.1: 118923544-118923544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140670116 Chr.1: 118923460-118923460 on Build GRCh38
rs771179864 Chr.1: 118923548-118923548 on Build GRCh38
rs761725095 Chr.1: 118923623-118923623 on Build GRCh38
rs763967043 Chr.1: 118923491-118923491 on Build GRCh38
rs775478038 Chr.1: 118923478-118923478 on Build GRCh38
rs748662087 Chr.1: 118923408-118923408 on Build GRCh38
rs778296371 Chr.1: 118923417-118923417 on Build GRCh38
rs983193881 Chr.1: 118923691-118923691 on Build GRCh38
rs892723950 Chr.1: 118923807-118923807 on Build GRCh38
rs145520486 Chr.1: 118923800-118923800 on Build GRCh38
rs769857016 Chr.1: 118923471-118923471 on Build GRCh38
rs780667920 Chr.1: 118923392-118923392 on Build GRCh38
rs772640867 Chr.1: 118923410-118923410 on Build GRCh38
rs889882160 Chr.1: 118923570-118923570 on Build GRCh38
rs534155441 Chr.1: 118923415-118923415 on Build GRCh38
rs867658460 Chr.1: 118923533-118923533 on Build GRCh38
rs758997437 Chr.1: 118923528-118923528 on Build GRCh38
rs753340888 Chr.1: 118923527-118923527 on Build GRCh38
rs761542851 Chr.1: 118923550-118923551 on Build GRCh38
rs199701328 Chr.1: 118923549-118923549 on Build GRCh38
rs776578922 Chr.1: 118923392-118923393 on Build GRCh38
rs749988915 Chr.1: 118923393-118923393 on Build GRCh38
rs192074581 Chr.1: 118923422-118923422 on Build GRCh38
rs770833571 Chr.1: 118923634-118923634 on Build GRCh38
rs371264178 Chr.1: 118923555-118923555 on Build GRCh38
rs756955286 Chr.1: 118923388-118923388 on Build GRCh38
rs543528579 Chr.1: 118923716-118923716 on Build GRCh38
rs775565793 Chr.1: 118923595-118923595 on Build GRCh38
rs189336136 Chr.1: 118923689-118923689 on Build GRCh38
rs145748292 Chr.1: 118923667-118923667 on Build GRCh38
rs1022643812 Chr.1: 118923663-118923663 on Build GRCh38
rs767213897 Chr.1: 118923631-118923631 on Build GRCh38
rs768619840 Chr.1: 118923611-118923611 on Build GRCh38
rs763877162 Chr.1: 118923385-118923385 on Build GRCh38
rs532278734 Chr.1: 118923357-118923357 on Build GRCh38
rs944163192 Chr.1: 118923759-118923759 on Build GRCh38
rs377226325 Chr.1: 118923647-118923647 on Build GRCh38
rs556249835 Chr.1: 118923521-118923521 on Build GRCh38
rs370064454 Chr.1: 118923430-118923430 on Build GRCh38
rs540708187 Chr.1: 118923792-118923792 on Build GRCh38
rs138218981 Chr.1: 118923569-118923569 on Build GRCh38
rs1766794 Chr.1: 118923816-118923816 on Build GRCh38
rs369246842 Chr.1: 118923404-118923404 on Build GRCh38
rs571821048 Chr.1: 118923366-118923366 on Build GRCh38
rs199611902 Chr.1: 118923545-118923545 on Build GRCh38
rs536026789 Chr.1: 118923386-118923386 on Build GRCh38
rs778105423 Chr.1: 118923535-118923535 on Build GRCh38
rs570607194 Chr.1: 118923695-118923695 on Build GRCh38
rs762887601 Chr.1: 118923486-118923486 on Build GRCh38
rs4659129 Chr.1: 118923652-118923652 on Build GRCh38
rs762029778 Chr.1: 118923381-118923381 on Build GRCh38
rs766610703 Chr.1: 118923763-118923763 on Build GRCh38
rs776689812 Chr.1: 118923425-118923425 on Build GRCh38
rs140463861 Chr.1: 118923505-118923505 on Build GRCh38
rs764713622 Chr.1: 118923648-118923648 on Build GRCh38
rs755662027 Chr.1: 118923397-118923397 on Build GRCh38
rs749326245 Chr.1: 118923610-118923610 on Build GRCh38
rs1039849347 Chr.1: 118923376-118923376 on Build GRCh38
rs781452293 Chr.1: 118923428-118923428 on Build GRCh38
rs556529627 Chr.1: 118923780-118923780 on Build GRCh38
rs184619664 Chr.1: 118923658-118923658 on Build GRCh38
rs559216526 Chr.1: 118923797-118923797 on Build GRCh38
rs577986490 Chr.1: 118923523-118923523 on Build GRCh38
rs199654302 Chr.1: 118923490-118923490 on Build GRCh38
rs961641788 Chr.1: 118923364-118923364 on Build GRCh38
rs914497854 Chr.1: 118923778-118923778 on Build GRCh38
rs565670285 Chr.1: 118923382-118923382 on Build GRCh38
rs1766793 Chr.1: 118923751-118923751 on Build GRCh38
rs377391020 Chr.1: 118923423-118923423 on Build GRCh38
rs766081546 Chr.1: 118923637-118923637 on Build GRCh38
rs374500071 Chr.1: 118923519-118923519 on Build GRCh38
rs769942045 Chr.1: 118923584-118923584 on Build GRCh38
rs576174909 Chr.1: 118923694-118923694 on Build GRCh38
rs993151027 Chr.1: 118923365-118923365 on Build GRCh38
rs374171713 Chr.1: 118923635-118923635 on Build GRCh38
rs774132238 Chr.1: 118923622-118923622 on Build GRCh38
rs73007559 Chr.1: 118923776-118923776 on Build GRCh38

View Full Product Details