Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 6909465-6909965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D14
Entrez Gene ID
57533
Gene Name
TBC1 domain family member 14
Gene Aliases
-
UniGene
Hs.518611
Gene Chromosome Location
Chr.4: 6909444-7033118 on Build GRCh38
Transcripts
NM_001113361.1 NM_020773.2 XM_005247986.2 XM_011513507.2 XM_011513508.1 XM_017008477.1

SNP Detail

SNP ID Chromosome Location
rs181057199 Chr.4: 6909705-6909705 on Build GRCh38
rs998611630 Chr.4: 6909844-6909844 on Build GRCh38
rs945713699 Chr.4: 6909551-6909551 on Build GRCh38
rs902138090 Chr.4: 6909631-6909631 on Build GRCh38
rs953173546 Chr.4: 6909514-6909514 on Build GRCh38
rs928275481 Chr.4: 6909694-6909694 on Build GRCh38
rs953557073 Chr.4: 6909648-6909648 on Build GRCh38
rs972132776 Chr.4: 6909917-6909917 on Build GRCh38
rs543504973 Chr.4: 6909535-6909535 on Build GRCh38
rs1035399107 Chr.4: 6909848-6909848 on Build GRCh38
rs372819697 Chr.4: 6909753-6909753 on Build GRCh38
rs915797565 Chr.4: 6909742-6909742 on Build GRCh38
rs1036352960 Chr.4: 6909608-6909608 on Build GRCh38
rs564973791 Chr.4: 6909557-6909557 on Build GRCh38
rs914295372 Chr.4: 6909530-6909530 on Build GRCh38
rs1048207437 Chr.4: 6909754-6909754 on Build GRCh38
rs570602991 Chr.4: 6909788-6909788 on Build GRCh38
rs928565337 Chr.4: 6909566-6909566 on Build GRCh38
rs1011719609 Chr.4: 6909652-6909652 on Build GRCh38
rs901855040 Chr.4: 6909798-6909798 on Build GRCh38
rs185242582 Chr.4: 6909831-6909831 on Build GRCh38
rs780539981 Chr.4: 6909492-6909492 on Build GRCh38
rs997722187 Chr.4: 6909639-6909639 on Build GRCh38
rs529923499 Chr.4: 6909668-6909668 on Build GRCh38
rs546859471 Chr.4: 6909860-6909860 on Build GRCh38
rs915854843 Chr.4: 6909870-6909870 on Build GRCh38
rs866612633 Chr.4: 6909761-6909761 on Build GRCh38
rs886989767 Chr.4: 6909537-6909537 on Build GRCh38
rs1050651785 Chr.4: 6909821-6909821 on Build GRCh38
rs910680041 Chr.4: 6909757-6909768 on Build GRCh38
rs568663566 Chr.4: 6909869-6909869 on Build GRCh38
rs997441019 Chr.4: 6909801-6909801 on Build GRCh38
rs552202183 Chr.4: 6909781-6909781 on Build GRCh38
rs563499437 Chr.4: 6909709-6909709 on Build GRCh38
rs1021808563 Chr.4: 6909654-6909654 on Build GRCh38
rs1026943906 Chr.4: 6909527-6909527 on Build GRCh38
rs1029111807 Chr.4: 6909509-6909509 on Build GRCh38
rs1050366389 Chr.4: 6909938-6909938 on Build GRCh38
rs559530730 Chr.4: 6909649-6909649 on Build GRCh38
rs967133651 Chr.4: 6909682-6909682 on Build GRCh38
rs541214912 Chr.4: 6909596-6909596 on Build GRCh38
rs939812213 Chr.4: 6909913-6909913 on Build GRCh38
rs989863857 Chr.4: 6909523-6909523 on Build GRCh38
rs959702132 Chr.4: 6909850-6909850 on Build GRCh38
rs966249521 Chr.4: 6909495-6909495 on Build GRCh38
rs573904060 Chr.4: 6909757-6909758 on Build GRCh38
rs535742051 Chr.4: 6909925-6909925 on Build GRCh38
rs1012180155 Chr.4: 6909836-6909836 on Build GRCh38
rs1005316933 Chr.4: 6909598-6909598 on Build GRCh38
rs1049619680 Chr.4: 6909593-6909593 on Build GRCh38
rs977518352 Chr.4: 6909693-6909693 on Build GRCh38
rs952646938 Chr.4: 6909889-6909889 on Build GRCh38
rs991406540 Chr.4: 6909714-6909714 on Build GRCh38
rs889391998 Chr.4: 6909943-6909944 on Build GRCh38
rs535486289 Chr.4: 6909597-6909598 on Build GRCh38
rs995154380 Chr.4: 6909518-6909518 on Build GRCh38
rs915743694 Chr.4: 6909587-6909587 on Build GRCh38
rs1036811964 Chr.4: 6909791-6909791 on Build GRCh38
rs938565485 Chr.4: 6909583-6909583 on Build GRCh38
rs908369516 Chr.4: 6909909-6909909 on Build GRCh38
rs984396616 Chr.4: 6909907-6909907 on Build GRCh38
rs577057070 Chr.4: 6909589-6909589 on Build GRCh38
rs1029162750 Chr.4: 6909647-6909647 on Build GRCh38
rs1021780256 Chr.4: 6909841-6909841 on Build GRCh38
rs976251496 Chr.4: 6909501-6909501 on Build GRCh38
rs530817786 Chr.4: 6909743-6909743 on Build GRCh38
rs190142339 Chr.4: 6909933-6909933 on Build GRCh38

View Full Product Details