Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7433027-7433530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSAPL1
Entrez Gene ID
768239
Gene Name
prosaposin-like 1 (gene/pseudogene)
Gene Aliases
-
UniGene
Hs.662523
Gene Chromosome Location
Chr.4: 7430294-7434973 on Build GRCh38
Transcripts
NM_001085382.1 NM_020777.2 XM_005247987.4 XM_011513514.2 XM_011513515.2 XM_011513516.2 XM_017008481.1

Gene Symbol
SORCS2
Entrez Gene ID
57537
Gene Name
sortilin related VPS10 domain containing receptor 2
Gene Aliases
-
UniGene
Hs.479099
Gene Chromosome Location
Chr.4: 7192607-7742837 on Build GRCh38
Transcripts
NM_001085382.1 NM_020777.2 XM_005247987.4 XM_011513514.2 XM_011513515.2 XM_011513516.2 XM_017008481.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7385841 c.1513T>A p.C505S Chr.4: 7433367-7433367 on Build GRCh38
6757360 c.596G>T p.G199V Chr.4: 7433480-7433480 on Build GRCh38
4405078 c.590A>G p.K197R Chr.4: 7433486-7433486 on Build GRCh38
5401518 c.1545C>T p.L515L Chr.4: 7433335-7433335 on Build GRCh38
5401520 c.1479G>A p.Q493Q Chr.4: 7433401-7433401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932429382 Chr.4: 7433458-7433458 on Build GRCh38
rs756186433 Chr.4: 7433352-7433352 on Build GRCh38
rs11721763 Chr.4: 7433286-7433286 on Build GRCh38
rs775026644 Chr.4: 7433403-7433403 on Build GRCh38
rs758855606 Chr.4: 7433507-7433507 on Build GRCh38
rs1056786507 Chr.4: 7433319-7433319 on Build GRCh38
rs561400494 Chr.4: 7433394-7433394 on Build GRCh38
rs866509461 Chr.4: 7433289-7433289 on Build GRCh38
rs763898194 Chr.4: 7433328-7433328 on Build GRCh38
rs539506664 Chr.4: 7433202-7433202 on Build GRCh38
rs756647155 Chr.4: 7433066-7433066 on Build GRCh38
rs764586181 Chr.4: 7433405-7433405 on Build GRCh38
rs545052742 Chr.4: 7433311-7433311 on Build GRCh38
rs752804073 Chr.4: 7433506-7433506 on Build GRCh38
rs866901018 Chr.4: 7433396-7433396 on Build GRCh38
rs1009422060 Chr.4: 7433423-7433423 on Build GRCh38
rs978837221 Chr.4: 7433080-7433080 on Build GRCh38
rs114025050 Chr.4: 7433109-7433109 on Build GRCh38
rs759095733 Chr.4: 7433317-7433317 on Build GRCh38
rs931101091 Chr.4: 7433295-7433295 on Build GRCh38
rs59409649 Chr.4: 7433282-7433282 on Build GRCh38
rs778982266 Chr.4: 7433447-7433447 on Build GRCh38
rs779261879 Chr.4: 7433503-7433503 on Build GRCh38
rs755270307 Chr.4: 7433446-7433446 on Build GRCh38
rs183139546 Chr.4: 7433264-7433264 on Build GRCh38
rs55987441 Chr.4: 7433452-7433452 on Build GRCh38
rs775671977 Chr.4: 7433280-7433280 on Build GRCh38
rs549335032 Chr.4: 7433355-7433355 on Build GRCh38
rs12498567 Chr.4: 7433467-7433467 on Build GRCh38
rs566624975 Chr.4: 7433246-7433246 on Build GRCh38
rs886867254 Chr.4: 7433164-7433164 on Build GRCh38
rs987463080 Chr.4: 7433269-7433269 on Build GRCh38
rs200029498 Chr.4: 7433395-7433395 on Build GRCh38
rs190241190 Chr.4: 7433199-7433199 on Build GRCh38
rs767506546 Chr.4: 7433428-7433428 on Build GRCh38
rs7439798 Chr.4: 7433053-7433053 on Build GRCh38
rs760649460 Chr.4: 7433299-7433299 on Build GRCh38
rs371858136 Chr.4: 7433443-7433443 on Build GRCh38
rs999012278 Chr.4: 7433332-7433332 on Build GRCh38
rs756203005 Chr.4: 7433442-7433442 on Build GRCh38
rs752331312 Chr.4: 7433322-7433322 on Build GRCh38
rs1057219140 Chr.4: 7433181-7433181 on Build GRCh38
rs931048792 Chr.4: 7433143-7433143 on Build GRCh38
rs767492629 Chr.4: 7433503-7433503 on Build GRCh38
rs73212523 Chr.4: 7433427-7433427 on Build GRCh38
rs527980462 Chr.4: 7433101-7433101 on Build GRCh38
rs1011814478 Chr.4: 7433108-7433108 on Build GRCh38
rs750384884 Chr.4: 7433480-7433480 on Build GRCh38
rs35116829 Chr.4: 7433258-7433259 on Build GRCh38
rs768409521 Chr.4: 7433474-7433474 on Build GRCh38
rs747468581 Chr.4: 7433464-7433464 on Build GRCh38
rs763620606 Chr.4: 7433411-7433411 on Build GRCh38
rs961521740 Chr.4: 7433413-7433413 on Build GRCh38
rs893474870 Chr.4: 7433313-7433313 on Build GRCh38
rs754000468 Chr.4: 7433495-7433495 on Build GRCh38
rs373713228 Chr.4: 7433374-7433374 on Build GRCh38
rs754999685 Chr.4: 7433498-7433498 on Build GRCh38
rs528093153 Chr.4: 7433163-7433163 on Build GRCh38
rs561774379 Chr.4: 7433195-7433195 on Build GRCh38
rs374876500 Chr.4: 7433398-7433398 on Build GRCh38
rs748605255 Chr.4: 7433449-7433449 on Build GRCh38
rs754863265 Chr.4: 7433366-7433375 on Build GRCh38
rs1013614219 Chr.4: 7433215-7433215 on Build GRCh38
rs1040789254 Chr.4: 7433079-7433079 on Build GRCh38
rs568088203 Chr.4: 7433124-7433124 on Build GRCh38
rs377581924 Chr.4: 7433290-7433290 on Build GRCh38
rs1057251961 Chr.4: 7433488-7433488 on Build GRCh38
rs987881110 Chr.4: 7433100-7433100 on Build GRCh38
rs200787392 Chr.4: 7433451-7433451 on Build GRCh38
rs991422283 Chr.4: 7433225-7433225 on Build GRCh38
rs562816477 Chr.4: 7433058-7433058 on Build GRCh38
rs371617705 Chr.4: 7433361-7433361 on Build GRCh38
rs750298624 Chr.4: 7433429-7433429 on Build GRCh38
rs757029999 Chr.4: 7433419-7433419 on Build GRCh38
rs921090710 Chr.4: 7433287-7433287 on Build GRCh38
rs952653133 Chr.4: 7433252-7433252 on Build GRCh38
rs781081903 Chr.4: 7433335-7433335 on Build GRCh38
rs533605750 Chr.4: 7433060-7433060 on Build GRCh38
rs376324046 Chr.4: 7433081-7433081 on Build GRCh38
rs750103765 Chr.4: 7433407-7433407 on Build GRCh38
rs147175966 Chr.4: 7433226-7433226 on Build GRCh38
rs1012934672 Chr.4: 7433356-7433356 on Build GRCh38
rs749506695 Chr.4: 7433444-7433444 on Build GRCh38
rs866397682 Chr.4: 7433166-7433166 on Build GRCh38
rs78445843 Chr.4: 7433266-7433266 on Build GRCh38
rs766198360 Chr.4: 7433494-7433494 on Build GRCh38
rs762344889 Chr.4: 7433470-7433470 on Build GRCh38
rs1039547209 Chr.4: 7433137-7433137 on Build GRCh38
rs968760350 Chr.4: 7433070-7433070 on Build GRCh38
rs908262123 Chr.4: 7433315-7433315 on Build GRCh38
rs985200510 Chr.4: 7433459-7433459 on Build GRCh38
rs761604402 Chr.4: 7433477-7433477 on Build GRCh38
rs551571989 Chr.4: 7433084-7433084 on Build GRCh38
rs1003843317 Chr.4: 7433179-7433179 on Build GRCh38
rs540184948 Chr.4: 7433194-7433194 on Build GRCh38
rs749471633 Chr.4: 7433359-7433359 on Build GRCh38
rs560283433 Chr.4: 7433096-7433096 on Build GRCh38
rs751416918 Chr.4: 7433414-7433414 on Build GRCh38
rs916912094 Chr.4: 7433500-7433500 on Build GRCh38
rs4318651 Chr.4: 7433261-7433261 on Build GRCh38
rs968894660 Chr.4: 7433243-7433243 on Build GRCh38
rs200338688 Chr.4: 7433469-7433469 on Build GRCh38
rs368073106 Chr.4: 7433425-7433425 on Build GRCh38
rs367780758 Chr.4: 7433384-7433384 on Build GRCh38
rs375187579 Chr.4: 7433330-7433330 on Build GRCh38
rs746001292 Chr.4: 7433342-7433342 on Build GRCh38
rs78599329 Chr.4: 7433283-7433283 on Build GRCh38
rs767245257 Chr.4: 7433478-7433478 on Build GRCh38
rs757005290 Chr.4: 7433320-7433320 on Build GRCh38
rs1044660420 Chr.4: 7433350-7433350 on Build GRCh38
rs1023067931 Chr.4: 7433229-7433229 on Build GRCh38
rs889342285 Chr.4: 7433106-7433106 on Build GRCh38
rs771521999 Chr.4: 7433485-7433485 on Build GRCh38
rs60816800 Chr.4: 7433331-7433331 on Build GRCh38
rs921123057 Chr.4: 7433142-7433142 on Build GRCh38
rs915897871 Chr.4: 7433234-7433234 on Build GRCh38
rs760567963 Chr.4: 7433492-7433492 on Build GRCh38
rs186026629 Chr.4: 7433318-7433318 on Build GRCh38
rs186517170 Chr.4: 7433169-7433169 on Build GRCh38
rs748430817 Chr.4: 7433379-7433379 on Build GRCh38
rs758140872 Chr.4: 7433325-7433325 on Build GRCh38

View Full Product Details