Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15442086-15442587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF7
Entrez Gene ID
114905
Gene Name
C1q and tumor necrosis factor related protein 7
Gene Aliases
CTRP7, ZACRP7
UniGene
Hs.153714
Gene Chromosome Location
Chr.4: 15339818-15446169 on Build GRCh38
Transcripts
NM_001135170.1 NM_001135171.1 NM_031911.4 XM_011513772.1 XM_011513773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5400647 c.464C>T p.T155I Chr.4: 15442393-15442393 on Build GRCh38
3726436 c.317T>A p.I106K Chr.4: 15442246-15442246 on Build GRCh38
5466073 c.241T>G p.L81V Chr.4: 15442170-15442170 on Build GRCh38
382322 c.378C>T p.D126D Chr.4: 15442307-15442307 on Build GRCh38
4931483 c.628C>A p.L210M Chr.4: 15442557-15442557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148159746 Chr.4: 15442188-15442188 on Build GRCh38
rs770057901 Chr.4: 15442227-15442227 on Build GRCh38
rs753537535 Chr.4: 15442189-15442189 on Build GRCh38
rs149975018 Chr.4: 15442384-15442384 on Build GRCh38
rs778294904 Chr.4: 15442436-15442436 on Build GRCh38
rs772582352 Chr.4: 15442331-15442331 on Build GRCh38
rs1055750154 Chr.4: 15442348-15442348 on Build GRCh38
rs768433452 Chr.4: 15442454-15442454 on Build GRCh38
rs773571400 Chr.4: 15442266-15442266 on Build GRCh38
rs372857232 Chr.4: 15442374-15442374 on Build GRCh38
rs780351928 Chr.4: 15442324-15442324 on Build GRCh38
rs779936437 Chr.4: 15442235-15442235 on Build GRCh38
rs781063980 Chr.4: 15442310-15442310 on Build GRCh38
rs146738721 Chr.4: 15442530-15442530 on Build GRCh38
rs199665810 Chr.4: 15442318-15442318 on Build GRCh38
rs201300459 Chr.4: 15442203-15442203 on Build GRCh38
rs376919397 Chr.4: 15442118-15442118 on Build GRCh38
rs773998639 Chr.4: 15442365-15442365 on Build GRCh38
rs747434592 Chr.4: 15442350-15442350 on Build GRCh38
rs148678610 Chr.4: 15442448-15442448 on Build GRCh38
rs556312158 Chr.4: 15442373-15442373 on Build GRCh38
rs376881645 Chr.4: 15442553-15442553 on Build GRCh38
rs755160883 Chr.4: 15442390-15442390 on Build GRCh38
rs773946027 Chr.4: 15442458-15442458 on Build GRCh38
rs201630756 Chr.4: 15442327-15442327 on Build GRCh38
rs769303337 Chr.4: 15442358-15442358 on Build GRCh38
rs374455303 Chr.4: 15442383-15442383 on Build GRCh38
rs748250423 Chr.4: 15442416-15442416 on Build GRCh38
rs202184168 Chr.4: 15442498-15442498 on Build GRCh38
rs147288877 Chr.4: 15442202-15442202 on Build GRCh38
rs763919521 Chr.4: 15442295-15442295 on Build GRCh38
rs763723180 Chr.4: 15442362-15442362 on Build GRCh38
rs758865104 Chr.4: 15442434-15442434 on Build GRCh38
rs746826403 Chr.4: 15442123-15442123 on Build GRCh38
rs772492592 Chr.4: 15442257-15442257 on Build GRCh38
rs1000706490 Chr.4: 15442426-15442426 on Build GRCh38
rs767529616 Chr.4: 15442465-15442465 on Build GRCh38
rs370149199 Chr.4: 15442242-15442242 on Build GRCh38
rs757308711 Chr.4: 15442451-15442451 on Build GRCh38
rs751608699 Chr.4: 15442303-15442303 on Build GRCh38
rs878999699 Chr.4: 15442447-15442447 on Build GRCh38
rs757110214 Chr.4: 15442309-15442309 on Build GRCh38
rs781548301 Chr.4: 15442210-15442210 on Build GRCh38
rs771240329 Chr.4: 15442131-15442131 on Build GRCh38
rs145116504 Chr.4: 15442400-15442400 on Build GRCh38
rs751372843 Chr.4: 15442206-15442206 on Build GRCh38
rs775265752 Chr.4: 15442360-15442360 on Build GRCh38
rs762567030 Chr.4: 15442284-15442284 on Build GRCh38
rs1020404919 Chr.4: 15442168-15442168 on Build GRCh38
rs200575694 Chr.4: 15442215-15442215 on Build GRCh38
rs909727073 Chr.4: 15442335-15442335 on Build GRCh38
rs1025735536 Chr.4: 15442504-15442504 on Build GRCh38
rs774529787 Chr.4: 15442176-15442176 on Build GRCh38
rs866407188 Chr.4: 15442201-15442201 on Build GRCh38
rs150846082 Chr.4: 15442328-15442328 on Build GRCh38
rs140339349 Chr.4: 15442557-15442557 on Build GRCh38
rs748587603 Chr.4: 15442330-15442330 on Build GRCh38
rs558870508 Chr.4: 15442243-15442243 on Build GRCh38
rs144010767 Chr.4: 15442190-15442190 on Build GRCh38
rs371471274 Chr.4: 15442559-15442559 on Build GRCh38
rs766263088 Chr.4: 15442502-15442502 on Build GRCh38
rs777038270 Chr.4: 15442143-15442143 on Build GRCh38
rs761781512 Chr.4: 15442369-15442369 on Build GRCh38
rs986356376 Chr.4: 15442244-15442244 on Build GRCh38
rs139272872 Chr.4: 15442340-15442340 on Build GRCh38
rs762195251 Chr.4: 15442181-15442181 on Build GRCh38
rs769216955 Chr.4: 15442167-15442167 on Build GRCh38
rs763300966 Chr.4: 15442161-15442161 on Build GRCh38
rs140824961 Chr.4: 15442281-15442281 on Build GRCh38
rs535929913 Chr.4: 15442361-15442361 on Build GRCh38
rs753791909 Chr.4: 15442441-15442441 on Build GRCh38
rs139379821 Chr.4: 15442453-15442453 on Build GRCh38
rs951057366 Chr.4: 15442511-15442511 on Build GRCh38
rs745924186 Chr.4: 15442150-15442150 on Build GRCh38
rs201187280 Chr.4: 15442315-15442315 on Build GRCh38
rs776056725 Chr.4: 15442154-15442154 on Build GRCh38
rs143385914 Chr.4: 15442463-15442463 on Build GRCh38
rs764586101 Chr.4: 15442193-15442193 on Build GRCh38
rs563271263 Chr.4: 15442449-15442449 on Build GRCh38
rs749406163 Chr.4: 15442239-15442239 on Build GRCh38
rs753589539 Chr.4: 15442525-15442525 on Build GRCh38
rs764008758 Chr.4: 15442419-15442420 on Build GRCh38
rs867027754 Chr.4: 15442404-15442404 on Build GRCh38
rs377604898 Chr.4: 15442280-15442280 on Build GRCh38
rs752111335 Chr.4: 15442282-15442282 on Build GRCh38
rs368300386 Chr.4: 15442466-15442466 on Build GRCh38
rs754147606 Chr.4: 15442389-15442389 on Build GRCh38
rs775005930 Chr.4: 15442450-15442450 on Build GRCh38
rs757171493 Chr.4: 15442209-15442209 on Build GRCh38
rs760373283 Chr.4: 15442501-15442501 on Build GRCh38
rs936745040 Chr.4: 15442272-15442272 on Build GRCh38
rs752265309 Chr.4: 15442197-15442197 on Build GRCh38

View Full Product Details