Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 17621476-17622011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED28
Entrez Gene ID
80306
Gene Name
mediator complex subunit 28
Gene Aliases
1500003D12Rik, EG1, magicin
UniGene
Hs.731966
Gene Chromosome Location
Chr.4: 17614628-17625628 on Build GRCh38
Transcripts
NM_025205.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7334643 c.247A>G p.K83E Chr.4: 17621607-17621607 on Build GRCh38
6469421 c.232G>T p.D78Y Chr.4: 17621592-17621592 on Build GRCh38
6529191 c.337G>C p.E113Q Chr.4: 17621697-17621697 on Build GRCh38
1053360 c.311T>C p.V104A Chr.4: 17621671-17621671 on Build GRCh38
733069 c.300G>T p.L100F Chr.4: 17621660-17621660 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776037740 Chr.4: 17621726-17621726 on Build GRCh38
rs549327734 Chr.4: 17621908-17621908 on Build GRCh38
rs768902937 Chr.4: 17621735-17621735 on Build GRCh38
rs980018832 Chr.4: 17621826-17621826 on Build GRCh38
rs368346412 Chr.4: 17621740-17621740 on Build GRCh38
rs780809258 Chr.4: 17621896-17621896 on Build GRCh38
rs1015319454 Chr.4: 17621768-17621768 on Build GRCh38
rs185810313 Chr.4: 17621764-17621764 on Build GRCh38
rs747212128 Chr.4: 17621690-17621690 on Build GRCh38
rs544924580 Chr.4: 17621778-17621778 on Build GRCh38
rs942042375 Chr.4: 17621971-17621971 on Build GRCh38
rs764510096 Chr.4: 17621616-17621616 on Build GRCh38
rs770247062 Chr.4: 17621561-17621561 on Build GRCh38
rs770728690 Chr.4: 17621893-17621893 on Build GRCh38
rs757102666 Chr.4: 17621746-17621746 on Build GRCh38
rs771135516 Chr.4: 17621697-17621697 on Build GRCh38
rs111645923 Chr.4: 17621718-17621718 on Build GRCh38
rs753346818 Chr.4: 17621650-17621650 on Build GRCh38
rs1044605134 Chr.4: 17621544-17621544 on Build GRCh38
rs764302492 Chr.4: 17621702-17621703 on Build GRCh38
rs913983375 Chr.4: 17621905-17621905 on Build GRCh38
rs924974078 Chr.4: 17621506-17621506 on Build GRCh38
rs929460243 Chr.4: 17621946-17621946 on Build GRCh38
rs752658347 Chr.4: 17621657-17621657 on Build GRCh38
rs373916760 Chr.4: 17621545-17621545 on Build GRCh38
rs760950057 Chr.4: 17621625-17621628 on Build GRCh38
rs966937129 Chr.4: 17621793-17621793 on Build GRCh38
rs752709409 Chr.4: 17621538-17621538 on Build GRCh38
rs191014499 Chr.4: 17621930-17621930 on Build GRCh38
rs368121116 Chr.4: 17621553-17621553 on Build GRCh38
rs145997779 Chr.4: 17621557-17621557 on Build GRCh38
rs925781074 Chr.4: 17621842-17621842 on Build GRCh38
rs759502263 Chr.4: 17621574-17621575 on Build GRCh38
rs34524628 Chr.4: 17621976-17621976 on Build GRCh38
rs935801235 Chr.4: 17621860-17621860 on Build GRCh38
rs1024848735 Chr.4: 17621792-17621792 on Build GRCh38
rs143222695 Chr.4: 17621721-17621721 on Build GRCh38
rs537705655 Chr.4: 17621949-17621949 on Build GRCh38
rs910966087 Chr.4: 17621889-17621889 on Build GRCh38
rs765816376 Chr.4: 17621646-17621646 on Build GRCh38
rs113177238 Chr.4: 17621751-17621751 on Build GRCh38
rs985091636 Chr.4: 17621888-17621888 on Build GRCh38
rs779034152 Chr.4: 17621654-17621654 on Build GRCh38
rs566180830 Chr.4: 17621666-17621666 on Build GRCh38
rs993747424 Chr.4: 17621598-17621598 on Build GRCh38
rs758184008 Chr.4: 17621674-17621674 on Build GRCh38
rs1047987561 Chr.4: 17621948-17621948 on Build GRCh38
rs767629499 Chr.4: 17621575-17621575 on Build GRCh38
rs1055092785 Chr.4: 17621976-17621976 on Build GRCh38
rs781161880 Chr.4: 17621711-17621711 on Build GRCh38
rs763360735 Chr.4: 17621599-17621599 on Build GRCh38
rs745839785 Chr.4: 17621900-17621900 on Build GRCh38
rs1041009285 Chr.4: 17621910-17621910 on Build GRCh38
rs370772273 Chr.4: 17621556-17621556 on Build GRCh38
rs976903328 Chr.4: 17621913-17621913 on Build GRCh38
rs1049528246 Chr.4: 17621531-17621531 on Build GRCh38
rs936307500 Chr.4: 17621525-17621525 on Build GRCh38
rs113614767 Chr.4: 17621716-17621716 on Build GRCh38
rs777559082 Chr.4: 17621689-17621689 on Build GRCh38
rs879072605 Chr.4: 17621564-17621564 on Build GRCh38
rs753523497 Chr.4: 17621709-17621710 on Build GRCh38
rs754996075 Chr.4: 17621652-17621652 on Build GRCh38
rs957706487 Chr.4: 17621845-17621845 on Build GRCh38
rs529286431 Chr.4: 17621906-17621906 on Build GRCh38
rs560568598 Chr.4: 17621783-17621783 on Build GRCh38
rs377649308 Chr.4: 17621728-17621728 on Build GRCh38
rs527517388 Chr.4: 17621575-17621576 on Build GRCh38
rs771961556 Chr.4: 17621542-17621542 on Build GRCh38
rs774952361 Chr.4: 17621553-17621553 on Build GRCh38
rs989556661 Chr.4: 17621861-17621861 on Build GRCh38
rs775949842 Chr.4: 17621593-17621593 on Build GRCh38
rs747240333 Chr.4: 17621550-17621550 on Build GRCh38
rs902006554 Chr.4: 17621573-17621573 on Build GRCh38
rs754316365 Chr.4: 17621799-17621799 on Build GRCh38
rs564815002 Chr.4: 17621862-17621862 on Build GRCh38
rs1032241838 Chr.4: 17621825-17621825 on Build GRCh38
rs370698867 Chr.4: 17621632-17621632 on Build GRCh38
rs750106365 Chr.4: 17621628-17621628 on Build GRCh38

View Full Product Details