Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 25268363-25268877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PI4K2B
Entrez Gene ID
55300
Gene Name
phosphatidylinositol 4-kinase type 2 beta
Gene Aliases
PI4KIIB, PIK42B
UniGene
Hs.191701
Gene Chromosome Location
Chr.4: 25234031-25279209 on Build GRCh38
Transcripts
NM_018323.3 XM_005248174.1 XM_005248175.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
117835 c.1143G>C p.L381F Chr.4: 25268507-25268507 on Build GRCh38
6745755 c.1143G>A p.L381L Chr.4: 25268507-25268507 on Build GRCh38
1054648 c.1108G>A p.A370T Chr.4: 25268472-25268472 on Build GRCh38
3603071 c.1091G>A p.W364* Chr.4: 25268455-25268455 on Build GRCh38
3940855 c.1184T>G p.L395* Chr.4: 25268548-25268548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571665999 Chr.4: 25268438-25268438 on Build GRCh38
rs772528277 Chr.4: 25268469-25268469 on Build GRCh38
rs779104326 Chr.4: 25268403-25268403 on Build GRCh38
rs373464866 Chr.4: 25268500-25268500 on Build GRCh38
rs150275485 Chr.4: 25268601-25268601 on Build GRCh38
rs538688973 Chr.4: 25268812-25268812 on Build GRCh38
rs755925769 Chr.4: 25268551-25268551 on Build GRCh38
rs774122949 Chr.4: 25268462-25268462 on Build GRCh38
rs553511251 Chr.4: 25268752-25268752 on Build GRCh38
rs1036630615 Chr.4: 25268698-25268698 on Build GRCh38
rs141398591 Chr.4: 25268412-25268412 on Build GRCh38
rs768247821 Chr.4: 25268575-25268575 on Build GRCh38
rs377424277 Chr.4: 25268513-25268513 on Build GRCh38
rs530145848 Chr.4: 25268429-25268430 on Build GRCh38
rs763646802 Chr.4: 25268725-25268725 on Build GRCh38
rs761698695 Chr.4: 25268604-25268604 on Build GRCh38
rs762564889 Chr.4: 25268409-25268410 on Build GRCh38
rs537395852 Chr.4: 25268511-25268511 on Build GRCh38
rs923620368 Chr.4: 25268396-25268396 on Build GRCh38
rs995199919 Chr.4: 25268716-25268716 on Build GRCh38
rs3115232 Chr.4: 25268389-25268389 on Build GRCh38
rs185281971 Chr.4: 25268404-25268404 on Build GRCh38
rs145883883 Chr.4: 25268683-25268683 on Build GRCh38
rs568386698 Chr.4: 25268411-25268411 on Build GRCh38
rs771770864 Chr.4: 25268603-25268603 on Build GRCh38
rs761033072 Chr.4: 25268478-25268478 on Build GRCh38
rs369722569 Chr.4: 25268464-25268464 on Build GRCh38
rs899081124 Chr.4: 25268707-25268707 on Build GRCh38
rs149385911 Chr.4: 25268474-25268474 on Build GRCh38
rs956837076 Chr.4: 25268436-25268436 on Build GRCh38
rs201675363 Chr.4: 25268616-25268616 on Build GRCh38
rs749285941 Chr.4: 25268568-25268568 on Build GRCh38
rs1012481756 Chr.4: 25268724-25268724 on Build GRCh38
rs780023892 Chr.4: 25268563-25268563 on Build GRCh38
rs778599082 Chr.4: 25268595-25268595 on Build GRCh38
rs908649731 Chr.4: 25268646-25268646 on Build GRCh38
rs1018675271 Chr.4: 25268843-25268843 on Build GRCh38
rs768335792 Chr.4: 25268446-25268446 on Build GRCh38
rs757013658 Chr.4: 25268409-25268409 on Build GRCh38
rs776108858 Chr.4: 25268630-25268630 on Build GRCh38
rs759806533 Chr.4: 25268481-25268481 on Build GRCh38
rs771261541 Chr.4: 25268625-25268625 on Build GRCh38
rs982038934 Chr.4: 25268845-25268845 on Build GRCh38
rs572304003 Chr.4: 25268827-25268827 on Build GRCh38
rs750236546 Chr.4: 25268547-25268547 on Build GRCh38
rs376801365 Chr.4: 25268439-25268439 on Build GRCh38
rs761099995 Chr.4: 25268617-25268617 on Build GRCh38
rs766914799 Chr.4: 25268486-25268486 on Build GRCh38
rs769784052 Chr.4: 25268437-25268437 on Build GRCh38
rs780792498 Chr.4: 25268536-25268536 on Build GRCh38
rs34963419 Chr.4: 25268631-25268632 on Build GRCh38
rs755127634 Chr.4: 25268402-25268402 on Build GRCh38
rs144713410 Chr.4: 25268516-25268516 on Build GRCh38
rs758640377 Chr.4: 25268523-25268523 on Build GRCh38

View Full Product Details