Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 25383179-25383580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC4
Entrez Gene ID
29945
Gene Name
anaphase promoting complex subunit 4
Gene Aliases
APC4
UniGene
Hs.152173
Gene Chromosome Location
Chr.4: 25377226-25418498 on Build GRCh38
Transcripts
NM_001286756.1 NM_013367.2 XM_011513838.1 XM_011513839.1 XM_017008138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1054666 c.349G>A p.E117K Chr.4: 25383374-25383374 on Build GRCh38
6545312 c.333C>T p.S111S Chr.4: 25383358-25383358 on Build GRCh38
1319344 c.236-2A>T p.? Chr.4: 25383259-25383259 on Build GRCh38
3603081 c.321G>A p.E107E Chr.4: 25383346-25383346 on Build GRCh38
3428446 c.250C>T p.L84F Chr.4: 25383275-25383275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374830999 Chr.4: 25383279-25383279 on Build GRCh38
rs1045828581 Chr.4: 25383538-25383538 on Build GRCh38
rs762596842 Chr.4: 25383307-25383307 on Build GRCh38
rs145334194 Chr.4: 25383472-25383472 on Build GRCh38
rs769312617 Chr.4: 25383293-25383293 on Build GRCh38
rs184722481 Chr.4: 25383488-25383488 on Build GRCh38
rs756223606 Chr.4: 25383362-25383362 on Build GRCh38
rs1034047370 Chr.4: 25383265-25383265 on Build GRCh38
rs771928831 Chr.4: 25383259-25383259 on Build GRCh38
rs762128577 Chr.4: 25383337-25383337 on Build GRCh38
rs748867356 Chr.4: 25383428-25383428 on Build GRCh38
rs773928073 Chr.4: 25383442-25383442 on Build GRCh38
rs750916286 Chr.4: 25383345-25383345 on Build GRCh38
rs746788136 Chr.4: 25383408-25383408 on Build GRCh38
rs183571387 Chr.4: 25383227-25383227 on Build GRCh38
rs941281660 Chr.4: 25383487-25383487 on Build GRCh38
rs188212244 Chr.4: 25383411-25383411 on Build GRCh38
rs759024464 Chr.4: 25383292-25383292 on Build GRCh38
rs750489087 Chr.4: 25383234-25383234 on Build GRCh38
rs891889262 Chr.4: 25383204-25383204 on Build GRCh38
rs201839196 Chr.4: 25383289-25383289 on Build GRCh38
rs753691993 Chr.4: 25383400-25383400 on Build GRCh38
rs13127230 Chr.4: 25383522-25383522 on Build GRCh38
rs756315710 Chr.4: 25383239-25383239 on Build GRCh38
rs376010530 Chr.4: 25383372-25383372 on Build GRCh38
rs147243013 Chr.4: 25383333-25383333 on Build GRCh38
rs745832603 Chr.4: 25383418-25383418 on Build GRCh38
rs114573919 Chr.4: 25383489-25383489 on Build GRCh38
rs944264910 Chr.4: 25383536-25383536 on Build GRCh38
rs1002034579 Chr.4: 25383245-25383245 on Build GRCh38
rs74863363 Chr.4: 25383260-25383260 on Build GRCh38
rs13127336 Chr.4: 25383324-25383324 on Build GRCh38
rs59314097 Chr.4: 25383548-25383549 on Build GRCh38
rs745672751 Chr.4: 25383244-25383245 on Build GRCh38
rs1030385877 Chr.4: 25383422-25383422 on Build GRCh38
rs1011645337 Chr.4: 25383208-25383208 on Build GRCh38
rs932863144 Chr.4: 25383493-25383493 on Build GRCh38
rs746836321 Chr.4: 25383274-25383274 on Build GRCh38
rs908388400 Chr.4: 25383466-25383466 on Build GRCh38
rs200095852 Chr.4: 25383410-25383410 on Build GRCh38
rs775080525 Chr.4: 25383299-25383299 on Build GRCh38
rs556214229 Chr.4: 25383426-25383426 on Build GRCh38
rs1049915330 Chr.4: 25383508-25383508 on Build GRCh38
rs758202065 Chr.4: 25383413-25383414 on Build GRCh38
rs13102409 Chr.4: 25383326-25383326 on Build GRCh38
rs564563091 Chr.4: 25383547-25383548 on Build GRCh38
rs749554010 Chr.4: 25383247-25383247 on Build GRCh38
rs371805695 Chr.4: 25383341-25383341 on Build GRCh38
rs780375315 Chr.4: 25383243-25383243 on Build GRCh38
rs967897206 Chr.4: 25383228-25383228 on Build GRCh38
rs779990618 Chr.4: 25383440-25383441 on Build GRCh38
rs780287319 Chr.4: 25383371-25383371 on Build GRCh38
rs575797971 Chr.4: 25383309-25383309 on Build GRCh38
rs576001123 Chr.4: 25383432-25383432 on Build GRCh38
rs776473600 Chr.4: 25383285-25383285 on Build GRCh38
rs779068339 Chr.4: 25383399-25383399 on Build GRCh38
rs755236879 Chr.4: 25383374-25383374 on Build GRCh38
rs948386734 Chr.4: 25383537-25383537 on Build GRCh38
rs181787523 Chr.4: 25383419-25383419 on Build GRCh38
rs367635861 Chr.4: 25383280-25383280 on Build GRCh38
rs777638838 Chr.4: 25383268-25383268 on Build GRCh38
rs77497792 Chr.4: 25383219-25383219 on Build GRCh38

View Full Product Details