Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 51993995-51994511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC66
Entrez Gene ID
339977
Gene Name
leucine rich repeat containing 66
Gene Aliases
-
UniGene
Hs.661450
Gene Chromosome Location
Chr.4: 51993640-52020311 on Build GRCh38
Transcripts
NM_001024611.2 XM_011534383.2 XM_017008162.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7190016 c.2626T>C p.S876P Chr.4: 51994396-51994396 on Build GRCh38
6100336 c.2606C>A p.A869D Chr.4: 51994416-51994416 on Build GRCh38
312628 c.2599G>A p.D867N Chr.4: 51994423-51994423 on Build GRCh38
6167469 c.2559G>T p.Q853H Chr.4: 51994463-51994463 on Build GRCh38
5951030 c.2550C>T p.D850D Chr.4: 51994472-51994472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949716953 Chr.4: 51994133-51994133 on Build GRCh38
rs767744197 Chr.4: 51994363-51994363 on Build GRCh38
rs913109787 Chr.4: 51994113-51994113 on Build GRCh38
rs200418036 Chr.4: 51994156-51994157 on Build GRCh38
rs1041667376 Chr.4: 51994145-51994145 on Build GRCh38
rs1055466487 Chr.4: 51994459-51994459 on Build GRCh38
rs184898548 Chr.4: 51994081-51994081 on Build GRCh38
rs770490201 Chr.4: 51994336-51994336 on Build GRCh38
rs546627549 Chr.4: 51994280-51994280 on Build GRCh38
rs150288718 Chr.4: 51994128-51994128 on Build GRCh38
rs753129496 Chr.4: 51994351-51994351 on Build GRCh38
rs762136683 Chr.4: 51994341-51994346 on Build GRCh38
rs1004908052 Chr.4: 51994328-51994328 on Build GRCh38
rs545220766 Chr.4: 51994159-51994159 on Build GRCh38
rs746733449 Chr.4: 51994413-51994413 on Build GRCh38
rs911882631 Chr.4: 51994054-51994054 on Build GRCh38
rs754983890 Chr.4: 51994384-51994384 on Build GRCh38
rs565118180 Chr.4: 51994160-51994160 on Build GRCh38
rs921887209 Chr.4: 51994065-51994065 on Build GRCh38
rs920378109 Chr.4: 51994090-51994090 on Build GRCh38
rs376265091 Chr.4: 51994454-51994454 on Build GRCh38
rs770316824 Chr.4: 51994449-51994449 on Build GRCh38
rs776170857 Chr.4: 51994435-51994435 on Build GRCh38
rs566621291 Chr.4: 51994356-51994356 on Build GRCh38
rs561027303 Chr.4: 51994238-51994238 on Build GRCh38
rs368493395 Chr.4: 51994482-51994482 on Build GRCh38
rs957309009 Chr.4: 51994375-51994375 on Build GRCh38
rs776176705 Chr.4: 51994088-51994088 on Build GRCh38
rs981164470 Chr.4: 51994420-51994420 on Build GRCh38
rs773799470 Chr.4: 51994471-51994471 on Build GRCh38
rs188186400 Chr.4: 51994166-51994166 on Build GRCh38
rs1001226313 Chr.4: 51994176-51994176 on Build GRCh38
rs745334377 Chr.4: 51994444-51994444 on Build GRCh38
rs750507814 Chr.4: 51994374-51994374 on Build GRCh38
rs758067621 Chr.4: 51994396-51994396 on Build GRCh38
rs966097081 Chr.4: 51994055-51994055 on Build GRCh38
rs924569196 Chr.4: 51994462-51994462 on Build GRCh38
rs776117486 Chr.4: 51994339-51994339 on Build GRCh38
rs142905075 Chr.4: 51994096-51994096 on Build GRCh38
rs375195073 Chr.4: 51994369-51994369 on Build GRCh38
rs1045536470 Chr.4: 51994174-51994174 on Build GRCh38
rs200802422 Chr.4: 51994162-51994165 on Build GRCh38
rs975043934 Chr.4: 51994056-51994056 on Build GRCh38
rs763187749 Chr.4: 51994355-51994355 on Build GRCh38
rs761291171 Chr.4: 51994391-51994391 on Build GRCh38
rs377708980 Chr.4: 51994214-51994214 on Build GRCh38
rs995373540 Chr.4: 51994215-51994215 on Build GRCh38
rs974780731 Chr.4: 51994366-51994366 on Build GRCh38
rs573555700 Chr.4: 51994045-51994045 on Build GRCh38
rs906849198 Chr.4: 51994175-51994175 on Build GRCh38
rs765714600 Chr.4: 51994349-51994349 on Build GRCh38
rs959515975 Chr.4: 51994398-51994399 on Build GRCh38
rs1055754774 Chr.4: 51994182-51994182 on Build GRCh38
rs775795774 Chr.4: 51994451-51994451 on Build GRCh38
rs371575450 Chr.4: 51994406-51994406 on Build GRCh38
rs928416573 Chr.4: 51994433-51994433 on Build GRCh38
rs550823746 Chr.4: 51994216-51994216 on Build GRCh38
rs774744159 Chr.4: 51994458-51994458 on Build GRCh38
rs913057374 Chr.4: 51994412-51994412 on Build GRCh38
rs1029749765 Chr.4: 51994262-51994262 on Build GRCh38
rs770543622 Chr.4: 51994415-51994415 on Build GRCh38
rs764431146 Chr.4: 51994455-51994455 on Build GRCh38
rs760081149 Chr.4: 51994343-51994343 on Build GRCh38
rs757547718 Chr.4: 51994357-51994357 on Build GRCh38
rs762175753 Chr.4: 51994472-51994472 on Build GRCh38
rs1004587721 Chr.4: 51994319-51994319 on Build GRCh38
rs955394109 Chr.4: 51994309-51994309 on Build GRCh38
rs775680531 Chr.4: 51994142-51994142 on Build GRCh38
rs770164485 Chr.4: 51994370-51994373 on Build GRCh38
rs950916916 Chr.4: 51994248-51994248 on Build GRCh38
rs181155286 Chr.4: 51994385-51994385 on Build GRCh38
rs751804532 Chr.4: 51994162-51994164 on Build GRCh38
rs529995822 Chr.4: 51994272-51994272 on Build GRCh38

View Full Product Details