Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56170107-56170546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1211
Entrez Gene ID
57482
Gene Name
KIAA1211
Gene Aliases
-
UniGene
Hs.596667
Gene Chromosome Location
Chr.4: 56170195-56330724 on Build GRCh38
Transcripts
NM_020722.1

SNP Detail

SNP ID Chromosome Location
rs912988091 Chr.4: 56170207-56170207 on Build GRCh38
rs965001115 Chr.4: 56170152-56170152 on Build GRCh38
rs991711230 Chr.4: 56170170-56170170 on Build GRCh38
rs532463364 Chr.4: 56170178-56170178 on Build GRCh38
rs546648299 Chr.4: 56170166-56170166 on Build GRCh38
rs183859159 Chr.4: 56170143-56170143 on Build GRCh38
rs187769060 Chr.4: 56170372-56170372 on Build GRCh38
rs1020827626 Chr.4: 56170132-56170132 on Build GRCh38
rs1049777009 Chr.4: 56170245-56170245 on Build GRCh38
rs1040563463 Chr.4: 56170351-56170351 on Build GRCh38
rs919833181 Chr.4: 56170278-56170278 on Build GRCh38
rs538005033 Chr.4: 56170268-56170268 on Build GRCh38
rs568960673 Chr.4: 56170238-56170238 on Build GRCh38
rs996857488 Chr.4: 56170168-56170168 on Build GRCh38
rs908718341 Chr.4: 56170192-56170192 on Build GRCh38
rs775225532 Chr.4: 56170159-56170159 on Build GRCh38
rs1019494686 Chr.4: 56170150-56170150 on Build GRCh38
rs891036529 Chr.4: 56170265-56170265 on Build GRCh38
rs746646717 Chr.4: 56170497-56170497 on Build GRCh38
rs1011900602 Chr.4: 56170339-56170339 on Build GRCh38
rs765624568 Chr.4: 56170134-56170134 on Build GRCh38
rs1038436738 Chr.4: 56170223-56170223 on Build GRCh38
rs902346312 Chr.4: 56170280-56170280 on Build GRCh38
rs1033264999 Chr.4: 56170493-56170493 on Build GRCh38
rs755116248 Chr.4: 56170419-56170419 on Build GRCh38
rs940069467 Chr.4: 56170196-56170196 on Build GRCh38
rs544048674 Chr.4: 56170206-56170206 on Build GRCh38
rs3822002 Chr.4: 56170326-56170326 on Build GRCh38
rs1000592629 Chr.4: 56170315-56170315 on Build GRCh38
rs78392512 Chr.4: 56170237-56170237 on Build GRCh38
rs562803716 Chr.4: 56170322-56170322 on Build GRCh38
rs981312319 Chr.4: 56170231-56170231 on Build GRCh38
rs1024682286 Chr.4: 56170343-56170343 on Build GRCh38
rs981720262 Chr.4: 56170398-56170398 on Build GRCh38
rs548068107 Chr.4: 56170271-56170271 on Build GRCh38
rs974242695 Chr.4: 56170482-56170482 on Build GRCh38
rs886529580 Chr.4: 56170340-56170340 on Build GRCh38
rs552982145 Chr.4: 56170480-56170480 on Build GRCh38
rs2276901 Chr.4: 56170140-56170140 on Build GRCh38
rs1009366526 Chr.4: 56170349-56170349 on Build GRCh38
rs959043693 Chr.4: 56170156-56170156 on Build GRCh38
rs1042577822 Chr.4: 56170273-56170273 on Build GRCh38
rs35798439 Chr.4: 56170275-56170276 on Build GRCh38
rs1016155851 Chr.4: 56170446-56170446 on Build GRCh38
rs972022605 Chr.4: 56170370-56170370 on Build GRCh38
rs147377878 Chr.4: 56170172-56170172 on Build GRCh38
rs766357765 Chr.4: 56170188-56170188 on Build GRCh38
rs141180107 Chr.4: 56170139-56170139 on Build GRCh38

View Full Product Details