Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56298128-56298635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1211
Entrez Gene ID
57482
Gene Name
KIAA1211
Gene Aliases
-
UniGene
Hs.596667
Gene Chromosome Location
Chr.4: 56170195-56330724 on Build GRCh38
Transcripts
NM_020722.1 XM_005265752.2 XM_011534397.2 XM_017008473.1 XM_017008474.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7373602 c.46G>A p.G16R Chr.4: 56298275-56298275 on Build GRCh38
6693831 c.101G>A p.G34D Chr.4: 56298330-56298330 on Build GRCh38
3301876 c.39C>T p.I13I Chr.4: 56298268-56298268 on Build GRCh38
6050677 c.113_114delCT p.L39fs*22 Chr.4: 56298342-56298343 on Build GRCh38
4929491 c.3G>A p.M1I Chr.4: 56298232-56298232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549249040 Chr.4: 56298571-56298571 on Build GRCh38
rs749168412 Chr.4: 56298555-56298555 on Build GRCh38
rs893928962 Chr.4: 56298514-56298514 on Build GRCh38
rs202033143 Chr.4: 56298275-56298275 on Build GRCh38
rs774665712 Chr.4: 56298374-56298374 on Build GRCh38
rs768049069 Chr.4: 56298268-56298268 on Build GRCh38
rs368564027 Chr.4: 56298257-56298257 on Build GRCh38
rs777213244 Chr.4: 56298202-56298202 on Build GRCh38
rs763160970 Chr.4: 56298240-56298240 on Build GRCh38
rs982089166 Chr.4: 56298213-56298213 on Build GRCh38
rs769059673 Chr.4: 56298252-56298252 on Build GRCh38
rs765885786 Chr.4: 56298297-56298297 on Build GRCh38
rs571278837 Chr.4: 56298600-56298600 on Build GRCh38
rs1035011363 Chr.4: 56298551-56298551 on Build GRCh38
rs201391712 Chr.4: 56298376-56298376 on Build GRCh38
rs934348896 Chr.4: 56298608-56298608 on Build GRCh38
rs199675859 Chr.4: 56298315-56298315 on Build GRCh38
rs528679131 Chr.4: 56298593-56298593 on Build GRCh38
rs768808654 Chr.4: 56298363-56298363 on Build GRCh38
rs752591162 Chr.4: 56298328-56298328 on Build GRCh38
rs1003875123 Chr.4: 56298574-56298574 on Build GRCh38
rs745895410 Chr.4: 56298333-56298333 on Build GRCh38
rs141400149 Chr.4: 56298239-56298239 on Build GRCh38
rs754811456 Chr.4: 56298191-56298191 on Build GRCh38
rs372368941 Chr.4: 56298200-56298200 on Build GRCh38
rs910285582 Chr.4: 56298583-56298583 on Build GRCh38
rs753536717 Chr.4: 56298317-56298317 on Build GRCh38
rs1056000496 Chr.4: 56298469-56298469 on Build GRCh38
rs755650255 Chr.4: 56298198-56298199 on Build GRCh38
rs59106370 Chr.4: 56298596-56298596 on Build GRCh38
rs1052616669 Chr.4: 56298509-56298509 on Build GRCh38
rs199976955 Chr.4: 56298339-56298339 on Build GRCh38
rs971907705 Chr.4: 56298277-56298277 on Build GRCh38
rs368462281 Chr.4: 56298197-56298197 on Build GRCh38
rs765114565 Chr.4: 56298322-56298322 on Build GRCh38
rs760251486 Chr.4: 56298290-56298290 on Build GRCh38
rs376241108 Chr.4: 56298411-56298411 on Build GRCh38
rs563660365 Chr.4: 56298558-56298558 on Build GRCh38
rs1011040630 Chr.4: 56298518-56298518 on Build GRCh38
rs1040087406 Chr.4: 56298412-56298412 on Build GRCh38
rs376578230 Chr.4: 56298330-56298330 on Build GRCh38
rs267600194 Chr.4: 56298295-56298295 on Build GRCh38
rs777832822 Chr.4: 56298547-56298547 on Build GRCh38
rs944331423 Chr.4: 56298405-56298405 on Build GRCh38
rs753523805 Chr.4: 56298190-56298190 on Build GRCh38
rs577962455 Chr.4: 56298438-56298438 on Build GRCh38
rs905302734 Chr.4: 56298541-56298541 on Build GRCh38
rs527690075 Chr.4: 56298570-56298570 on Build GRCh38
rs901413759 Chr.4: 56298422-56298422 on Build GRCh38
rs746651149 Chr.4: 56298503-56298503 on Build GRCh38
rs758380366 Chr.4: 56298329-56298329 on Build GRCh38
rs372984808 Chr.4: 56298353-56298353 on Build GRCh38
rs762283368 Chr.4: 56298260-56298260 on Build GRCh38
rs368258642 Chr.4: 56298399-56298399 on Build GRCh38
rs993861005 Chr.4: 56298556-56298556 on Build GRCh38
rs745987405 Chr.4: 56298231-56298231 on Build GRCh38
rs781401465 Chr.4: 56298230-56298230 on Build GRCh38
rs150825655 Chr.4: 56298454-56298454 on Build GRCh38
rs542245153 Chr.4: 56298578-56298578 on Build GRCh38
rs111726051 Chr.4: 56298505-56298505 on Build GRCh38
rs184995318 Chr.4: 56298573-56298573 on Build GRCh38
rs749512264 Chr.4: 56298356-56298356 on Build GRCh38
rs772196808 Chr.4: 56298223-56298223 on Build GRCh38
rs754657206 Chr.4: 56298319-56298319 on Build GRCh38
rs769868355 Chr.4: 56298236-56298236 on Build GRCh38
rs748540597 Chr.4: 56298375-56298375 on Build GRCh38
rs1023644168 Chr.4: 56298537-56298537 on Build GRCh38

View Full Product Details