Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56908741-56909237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REST
Entrez Gene ID
5978
Gene Name
RE1 silencing transcription factor
Gene Aliases
NRSF, WT6, XBR
UniGene
Hs.307836
Gene Chromosome Location
Chr.4: 56907876-56935847 on Build GRCh38
Transcripts
NM_001193508.1 NM_005612.4 XM_011534401.2 XM_017008527.1

SNP Detail

SNP ID Chromosome Location
rs529794810 Chr.4: 56908768-56908768 on Build GRCh38
rs990232424 Chr.4: 56908954-56908954 on Build GRCh38
rs113580476 Chr.4: 56909144-56909144 on Build GRCh38
rs922233445 Chr.4: 56909078-56909078 on Build GRCh38
rs1024859473 Chr.4: 56909196-56909196 on Build GRCh38
rs959897470 Chr.4: 56909185-56909185 on Build GRCh38
rs368111764 Chr.4: 56908790-56908790 on Build GRCh38
rs1004858819 Chr.4: 56909209-56909209 on Build GRCh38
rs914649759 Chr.4: 56908957-56908957 on Build GRCh38
rs903685720 Chr.4: 56909010-56909010 on Build GRCh38
rs116000804 Chr.4: 56909204-56909204 on Build GRCh38
rs959526757 Chr.4: 56909043-56909043 on Build GRCh38
rs1013500165 Chr.4: 56909193-56909193 on Build GRCh38
rs1000785002 Chr.4: 56908837-56908837 on Build GRCh38
rs542702832 Chr.4: 56909003-56909003 on Build GRCh38
rs765702626 Chr.4: 56909030-56909030 on Build GRCh38
rs1005043555 Chr.4: 56908909-56908909 on Build GRCh38
rs959407535 Chr.4: 56908844-56908844 on Build GRCh38
rs564949763 Chr.4: 56909118-56909118 on Build GRCh38
rs1055839842 Chr.4: 56908856-56908856 on Build GRCh38
rs1011519538 Chr.4: 56909128-56909128 on Build GRCh38
rs967969468 Chr.4: 56908832-56908832 on Build GRCh38
rs372788537 Chr.4: 56909053-56909053 on Build GRCh38
rs887981104 Chr.4: 56908907-56908907 on Build GRCh38
rs918022801 Chr.4: 56908863-56908863 on Build GRCh38
rs6852182 Chr.4: 56908950-56908950 on Build GRCh38
rs149300866 Chr.4: 56908798-56908798 on Build GRCh38
rs975294536 Chr.4: 56909071-56909071 on Build GRCh38
rs1028653061 Chr.4: 56908985-56909004 on Build GRCh38
rs866205580 Chr.4: 56909013-56909013 on Build GRCh38
rs950760155 Chr.4: 56908875-56908875 on Build GRCh38
rs895092719 Chr.4: 56909182-56909182 on Build GRCh38
rs938380099 Chr.4: 56908847-56908847 on Build GRCh38
rs966670631 Chr.4: 56908928-56908928 on Build GRCh38
rs995216962 Chr.4: 56908872-56908872 on Build GRCh38
rs932354951 Chr.4: 56908990-56908990 on Build GRCh38
rs1029373922 Chr.4: 56908935-56908935 on Build GRCh38
rs1024967070 Chr.4: 56909045-56909045 on Build GRCh38
rs1041651360 Chr.4: 56908829-56908829 on Build GRCh38
rs544765029 Chr.4: 56909113-56909113 on Build GRCh38
rs558075233 Chr.4: 56908803-56908803 on Build GRCh38
rs992539779 Chr.4: 56908861-56908861 on Build GRCh38
rs1005071160 Chr.4: 56909072-56909072 on Build GRCh38
rs976119573 Chr.4: 56908929-56908929 on Build GRCh38
rs998134873 Chr.4: 56909115-56909115 on Build GRCh38
rs1038008136 Chr.4: 56909122-56909122 on Build GRCh38
rs983648237 Chr.4: 56908916-56908916 on Build GRCh38
rs928419137 Chr.4: 56909160-56909160 on Build GRCh38
rs1042127011 Chr.4: 56909125-56909125 on Build GRCh38
rs529758326 Chr.4: 56909211-56909211 on Build GRCh38
rs188609417 Chr.4: 56909047-56909047 on Build GRCh38
rs886841619 Chr.4: 56909064-56909064 on Build GRCh38
rs573984914 Chr.4: 56908971-56908971 on Build GRCh38
rs1021687715 Chr.4: 56909131-56909131 on Build GRCh38
rs546754313 Chr.4: 56908789-56908789 on Build GRCh38
rs372502102 Chr.4: 56909097-56909097 on Build GRCh38
rs6835138 Chr.4: 56908898-56908898 on Build GRCh38
rs544209538 Chr.4: 56909152-56909152 on Build GRCh38
rs1013616011 Chr.4: 56909044-56909044 on Build GRCh38
rs1050754822 Chr.4: 56909102-56909102 on Build GRCh38
rs1050701714 Chr.4: 56908998-56908998 on Build GRCh38
rs909263470 Chr.4: 56909052-56909052 on Build GRCh38
rs932037857 Chr.4: 56908774-56908774 on Build GRCh38
rs991851184 Chr.4: 56909201-56909201 on Build GRCh38
rs922149543 Chr.4: 56908984-56908984 on Build GRCh38
rs909377944 Chr.4: 56908927-56908927 on Build GRCh38
rs1033973758 Chr.4: 56908842-56908842 on Build GRCh38
rs963592337 Chr.4: 56909069-56909069 on Build GRCh38
rs1009382093 Chr.4: 56908797-56908797 on Build GRCh38
rs1042238848 Chr.4: 56909008-56909008 on Build GRCh38
rs933557407 Chr.4: 56909093-56909093 on Build GRCh38
rs983537962 Chr.4: 56909049-56909049 on Build GRCh38
rs77446287 Chr.4: 56909158-56909158 on Build GRCh38
rs1000722978 Chr.4: 56909150-56909150 on Build GRCh38
rs1001071790 Chr.4: 56909022-56909022 on Build GRCh38
rs573005069 Chr.4: 56909099-56909099 on Build GRCh38
rs890907450 Chr.4: 56908999-56908999 on Build GRCh38
rs77252487 Chr.4: 56909157-56909157 on Build GRCh38
rs546619675 Chr.4: 56909212-56909212 on Build GRCh38
rs996882332 Chr.4: 56908784-56908784 on Build GRCh38
rs537365839 Chr.4: 56908922-56908922 on Build GRCh38

View Full Product Details