Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70196592-70197091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ODAM
Entrez Gene ID
54959
Gene Name
odontogenic, ameloblast asssociated
Gene Aliases
APIN
UniGene
Hs.143811
Gene Chromosome Location
Chr.4: 70195728-70204576 on Build GRCh38
Transcripts
NM_017855.3 XM_005265698.3 XM_006714245.3 XM_017008331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6398305 c.74C>A p.S25Y Chr.4: 70196714-70196714 on Build GRCh38
7221077 c.69C>T p.L23L Chr.4: 70196709-70196709 on Build GRCh38
3409447 c.62A>T p.Q21L Chr.4: 70196702-70196702 on Build GRCh38
6101081 c.91G>A p.E31K Chr.4: 70196731-70196731 on Build GRCh38
71917 c.73T>A p.S25T Chr.4: 70196713-70196713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762559764 Chr.4: 70196746-70196746 on Build GRCh38
rs755122598 Chr.4: 70196692-70196692 on Build GRCh38
rs771449053 Chr.4: 70196630-70196630 on Build GRCh38
rs1024713586 Chr.4: 70196784-70196784 on Build GRCh38
rs921500374 Chr.4: 70197054-70197054 on Build GRCh38
rs542967672 Chr.4: 70196667-70196668 on Build GRCh38
rs957868949 Chr.4: 70196846-70196846 on Build GRCh38
rs773623963 Chr.4: 70196627-70196627 on Build GRCh38
rs887948658 Chr.4: 70196712-70196712 on Build GRCh38
rs761676461 Chr.4: 70196752-70196752 on Build GRCh38
rs763672375 Chr.4: 70196709-70196709 on Build GRCh38
rs3756130 Chr.4: 70196811-70196811 on Build GRCh38
rs980599538 Chr.4: 70196823-70196823 on Build GRCh38
rs376371480 Chr.4: 70196743-70196743 on Build GRCh38
rs760461159 Chr.4: 70196668-70196668 on Build GRCh38
rs753237683 Chr.4: 70196764-70196765 on Build GRCh38
rs763832702 Chr.4: 70196748-70196748 on Build GRCh38
rs767363966 Chr.4: 70196754-70196754 on Build GRCh38
rs748387440 Chr.4: 70196704-70196704 on Build GRCh38
rs987040257 Chr.4: 70197064-70197064 on Build GRCh38
rs542475015 Chr.4: 70196946-70196946 on Build GRCh38
rs758616960 Chr.4: 70196705-70196705 on Build GRCh38
rs750323696 Chr.4: 70196691-70196691 on Build GRCh38
rs188812959 Chr.4: 70196734-70196734 on Build GRCh38
rs777135540 Chr.4: 70196711-70196711 on Build GRCh38
rs756956184 Chr.4: 70196659-70196659 on Build GRCh38
rs897452459 Chr.4: 70196927-70196927 on Build GRCh38
rs200439171 Chr.4: 70196617-70196617 on Build GRCh38
rs17704351 Chr.4: 70196709-70196709 on Build GRCh38
rs969580243 Chr.4: 70196818-70196818 on Build GRCh38
rs556354154 Chr.4: 70196891-70196891 on Build GRCh38
rs564899447 Chr.4: 70197005-70197005 on Build GRCh38
rs372000636 Chr.4: 70196721-70196721 on Build GRCh38
rs531986434 Chr.4: 70197030-70197030 on Build GRCh38
rs3756131 Chr.4: 70196625-70196625 on Build GRCh38
rs944454462 Chr.4: 70196882-70196882 on Build GRCh38
rs550668503 Chr.4: 70197031-70197031 on Build GRCh38
rs372202605 Chr.4: 70196648-70196648 on Build GRCh38
rs777807789 Chr.4: 70196708-70196708 on Build GRCh38
rs759051239 Chr.4: 70196635-70196635 on Build GRCh38
rs184431039 Chr.4: 70197042-70197042 on Build GRCh38
rs898101075 Chr.4: 70197068-70197068 on Build GRCh38
rs773839802 Chr.4: 70196750-70196750 on Build GRCh38
rs572817738 Chr.4: 70196989-70196989 on Build GRCh38
rs779283484 Chr.4: 70196618-70196618 on Build GRCh38
rs193206771 Chr.4: 70197001-70197001 on Build GRCh38
rs913829846 Chr.4: 70196869-70196869 on Build GRCh38
rs779124059 Chr.4: 70196701-70196701 on Build GRCh38
rs72856564 Chr.4: 70196975-70196975 on Build GRCh38
rs764666343 Chr.4: 70196636-70196636 on Build GRCh38
rs1036112193 Chr.4: 70196902-70196902 on Build GRCh38
rs1051221723 Chr.4: 70196833-70196833 on Build GRCh38
rs1034839138 Chr.4: 70196986-70196986 on Build GRCh38
rs771238018 Chr.4: 70196825-70196825 on Build GRCh38
rs751349409 Chr.4: 70196657-70196657 on Build GRCh38
rs377655517 Chr.4: 70197025-70197025 on Build GRCh38
rs912752536 Chr.4: 70196841-70196841 on Build GRCh38
rs771218035 Chr.4: 70196710-70196710 on Build GRCh38
rs202029273 Chr.4: 70196639-70196639 on Build GRCh38
rs747517020 Chr.4: 70196628-70196628 on Build GRCh38
rs887919241 Chr.4: 70196996-70196997 on Build GRCh38
rs750444449 Chr.4: 70196767-70196767 on Build GRCh38
rs777082247 Chr.4: 70196634-70196634 on Build GRCh38
rs921520540 Chr.4: 70196803-70196803 on Build GRCh38
rs201390555 Chr.4: 70196664-70196664 on Build GRCh38
rs988032277 Chr.4: 70196862-70196862 on Build GRCh38
rs954275532 Chr.4: 70197062-70197062 on Build GRCh38
rs748345678 Chr.4: 70196624-70196624 on Build GRCh38
rs529718278 Chr.4: 70197047-70197047 on Build GRCh38
rs575100805 Chr.4: 70196945-70196945 on Build GRCh38
rs905127026 Chr.4: 70196959-70196959 on Build GRCh38
rs756961220 Chr.4: 70196765-70196765 on Build GRCh38

View Full Product Details