Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99529490-99529999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf17
Entrez Gene ID
84103
Gene Name
chromosome 4 open reading frame 17
Gene Aliases
-
UniGene
Hs.97501
Gene Chromosome Location
Chr.4: 99511004-99542303 on Build GRCh38
Transcripts
NM_032149.2 XM_011532315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
254591 c.415G>A p.A139T Chr.4: 99529827-99529827 on Build GRCh38
6165964 c.532G>T p.D178Y Chr.4: 99529944-99529944 on Build GRCh38
5430117 c.422G>T p.R141I Chr.4: 99529834-99529834 on Build GRCh38
1049425 c.532G>A p.D178N Chr.4: 99529944-99529944 on Build GRCh38
1049424 c.463T>C p.C155R Chr.4: 99529875-99529875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41275703 Chr.4: 99529800-99529800 on Build GRCh38
rs774297173 Chr.4: 99529857-99529857 on Build GRCh38
rs529127629 Chr.4: 99529713-99529713 on Build GRCh38
rs884275 Chr.4: 99529555-99529555 on Build GRCh38
rs1033353832 Chr.4: 99529561-99529561 on Build GRCh38
rs981198294 Chr.4: 99529661-99529661 on Build GRCh38
rs750685200 Chr.4: 99529749-99529749 on Build GRCh38
rs755603949 Chr.4: 99529769-99529769 on Build GRCh38
rs915169041 Chr.4: 99529686-99529686 on Build GRCh38
rs767101343 Chr.4: 99529864-99529864 on Build GRCh38
rs45440196 Chr.4: 99529970-99529970 on Build GRCh38
rs884276 Chr.4: 99529616-99529616 on Build GRCh38
rs756064867 Chr.4: 99529521-99529521 on Build GRCh38
rs912866178 Chr.4: 99529620-99529620 on Build GRCh38
rs556573313 Chr.4: 99529893-99529894 on Build GRCh38
rs764543947 Chr.4: 99529922-99529922 on Build GRCh38
rs769958125 Chr.4: 99529813-99529813 on Build GRCh38
rs565275269 Chr.4: 99529596-99529596 on Build GRCh38
rs768226123 Chr.4: 99529805-99529809 on Build GRCh38
rs757665778 Chr.4: 99529941-99529941 on Build GRCh38
rs927984007 Chr.4: 99529652-99529652 on Build GRCh38
rs547671748 Chr.4: 99529551-99529551 on Build GRCh38
rs142676237 Chr.4: 99529923-99529923 on Build GRCh38
rs148607252 Chr.4: 99529894-99529894 on Build GRCh38
rs982196102 Chr.4: 99529648-99529648 on Build GRCh38
rs569488998 Chr.4: 99529658-99529658 on Build GRCh38
rs920190334 Chr.4: 99529759-99529759 on Build GRCh38
rs768749789 Chr.4: 99529963-99529963 on Build GRCh38
rs562665798 Chr.4: 99529950-99529950 on Build GRCh38
rs1038507269 Chr.4: 99529728-99529728 on Build GRCh38
rs778119354 Chr.4: 99529792-99529792 on Build GRCh38
rs758885644 Chr.4: 99529786-99529786 on Build GRCh38
rs749318661 Chr.4: 99529962-99529962 on Build GRCh38
rs753225264 Chr.4: 99529895-99529895 on Build GRCh38
rs781424563 Chr.4: 99529801-99529801 on Build GRCh38
rs760236429 Chr.4: 99529874-99529874 on Build GRCh38
rs780196382 Chr.4: 99529960-99529960 on Build GRCh38
rs371184535 Chr.4: 99529949-99529949 on Build GRCh38
rs957285273 Chr.4: 99529575-99529575 on Build GRCh38
rs776318381 Chr.4: 99529882-99529882 on Build GRCh38
rs145022283 Chr.4: 99529883-99529894 on Build GRCh38
rs758910207 Chr.4: 99529918-99529918 on Build GRCh38
rs779397720 Chr.4: 99529773-99529773 on Build GRCh38
rs145411536 Chr.4: 99529873-99529873 on Build GRCh38
rs377453185 Chr.4: 99529803-99529803 on Build GRCh38
rs927029059 Chr.4: 99529697-99529697 on Build GRCh38
rs575779535 Chr.4: 99529848-99529848 on Build GRCh38
rs761663082 Chr.4: 99529861-99529861 on Build GRCh38
rs386677741 Chr.4: 99529882-99529894 on Build GRCh38
rs555480244 Chr.4: 99529882-99529891 on Build GRCh38
rs747307329 Chr.4: 99529794-99529794 on Build GRCh38
rs774385326 Chr.4: 99529969-99529969 on Build GRCh38
rs753311460 Chr.4: 99529782-99529782 on Build GRCh38
rs766755696 Chr.4: 99529957-99529960 on Build GRCh38
rs768660292 Chr.4: 99529856-99529856 on Build GRCh38
rs884274 Chr.4: 99529536-99529536 on Build GRCh38
rs756278051 Chr.4: 99529954-99529954 on Build GRCh38
rs988064373 Chr.4: 99529664-99529664 on Build GRCh38
rs200233951 Chr.4: 99529884-99529894 on Build GRCh38
rs866920825 Chr.4: 99529882-99529882 on Build GRCh38
rs1048737638 Chr.4: 99529865-99529865 on Build GRCh38
rs150415629 Chr.4: 99529726-99529726 on Build GRCh38
rs775429541 Chr.4: 99529842-99529842 on Build GRCh38

View Full Product Details