Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 108811659-108812165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL25A1
Entrez Gene ID
84570
Gene Name
collagen type XXV alpha 1
Gene Aliases
AMY, CFEOM5, CLAC, CLAC-P, CLACP
UniGene
Hs.658842
Gene Chromosome Location
Chr.4: 108808725-109302737 on Build GRCh38
Transcripts
NM_198721.3 NR_045756.2 XM_011532333.2 XM_011532334.2 XM_011532335.2 XM_011532338.2 XM_011532355.2 XM_011532356.2 XM_011532358.2 XM_017008735.1 XM_017008736.1 XM_017008737.1

SNP Detail

SNP ID Chromosome Location
rs1035406786 Chr.4: 108811784-108811784 on Build GRCh38
rs577336412 Chr.4: 108811777-108811777 on Build GRCh38
rs985026422 Chr.4: 108811839-108811839 on Build GRCh38
rs186200886 Chr.4: 108812056-108812056 on Build GRCh38
rs967063377 Chr.4: 108811725-108811725 on Build GRCh38
rs562405607 Chr.4: 108811942-108811942 on Build GRCh38
rs952212307 Chr.4: 108811836-108811836 on Build GRCh38
rs968612614 Chr.4: 108811971-108811971 on Build GRCh38
rs13151012 Chr.4: 108812016-108812016 on Build GRCh38
rs1021992948 Chr.4: 108811944-108811944 on Build GRCh38
rs563811286 Chr.4: 108811696-108811696 on Build GRCh38
rs923354892 Chr.4: 108811887-108811887 on Build GRCh38
rs1049145377 Chr.4: 108811785-108811785 on Build GRCh38
rs892961034 Chr.4: 108812017-108812017 on Build GRCh38
rs918660322 Chr.4: 108812045-108812045 on Build GRCh38
rs979947612 Chr.4: 108811984-108811984 on Build GRCh38
rs938656492 Chr.4: 108812023-108812023 on Build GRCh38
rs1040778112 Chr.4: 108811851-108811851 on Build GRCh38
rs528858467 Chr.4: 108811707-108811707 on Build GRCh38
rs539917571 Chr.4: 108812090-108812090 on Build GRCh38
rs930146426 Chr.4: 108811755-108811755 on Build GRCh38
rs137922426 Chr.4: 108811850-108811850 on Build GRCh38
rs916768481 Chr.4: 108811835-108811835 on Build GRCh38
rs1030055520 Chr.4: 108811894-108811894 on Build GRCh38
rs1005925996 Chr.4: 108811816-108811816 on Build GRCh38
rs938769774 Chr.4: 108811699-108811699 on Build GRCh38
rs956097214 Chr.4: 108811914-108811914 on Build GRCh38
rs142622339 Chr.4: 108811810-108811810 on Build GRCh38
rs568626659 Chr.4: 108812018-108812018 on Build GRCh38
rs570891790 Chr.4: 108812078-108812078 on Build GRCh38
rs1017290014 Chr.4: 108811832-108811832 on Build GRCh38
rs150263923 Chr.4: 108811849-108811849 on Build GRCh38
rs992900435 Chr.4: 108812041-108812041 on Build GRCh38
rs932088579 Chr.4: 108811978-108811978 on Build GRCh38
rs1057119494 Chr.4: 108811708-108811708 on Build GRCh38
rs897328675 Chr.4: 108811738-108811738 on Build GRCh38
rs1048549279 Chr.4: 108812075-108812075 on Build GRCh38
rs548556003 Chr.4: 108811929-108811929 on Build GRCh38
rs958708246 Chr.4: 108811799-108811799 on Build GRCh38
rs774661723 Chr.4: 108811706-108811706 on Build GRCh38
rs1020188430 Chr.4: 108811689-108811689 on Build GRCh38
rs377002199 Chr.4: 108811985-108811985 on Build GRCh38
rs1002993097 Chr.4: 108811768-108811768 on Build GRCh38
rs35191642 Chr.4: 108811753-108811753 on Build GRCh38
rs532918866 Chr.4: 108812103-108812103 on Build GRCh38
rs772497440 Chr.4: 108811741-108811741 on Build GRCh38

View Full Product Details