Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 118294833-118295330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS12
Entrez Gene ID
8492
Gene Name
protease, serine 12
Gene Aliases
BSSP-3, BSSP3, MRT1
UniGene
Hs.445857
Gene Chromosome Location
Chr.4: 118280038-118353042 on Build GRCh38
Transcripts
NM_003619.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4718195 c.1949T>A p.L650Q Chr.4: 118295029-118295029 on Build GRCh38
7078949 c.2016C>T p.L672L Chr.4: 118294962-118294962 on Build GRCh38
6512484 c.1963G>T p.G655* Chr.4: 118295015-118295015 on Build GRCh38
4630920 c.1992G>A p.T664T Chr.4: 118294986-118294986 on Build GRCh38
4969611 c.2010G>A p.W670* Chr.4: 118294968-118294968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775145863 Chr.4: 118294972-118294973 on Build GRCh38
rs1024837016 Chr.4: 118295227-118295227 on Build GRCh38
rs371384574 Chr.4: 118295004-118295004 on Build GRCh38
rs773838679 Chr.4: 118295114-118295114 on Build GRCh38
rs142551296 Chr.4: 118294944-118294944 on Build GRCh38
rs768700323 Chr.4: 118295005-118295005 on Build GRCh38
rs34325701 Chr.4: 118295236-118295237 on Build GRCh38
rs911330406 Chr.4: 118295296-118295296 on Build GRCh38
rs1039791622 Chr.4: 118295237-118295237 on Build GRCh38
rs79009259 Chr.4: 118294871-118294871 on Build GRCh38
rs923915777 Chr.4: 118295264-118295264 on Build GRCh38
rs991096160 Chr.4: 118295219-118295219 on Build GRCh38
rs756760181 Chr.4: 118294994-118294994 on Build GRCh38
rs944121410 Chr.4: 118294890-118294890 on Build GRCh38
rs541113476 Chr.4: 118294953-118294953 on Build GRCh38
rs936684321 Chr.4: 118295275-118295275 on Build GRCh38
rs1005933414 Chr.4: 118295157-118295157 on Build GRCh38
rs370409320 Chr.4: 118294980-118294980 on Build GRCh38
rs542351163 Chr.4: 118295059-118295059 on Build GRCh38
rs753518116 Chr.4: 118295037-118295037 on Build GRCh38
rs775755695 Chr.4: 118295235-118295235 on Build GRCh38
rs780905213 Chr.4: 118295089-118295089 on Build GRCh38
rs989577172 Chr.4: 118295276-118295276 on Build GRCh38
rs748357638 Chr.4: 118295110-118295110 on Build GRCh38
rs776520246 Chr.4: 118294950-118294950 on Build GRCh38
rs376368168 Chr.4: 118294958-118294958 on Build GRCh38
rs758278960 Chr.4: 118295075-118295075 on Build GRCh38
rs771497164 Chr.4: 118295032-118295032 on Build GRCh38
rs377739049 Chr.4: 118295003-118295003 on Build GRCh38
rs770190361 Chr.4: 118295070-118295070 on Build GRCh38
rs756956608 Chr.4: 118295042-118295042 on Build GRCh38
rs748610764 Chr.4: 118294979-118294979 on Build GRCh38
rs899653840 Chr.4: 118294898-118294898 on Build GRCh38
rs748852707 Chr.4: 118294895-118294895 on Build GRCh38
rs200793018 Chr.4: 118294893-118294893 on Build GRCh38
rs969526761 Chr.4: 118295127-118295127 on Build GRCh38
rs886117210 Chr.4: 118295143-118295143 on Build GRCh38
rs771843623 Chr.4: 118294922-118294922 on Build GRCh38
rs759951105 Chr.4: 118295033-118295033 on Build GRCh38
rs780052141 Chr.4: 118295081-118295081 on Build GRCh38
rs901452844 Chr.4: 118295086-118295086 on Build GRCh38
rs925524885 Chr.4: 118295149-118295149 on Build GRCh38
rs767839848 Chr.4: 118295035-118295035 on Build GRCh38
rs750961138 Chr.4: 118294957-118294957 on Build GRCh38
rs545871382 Chr.4: 118295162-118295162 on Build GRCh38
rs763514450 Chr.4: 118295026-118295026 on Build GRCh38
rs754797015 Chr.4: 118295085-118295085 on Build GRCh38
rs950469719 Chr.4: 118294934-118294934 on Build GRCh38
rs776916062 Chr.4: 118295010-118295010 on Build GRCh38
rs749963084 Chr.4: 118295070-118295070 on Build GRCh38
rs370102404 Chr.4: 118294929-118294929 on Build GRCh38
rs775316328 Chr.4: 118294936-118294936 on Build GRCh38
rs374737501 Chr.4: 118294987-118294987 on Build GRCh38
rs544110238 Chr.4: 118295014-118295014 on Build GRCh38
rs770717600 Chr.4: 118294896-118294896 on Build GRCh38
rs745564161 Chr.4: 118294908-118294908 on Build GRCh38
rs959235184 Chr.4: 118295215-118295215 on Build GRCh38
rs778454527 Chr.4: 118294995-118294995 on Build GRCh38
rs959473801 Chr.4: 118294956-118294956 on Build GRCh38
rs771669453 Chr.4: 118294915-118294918 on Build GRCh38
rs745970015 Chr.4: 118295165-118295165 on Build GRCh38
rs201706327 Chr.4: 118294939-118294939 on Build GRCh38
rs769936150 Chr.4: 118295169-118295169 on Build GRCh38
rs762721819 Chr.4: 118295236-118295236 on Build GRCh38
rs376037431 Chr.4: 118295090-118295090 on Build GRCh38
rs575065414 Chr.4: 118295013-118295013 on Build GRCh38
rs1054293043 Chr.4: 118295062-118295062 on Build GRCh38
rs765772252 Chr.4: 118294955-118294955 on Build GRCh38
rs887056461 Chr.4: 118294959-118294959 on Build GRCh38
rs754208404 Chr.4: 118295083-118295083 on Build GRCh38
rs755675973 Chr.4: 118294983-118294983 on Build GRCh38
rs745877442 Chr.4: 118294916-118294916 on Build GRCh38
rs746711778 Chr.4: 118295082-118295082 on Build GRCh38
rs957180305 Chr.4: 118295167-118295167 on Build GRCh38
rs781080768 Chr.4: 118295138-118295138 on Build GRCh38
rs3828500 Chr.4: 118294899-118294899 on Build GRCh38

View Full Product Details