Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 134198591-134199140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PABPC4L
Entrez Gene ID
132430
Gene Name
poly(A) binding protein cytoplasmic 4 like
Gene Aliases
-
UniGene
Hs.49889
Gene Chromosome Location
Chr.4: 133948459-134201748 on Build GRCh38
Transcripts
NM_001114734.1 XM_017007727.1

SNP Detail

SNP ID Chromosome Location
rs995356419 Chr.4: 134198913-134198913 on Build GRCh38
rs1015596542 Chr.4: 134198740-134198740 on Build GRCh38
rs192219867 Chr.4: 134199048-134199048 on Build GRCh38
rs1043142294 Chr.4: 134198876-134198876 on Build GRCh38
rs974172991 Chr.4: 134199073-134199073 on Build GRCh38
rs1029254597 Chr.4: 134199026-134199026 on Build GRCh38
rs1035964238 Chr.4: 134198662-134198662 on Build GRCh38
rs954031156 Chr.4: 134199087-134199087 on Build GRCh38
rs992203833 Chr.4: 134198720-134198720 on Build GRCh38
rs112656129 Chr.4: 134199085-134199085 on Build GRCh38
rs1001899754 Chr.4: 134198887-134198887 on Build GRCh38
rs529510729 Chr.4: 134199068-134199068 on Build GRCh38
rs567265618 Chr.4: 134198660-134198660 on Build GRCh38
rs1018980868 Chr.4: 134198969-134198969 on Build GRCh38
rs1004958603 Chr.4: 134198966-134198966 on Build GRCh38
rs73858049 Chr.4: 134198866-134198866 on Build GRCh38
rs183488621 Chr.4: 134199101-134199101 on Build GRCh38
rs566597148 Chr.4: 134198878-134198878 on Build GRCh38
rs968664581 Chr.4: 134199041-134199041 on Build GRCh38
rs145573571 Chr.4: 134198930-134198930 on Build GRCh38
rs35497148 Chr.4: 134198769-134198770 on Build GRCh38
rs891526638 Chr.4: 134198839-134198839 on Build GRCh38
rs944152886 Chr.4: 134198805-134198805 on Build GRCh38
rs966312193 Chr.4: 134198723-134198723 on Build GRCh38
rs996551565 Chr.4: 134198999-134198999 on Build GRCh38
rs956481788 Chr.4: 134199047-134199047 on Build GRCh38
rs912789410 Chr.4: 134199102-134199102 on Build GRCh38
rs912737219 Chr.4: 134198785-134198785 on Build GRCh38
rs74405964 Chr.4: 134198915-134198915 on Build GRCh38
rs1015650596 Chr.4: 134198971-134198971 on Build GRCh38
rs979973173 Chr.4: 134198758-134198758 on Build GRCh38
rs974456363 Chr.4: 134198749-134198749 on Build GRCh38
rs112557752 Chr.4: 134198762-134198762 on Build GRCh38
rs867271408 Chr.4: 134198751-134198751 on Build GRCh38
rs1027439927 Chr.4: 134199080-134199080 on Build GRCh38
rs991849984 Chr.4: 134199062-134199062 on Build GRCh38
rs901863442 Chr.4: 134198988-134198988 on Build GRCh38
rs1031119523 Chr.4: 134199046-134199046 on Build GRCh38
rs1018950148 Chr.4: 134198706-134198706 on Build GRCh38
rs569088812 Chr.4: 134199043-134199043 on Build GRCh38
rs917639810 Chr.4: 134199063-134199063 on Build GRCh38
rs975004879 Chr.4: 134198739-134198739 on Build GRCh38
rs554889907 Chr.4: 134199072-134199072 on Build GRCh38
rs947756897 Chr.4: 134199065-134199065 on Build GRCh38
rs372857020 Chr.4: 134199082-134199082 on Build GRCh38
rs985727797 Chr.4: 134198779-134198779 on Build GRCh38
rs1052867476 Chr.4: 134198826-134198826 on Build GRCh38
rs961371402 Chr.4: 134198743-134198743 on Build GRCh38
rs959860880 Chr.4: 134198663-134198663 on Build GRCh38
rs1035533801 Chr.4: 134198888-134198888 on Build GRCh38

View Full Product Details