Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152960399-152960941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHDC1
Entrez Gene ID
85462
Gene Name
FH2 domain containing 1
Gene Aliases
-
UniGene
Hs.132629
Gene Chromosome Location
Chr.4: 152911570-152979696 on Build GRCh38
Transcripts
NM_033393.2 XM_005263319.3 XM_006714393.3 XM_011532388.1 XM_011532389.1 XM_017008787.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3128177 c.742G>T p.V248L Chr.4: 152960643-152960643 on Build GRCh38
4509521 c.809C>T p.S270F Chr.4: 152960803-152960803 on Build GRCh38
5671327 c.787G>C p.E263Q Chr.4: 152960781-152960781 on Build GRCh38
1052179 c.690C>T p.G230G Chr.4: 152960591-152960591 on Build GRCh38
1052180 c.769G>A p.A257T Chr.4: 152960763-152960763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758411160 Chr.4: 152960606-152960606 on Build GRCh38
rs774821132 Chr.4: 152960790-152960790 on Build GRCh38
rs754973713 Chr.4: 152960857-152960857 on Build GRCh38
rs754175402 Chr.4: 152960837-152960837 on Build GRCh38
rs775916311 Chr.4: 152960891-152960891 on Build GRCh38
rs780794533 Chr.4: 152960861-152960861 on Build GRCh38
rs745830413 Chr.4: 152960670-152960672 on Build GRCh38
rs754796627 Chr.4: 152960727-152960727 on Build GRCh38
rs143448808 Chr.4: 152960600-152960600 on Build GRCh38
rs373511952 Chr.4: 152960552-152960552 on Build GRCh38
rs760748106 Chr.4: 152960817-152960817 on Build GRCh38
rs751935867 Chr.4: 152960591-152960591 on Build GRCh38
rs552055341 Chr.4: 152960431-152960431 on Build GRCh38
rs879216627 Chr.4: 152960449-152960449 on Build GRCh38
rs181223640 Chr.4: 152960424-152960424 on Build GRCh38
rs978509112 Chr.4: 152960463-152960463 on Build GRCh38
rs200501757 Chr.4: 152960849-152960849 on Build GRCh38
rs756128733 Chr.4: 152960599-152960599 on Build GRCh38
rs745870340 Chr.4: 152960536-152960536 on Build GRCh38
rs369048099 Chr.4: 152960541-152960541 on Build GRCh38
rs146915636 Chr.4: 152960592-152960592 on Build GRCh38
rs141616723 Chr.4: 152960612-152960612 on Build GRCh38
rs371561970 Chr.4: 152960522-152960522 on Build GRCh38
rs770472175 Chr.4: 152960769-152960769 on Build GRCh38
rs781060983 Chr.4: 152960730-152960730 on Build GRCh38
rs745749523 Chr.4: 152960603-152960603 on Build GRCh38
rs761953138 Chr.4: 152960840-152960840 on Build GRCh38
rs917498928 Chr.4: 152960494-152960494 on Build GRCh38
rs112307773 Chr.4: 152960850-152960850 on Build GRCh38
rs764011436 Chr.4: 152960687-152960687 on Build GRCh38
rs777847344 Chr.4: 152960540-152960540 on Build GRCh38
rs374335444 Chr.4: 152960866-152960866 on Build GRCh38
rs570307116 Chr.4: 152960906-152960906 on Build GRCh38
rs533371393 Chr.4: 152960881-152960881 on Build GRCh38
rs769501188 Chr.4: 152960746-152960746 on Build GRCh38
rs561245289 Chr.4: 152960473-152960473 on Build GRCh38
rs761224747 Chr.4: 152960565-152960565 on Build GRCh38
rs752524912 Chr.4: 152960863-152960863 on Build GRCh38
rs576289636 Chr.4: 152960703-152960703 on Build GRCh38
rs531155459 Chr.4: 152960910-152960910 on Build GRCh38
rs376050246 Chr.4: 152960714-152960714 on Build GRCh38
rs924379514 Chr.4: 152960634-152960634 on Build GRCh38
rs760061037 Chr.4: 152960794-152960794 on Build GRCh38
rs776116429 Chr.4: 152960659-152960659 on Build GRCh38
rs934312027 Chr.4: 152960480-152960480 on Build GRCh38
rs545811494 Chr.4: 152960539-152960540 on Build GRCh38
rs1006151658 Chr.4: 152960575-152960575 on Build GRCh38
rs770873640 Chr.4: 152960649-152960652 on Build GRCh38
rs978981118 Chr.4: 152960468-152960468 on Build GRCh38
rs1020342682 Chr.4: 152960898-152960899 on Build GRCh38
rs780711170 Chr.4: 152960470-152960470 on Build GRCh38
rs903259134 Chr.4: 152960905-152960905 on Build GRCh38
rs112421953 Chr.4: 152960595-152960595 on Build GRCh38
rs764360863 Chr.4: 152960821-152960821 on Build GRCh38
rs774234954 Chr.4: 152960659-152960660 on Build GRCh38
rs4696369 Chr.4: 152960905-152960905 on Build GRCh38
rs910414620 Chr.4: 152960434-152960434 on Build GRCh38
rs772643352 Chr.4: 152960890-152960890 on Build GRCh38
rs763484924 Chr.4: 152960588-152960588 on Build GRCh38
rs148225074 Chr.4: 152960755-152960755 on Build GRCh38
rs751586117 Chr.4: 152960720-152960720 on Build GRCh38
rs768431824 Chr.4: 152960556-152960556 on Build GRCh38
rs142466283 Chr.4: 152960728-152960729 on Build GRCh38
rs778427895 Chr.4: 152960876-152960876 on Build GRCh38
rs768294294 Chr.4: 152960437-152960437 on Build GRCh38
rs191004080 Chr.4: 152960719-152960719 on Build GRCh38
rs372587056 Chr.4: 152960765-152960765 on Build GRCh38
rs768639500 Chr.4: 152960656-152960656 on Build GRCh38
rs979012592 Chr.4: 152960620-152960620 on Build GRCh38
rs767725606 Chr.4: 152960594-152960594 on Build GRCh38
rs926211801 Chr.4: 152960471-152960472 on Build GRCh38
rs141244256 Chr.4: 152960815-152960815 on Build GRCh38
rs376255247 Chr.4: 152960665-152960665 on Build GRCh38
rs555505077 Chr.4: 152960523-152960523 on Build GRCh38
rs115796267 Chr.4: 152960867-152960867 on Build GRCh38
rs371769118 Chr.4: 152960684-152960684 on Build GRCh38
rs1017956108 Chr.4: 152960605-152960605 on Build GRCh38
rs747883162 Chr.4: 152960663-152960663 on Build GRCh38
rs896333447 Chr.4: 152960626-152960626 on Build GRCh38
rs982528163 Chr.4: 152960800-152960800 on Build GRCh38
rs374612781 Chr.4: 152960582-152960582 on Build GRCh38
rs954647929 Chr.4: 152960430-152960430 on Build GRCh38
rs766858840 Chr.4: 152960675-152960675 on Build GRCh38
rs553614897 Chr.4: 152960807-152960807 on Build GRCh38
rs1056761433 Chr.4: 152960493-152960493 on Build GRCh38
rs3811834 Chr.4: 152960683-152960683 on Build GRCh38
rs770383925 Chr.4: 152960870-152960870 on Build GRCh38
rs202120483 Chr.4: 152960560-152960560 on Build GRCh38
rs746783537 Chr.4: 152960616-152960616 on Build GRCh38
rs756392174 Chr.4: 152960686-152960686 on Build GRCh38
rs200988003 Chr.4: 152960664-152960664 on Build GRCh38
rs766664654 Chr.4: 152960590-152960590 on Build GRCh38
rs569922855 Chr.4: 152960478-152960478 on Build GRCh38
rs747690266 Chr.4: 152960745-152960745 on Build GRCh38
rs149221149 Chr.4: 152960581-152960581 on Build GRCh38
rs748163006 Chr.4: 152960519-152960519 on Build GRCh38
rs561578259 Chr.4: 152960754-152960754 on Build GRCh38
rs774905152 Chr.4: 152960682-152960682 on Build GRCh38
rs749380139 Chr.4: 152960547-152960547 on Build GRCh38
rs775454602 Chr.4: 152960545-152960545 on Build GRCh38

View Full Product Details