Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158680235-158680677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETFDH
Entrez Gene ID
2110
Gene Name
electron transfer flavoprotein dehydrogenase
Gene Aliases
ETFQO, MADD
UniGene
Hs.155729
Gene Chromosome Location
Chr.4: 158672101-158708713 on Build GRCh38
Transcripts
NM_001281737.1 NM_004453.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4158888 c.92C>T p.T31I Chr.4: 158680524-158680524 on Build GRCh38
4822135 c.49C>T p.H17Y Chr.4: 158680481-158680481 on Build GRCh38
1428302 c.82C>A p.L28I Chr.4: 158680514-158680514 on Build GRCh38
232269 c.64A>G p.K22E Chr.4: 158680496-158680496 on Build GRCh38
3674125 c.53C>T p.A18V Chr.4: 158680485-158680485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182144074 Chr.4: 158680523-158680523 on Build GRCh38
rs201823591 Chr.4: 158680574-158680574 on Build GRCh38
rs368345149 Chr.4: 158680436-158680436 on Build GRCh38
rs750425050 Chr.4: 158680590-158680590 on Build GRCh38
rs778798030 Chr.4: 158680469-158680469 on Build GRCh38
rs771759141 Chr.4: 158680653-158680655 on Build GRCh38
rs199546824 Chr.4: 158680556-158680556 on Build GRCh38
rs778179798 Chr.4: 158680454-158680454 on Build GRCh38
rs534388496 Chr.4: 158680584-158680584 on Build GRCh38
rs781151266 Chr.4: 158680610-158680610 on Build GRCh38
rs917824297 Chr.4: 158680600-158680600 on Build GRCh38
rs796051964 Chr.4: 158680483-158680484 on Build GRCh38
rs144283843 Chr.4: 158680626-158680626 on Build GRCh38
rs1016190950 Chr.4: 158680377-158680377 on Build GRCh38
rs150105001 Chr.4: 158680554-158680554 on Build GRCh38
rs753999138 Chr.4: 158680461-158680461 on Build GRCh38
rs1053865022 Chr.4: 158680516-158680516 on Build GRCh38
rs878853006 Chr.4: 158680589-158680589 on Build GRCh38
rs558014701 Chr.4: 158680372-158680372 on Build GRCh38
rs768442787 Chr.4: 158680475-158680475 on Build GRCh38
rs774060297 Chr.4: 158680560-158680560 on Build GRCh38
rs1035530852 Chr.4: 158680330-158680330 on Build GRCh38
rs766553154 Chr.4: 158680425-158680425 on Build GRCh38
rs530680991 Chr.4: 158680311-158680311 on Build GRCh38
rs949249162 Chr.4: 158680603-158680603 on Build GRCh38
rs748076363 Chr.4: 158680618-158680618 on Build GRCh38
rs776108657 Chr.4: 158680565-158680565 on Build GRCh38
rs201254467 Chr.4: 158680468-158680468 on Build GRCh38
rs758453563 Chr.4: 158680607-158680607 on Build GRCh38
rs537038850 Chr.4: 158680511-158680511 on Build GRCh38
rs569257408 Chr.4: 158680531-158680531 on Build GRCh38
rs1013828853 Chr.4: 158680349-158680349 on Build GRCh38
rs952420339 Chr.4: 158680389-158680389 on Build GRCh38
rs903142172 Chr.4: 158680269-158680269 on Build GRCh38
rs780991832 Chr.4: 158680493-158680493 on Build GRCh38
rs761186585 Chr.4: 158680570-158680570 on Build GRCh38
rs550870904 Chr.4: 158680383-158680383 on Build GRCh38
rs759903159 Chr.4: 158680435-158680435 on Build GRCh38
rs768739704 Chr.4: 158680464-158680465 on Build GRCh38
rs746966542 Chr.4: 158680470-158680470 on Build GRCh38
rs1012681048 Chr.4: 158680342-158680342 on Build GRCh38
rs569876455 Chr.4: 158680386-158680386 on Build GRCh38
rs187248590 Chr.4: 158680583-158680583 on Build GRCh38
rs76669690 Chr.4: 158680612-158680612 on Build GRCh38
rs954937309 Chr.4: 158680449-158680449 on Build GRCh38
rs773668457 Chr.4: 158680553-158680553 on Build GRCh38
rs896346722 Chr.4: 158680317-158680317 on Build GRCh38
rs1006186398 Chr.4: 158680374-158680374 on Build GRCh38
rs958759984 Chr.4: 158680338-158680338 on Build GRCh38
rs938629880 Chr.4: 158680619-158680619 on Build GRCh38
rs1026438581 Chr.4: 158680362-158680362 on Build GRCh38
rs778154689 Chr.4: 158680625-158680625 on Build GRCh38
rs924962456 Chr.4: 158680571-158680571 on Build GRCh38
rs566605780 Chr.4: 158680495-158680495 on Build GRCh38
rs572956173 Chr.4: 158680629-158680629 on Build GRCh38
rs529832630 Chr.4: 158680452-158680452 on Build GRCh38
rs751894183 Chr.4: 158680440-158680442 on Build GRCh38
rs796051961 Chr.4: 158680608-158680608 on Build GRCh38
rs567085256 Chr.4: 158680451-158680451 on Build GRCh38
rs11559290 Chr.4: 158680524-158680524 on Build GRCh38
rs140731939 Chr.4: 158680540-158680540 on Build GRCh38
rs932843866 Chr.4: 158680499-158680499 on Build GRCh38
rs752167194 Chr.4: 158680416-158680416 on Build GRCh38
rs1013187846 Chr.4: 158680647-158680647 on Build GRCh38
rs748975378 Chr.4: 158680644-158680644 on Build GRCh38
rs937646249 Chr.4: 158680580-158680580 on Build GRCh38

View Full Product Details