Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168216105-168216612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX60
Entrez Gene ID
55601
Gene Name
DEXD/H-box helicase 60
Gene Aliases
-
UniGene
Hs.591710
Gene Chromosome Location
Chr.4: 168216291-168318807 on Build GRCh38
Transcripts
NM_017631.5

SNP Detail

SNP ID Chromosome Location
rs182991002 Chr.4: 168216134-168216134 on Build GRCh38
rs950454534 Chr.4: 168216505-168216505 on Build GRCh38
rs923998227 Chr.4: 168216307-168216307 on Build GRCh38
rs1000103846 Chr.4: 168216135-168216135 on Build GRCh38
rs916397985 Chr.4: 168216217-168216217 on Build GRCh38
rs545199670 Chr.4: 168216401-168216401 on Build GRCh38
rs927911012 Chr.4: 168216204-168216204 on Build GRCh38
rs917037386 Chr.4: 168216374-168216374 on Build GRCh38
rs560285388 Chr.4: 168216419-168216419 on Build GRCh38
rs991146947 Chr.4: 168216582-168216582 on Build GRCh38
rs527991512 Chr.4: 168216430-168216430 on Build GRCh38
rs562830726 Chr.4: 168216335-168216335 on Build GRCh38
rs746292970 Chr.4: 168216587-168216587 on Build GRCh38
rs573930158 Chr.4: 168216176-168216176 on Build GRCh38
rs375430839 Chr.4: 168216383-168216383 on Build GRCh38
rs950402500 Chr.4: 168216167-168216167 on Build GRCh38
rs996906454 Chr.4: 168216556-168216556 on Build GRCh38
rs1028928454 Chr.4: 168216558-168216558 on Build GRCh38
rs1028476645 Chr.4: 168216277-168216277 on Build GRCh38
rs901280838 Chr.4: 168216553-168216553 on Build GRCh38
rs960896322 Chr.4: 168216211-168216211 on Build GRCh38
rs1009604435 Chr.4: 168216473-168216473 on Build GRCh38
rs992447652 Chr.4: 168216359-168216359 on Build GRCh38
rs891409855 Chr.4: 168216365-168216365 on Build GRCh38
rs1007549564 Chr.4: 168216399-168216399 on Build GRCh38
rs990704252 Chr.4: 168216214-168216214 on Build GRCh38
rs139771533 Chr.4: 168216435-168216435 on Build GRCh38
rs549711813 Chr.4: 168216516-168216516 on Build GRCh38
rs10975 Chr.4: 168216436-168216436 on Build GRCh38
rs369983691 Chr.4: 168216561-168216561 on Build GRCh38
rs994547016 Chr.4: 168216141-168216141 on Build GRCh38
rs1043846245 Chr.4: 168216304-168216304 on Build GRCh38
rs1009655167 Chr.4: 168216201-168216201 on Build GRCh38
rs777928174 Chr.4: 168216403-168216403 on Build GRCh38
rs544740218 Chr.4: 168216294-168216294 on Build GRCh38
rs533423277 Chr.4: 168216342-168216342 on Build GRCh38
rs946532711 Chr.4: 168216281-168216281 on Build GRCh38
rs948457028 Chr.4: 168216386-168216386 on Build GRCh38
rs997055504 Chr.4: 168216253-168216253 on Build GRCh38
rs1027328410 Chr.4: 168216160-168216160 on Build GRCh38
rs186667381 Chr.4: 168216173-168216173 on Build GRCh38
rs1035090861 Chr.4: 168216531-168216531 on Build GRCh38
rs531204657 Chr.4: 168216470-168216470 on Build GRCh38
rs925778503 Chr.4: 168216404-168216404 on Build GRCh38

View Full Product Details