Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 173526799-173527296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAND2
Entrez Gene ID
9464
Gene Name
heart and neural crest derivatives expressed 2
Gene Aliases
DHAND2, Hed, Thing2, bHLHa26, dHand
UniGene
Hs.388245
Gene Chromosome Location
Chr.4: 173526501-173530227 on Build GRCh38
Transcripts
NM_021973.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6569167 c.640G>C p.E214Q Chr.4: 173527291-173527291 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773683304 Chr.4: 173527202-173527202 on Build GRCh38
rs1046485086 Chr.4: 173527013-173527013 on Build GRCh38
rs200090785 Chr.4: 173527260-173527260 on Build GRCh38
rs770141985 Chr.4: 173527267-173527269 on Build GRCh38
rs186309406 Chr.4: 173527234-173527234 on Build GRCh38
rs747041178 Chr.4: 173527190-173527190 on Build GRCh38
rs549352518 Chr.4: 173527183-173527183 on Build GRCh38
rs781057263 Chr.4: 173527070-173527070 on Build GRCh38
rs1031031775 Chr.4: 173526920-173526920 on Build GRCh38
rs1046281990 Chr.4: 173527048-173527048 on Build GRCh38
rs535075696 Chr.4: 173526944-173526944 on Build GRCh38
rs773256616 Chr.4: 173527073-173527073 on Build GRCh38
rs957979380 Chr.4: 173526855-173526855 on Build GRCh38
rs373950315 Chr.4: 173527228-173527228 on Build GRCh38
rs982965349 Chr.4: 173527181-173527181 on Build GRCh38
rs79766724 Chr.4: 173527252-173527252 on Build GRCh38
rs768929734 Chr.4: 173526980-173526980 on Build GRCh38
rs963812160 Chr.4: 173527114-173527114 on Build GRCh38
rs752774713 Chr.4: 173527081-173527081 on Build GRCh38
rs906404242 Chr.4: 173527066-173527066 on Build GRCh38
rs989882574 Chr.4: 173526946-173526946 on Build GRCh38
rs59706979 Chr.4: 173527268-173527268 on Build GRCh38
rs748610542 Chr.4: 173527265-173527266 on Build GRCh38
rs772042494 Chr.4: 173526896-173526896 on Build GRCh38
rs759510645 Chr.4: 173527184-173527184 on Build GRCh38
rs941716799 Chr.4: 173526998-173526998 on Build GRCh38
rs748418989 Chr.4: 173527195-173527195 on Build GRCh38
rs543928823 Chr.4: 173526957-173526957 on Build GRCh38
rs778531737 Chr.4: 173527250-173527251 on Build GRCh38
rs373720948 Chr.4: 173527240-173527240 on Build GRCh38
rs746694301 Chr.4: 173526876-173526876 on Build GRCh38
rs749290649 Chr.4: 173526887-173526887 on Build GRCh38
rs145372456 Chr.4: 173526991-173526991 on Build GRCh38
rs202001355 Chr.4: 173527276-173527276 on Build GRCh38
rs748365783 Chr.4: 173527247-173527247 on Build GRCh38
rs564671273 Chr.4: 173527018-173527018 on Build GRCh38
rs1000057185 Chr.4: 173527051-173527051 on Build GRCh38
rs139852692 Chr.4: 173526877-173526877 on Build GRCh38
rs976993195 Chr.4: 173526972-173526972 on Build GRCh38
rs375061640 Chr.4: 173527020-173527020 on Build GRCh38
rs774861958 Chr.4: 173527218-173527218 on Build GRCh38
rs771251278 Chr.4: 173527264-173527264 on Build GRCh38
rs754514186 Chr.4: 173527124-173527124 on Build GRCh38
rs777159348 Chr.4: 173527264-173527266 on Build GRCh38
rs757033397 Chr.4: 173527234-173527234 on Build GRCh38
rs752606032 Chr.4: 173527069-173527070 on Build GRCh38
rs370677207 Chr.4: 173527217-173527217 on Build GRCh38
rs777861282 Chr.4: 173527257-173527257 on Build GRCh38
rs781487306 Chr.4: 173527243-173527243 on Build GRCh38
rs563513362 Chr.4: 173526969-173526969 on Build GRCh38
rs755875700 Chr.4: 173527152-173527152 on Build GRCh38
rs942282473 Chr.4: 173526990-173526990 on Build GRCh38
rs895052855 Chr.4: 173526849-173526849 on Build GRCh38
rs749172717 Chr.4: 173527173-173527173 on Build GRCh38
rs1016205913 Chr.4: 173526836-173526836 on Build GRCh38
rs577670901 Chr.4: 173526902-173526902 on Build GRCh38
rs771732687 Chr.4: 173527251-173527252 on Build GRCh38
rs529454991 Chr.4: 173527165-173527165 on Build GRCh38
rs778716646 Chr.4: 173527189-173527189 on Build GRCh38
rs763671620 Chr.4: 173527233-173527233 on Build GRCh38
rs552630815 Chr.4: 173526865-173526865 on Build GRCh38
rs1037766874 Chr.4: 173527005-173527005 on Build GRCh38
rs1018051648 Chr.4: 173526888-173526888 on Build GRCh38
rs777442218 Chr.4: 173527110-173527110 on Build GRCh38
rs75706356 Chr.4: 173527226-173527226 on Build GRCh38
rs958053406 Chr.4: 173526933-173526934 on Build GRCh38
rs747102620 Chr.4: 173526966-173526966 on Build GRCh38
rs773506964 Chr.4: 173527266-173527267 on Build GRCh38
rs997977100 Chr.4: 173527121-173527121 on Build GRCh38
rs775701161 Chr.4: 173526901-173526901 on Build GRCh38
rs763240197 Chr.4: 173527214-173527214 on Build GRCh38
rs374459506 Chr.4: 173527227-173527227 on Build GRCh38
rs894777231 Chr.4: 173527011-173527011 on Build GRCh38
rs935197018 Chr.4: 173527255-173527255 on Build GRCh38
rs770047887 Chr.4: 173527201-173527201 on Build GRCh38
rs951080283 Chr.4: 173526953-173526953 on Build GRCh38
rs768876718 Chr.4: 173527194-173527194 on Build GRCh38
rs752396649 Chr.4: 173527150-173527150 on Build GRCh38
rs560557491 Chr.4: 173527125-173527125 on Build GRCh38
rs879129778 Chr.4: 173527055-173527055 on Build GRCh38
rs774810701 Chr.4: 173527270-173527270 on Build GRCh38
rs1012594633 Chr.4: 173527024-173527024 on Build GRCh38
rs767110094 Chr.4: 173527127-173527127 on Build GRCh38
rs267600084 Chr.4: 173527236-173527236 on Build GRCh38
rs751706907 Chr.4: 173526934-173526934 on Build GRCh38
rs1029946645 Chr.4: 173527131-173527131 on Build GRCh38
rs928157188 Chr.4: 173527244-173527244 on Build GRCh38
rs748952880 Chr.4: 173527147-173527156 on Build GRCh38
rs745417688 Chr.4: 173527252-173527254 on Build GRCh38
rs1031296169 Chr.4: 173526830-173526830 on Build GRCh38
rs544592345 Chr.4: 173526997-173526997 on Build GRCh38
rs964056062 Chr.4: 173527203-173527203 on Build GRCh38
rs10024737 Chr.4: 173526992-173526992 on Build GRCh38
rs533234480 Chr.4: 173526828-173526828 on Build GRCh38
rs879069138 Chr.4: 173527206-173527206 on Build GRCh38
rs1052438981 Chr.4: 173527070-173527070 on Build GRCh38
rs760105729 Chr.4: 173527272-173527272 on Build GRCh38
rs578094346 Chr.4: 173527042-173527042 on Build GRCh38
rs759026741 Chr.4: 173527220-173527220 on Build GRCh38
rs1018159376 Chr.4: 173527092-173527092 on Build GRCh38
rs529335909 Chr.4: 173526987-173526987 on Build GRCh38
rs371100365 Chr.4: 173527237-173527237 on Build GRCh38
rs755075499 Chr.4: 173527242-173527242 on Build GRCh38
rs751537957 Chr.4: 173527267-173527267 on Build GRCh38
rs147672639 Chr.4: 173527266-173527266 on Build GRCh38
rs181585513 Chr.4: 173527079-173527079 on Build GRCh38
rs778857884 Chr.4: 173527235-173527235 on Build GRCh38
rs754881292 Chr.4: 173526966-173526966 on Build GRCh38
rs77946068 Chr.4: 173527274-173527274 on Build GRCh38
rs767055146 Chr.4: 173527222-173527222 on Build GRCh38
rs765319090 Chr.4: 173527033-173527033 on Build GRCh38

View Full Product Details