Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183905274-183905772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STOX2
Entrez Gene ID
56977
Gene Name
storkhead box 2
Gene Aliases
-
UniGene
Hs.21958
Gene Chromosome Location
Chr.4: 183796331-184023530 on Build GRCh38
Transcripts
NM_020225.2 NR_132761.1 XM_011532129.1 XM_011532130.1 XM_017008466.1

SNP Detail

SNP ID Chromosome Location
rs1046949494 Chr.4: 183905478-183905478 on Build GRCh38
rs989769510 Chr.4: 183905680-183905680 on Build GRCh38
rs533394863 Chr.4: 183905504-183905504 on Build GRCh38
rs535952066 Chr.4: 183905678-183905678 on Build GRCh38
rs992111055 Chr.4: 183905347-183905347 on Build GRCh38
rs372128504 Chr.4: 183905560-183905560 on Build GRCh38
rs1030524517 Chr.4: 183905580-183905580 on Build GRCh38
rs954468401 Chr.4: 183905584-183905584 on Build GRCh38
rs984872922 Chr.4: 183905707-183905707 on Build GRCh38
rs754646260 Chr.4: 183905475-183905475 on Build GRCh38
rs886245757 Chr.4: 183905516-183905516 on Build GRCh38
rs1031176159 Chr.4: 183905653-183905653 on Build GRCh38
rs940686217 Chr.4: 183905461-183905461 on Build GRCh38
rs940695973 Chr.4: 183905713-183905713 on Build GRCh38
rs924213171 Chr.4: 183905435-183905435 on Build GRCh38
rs1030749987 Chr.4: 183905575-183905575 on Build GRCh38
rs980981955 Chr.4: 183905363-183905363 on Build GRCh38
rs549894988 Chr.4: 183905517-183905517 on Build GRCh38
rs1049118187 Chr.4: 183905417-183905417 on Build GRCh38
rs969684901 Chr.4: 183905612-183905612 on Build GRCh38
rs948062767 Chr.4: 183905714-183905714 on Build GRCh38
rs1024907018 Chr.4: 183905606-183905606 on Build GRCh38
rs936918847 Chr.4: 183905445-183905445 on Build GRCh38
rs978214955 Chr.4: 183905643-183905643 on Build GRCh38
rs1047574140 Chr.4: 183905507-183905507 on Build GRCh38
rs747695809 Chr.4: 183905740-183905740 on Build GRCh38
rs969252011 Chr.4: 183905361-183905361 on Build GRCh38
rs980649586 Chr.4: 183905656-183905656 on Build GRCh38
rs34792871 Chr.4: 183905482-183905483 on Build GRCh38
rs996367199 Chr.4: 183905487-183905487 on Build GRCh38
rs906602599 Chr.4: 183905500-183905500 on Build GRCh38
rs183274100 Chr.4: 183905701-183905701 on Build GRCh38
rs879848014 Chr.4: 183905642-183905642 on Build GRCh38
rs995791021 Chr.4: 183905574-183905574 on Build GRCh38
rs149626442 Chr.4: 183905585-183905585 on Build GRCh38
rs561540380 Chr.4: 183905301-183905301 on Build GRCh38
rs907959768 Chr.4: 183905710-183905710 on Build GRCh38
rs955080402 Chr.4: 183905371-183905371 on Build GRCh38
rs570770043 Chr.4: 183905496-183905496 on Build GRCh38
rs77422896 Chr.4: 183905411-183905411 on Build GRCh38
rs964315270 Chr.4: 183905316-183905316 on Build GRCh38
rs917837119 Chr.4: 183905354-183905354 on Build GRCh38
rs919779680 Chr.4: 183905676-183905676 on Build GRCh38
rs114309272 Chr.4: 183905339-183905339 on Build GRCh38
rs903620015 Chr.4: 183905520-183905520 on Build GRCh38
rs780799702 Chr.4: 183905588-183905588 on Build GRCh38
rs916586967 Chr.4: 183905460-183905460 on Build GRCh38
rs1000548393 Chr.4: 183905303-183905303 on Build GRCh38
rs1013219543 Chr.4: 183905592-183905592 on Build GRCh38
rs930700680 Chr.4: 183905378-183905378 on Build GRCh38
rs958152584 Chr.4: 183905654-183905654 on Build GRCh38
rs900219247 Chr.4: 183905563-183905563 on Build GRCh38
rs989696294 Chr.4: 183905447-183905447 on Build GRCh38
rs974469893 Chr.4: 183905319-183905319 on Build GRCh38
rs948032912 Chr.4: 183905465-183905465 on Build GRCh38

View Full Product Details