Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 1243443-1243935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC6A18
Entrez Gene ID
348932
Gene Name
solute carrier family 6 member 18
Gene Aliases
Xtrp2
UniGene
Hs.213284
Gene Chromosome Location
Chr.5: 1225355-1246189 on Build GRCh38
Transcripts
NM_182632.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7390746 c.1335T>C p.T445T Chr.5: 1243758-1243758 on Build GRCh38
3135431 c.1243C>T p.L415L Chr.5: 1243666-1243666 on Build GRCh38
3608071 c.1257C>T p.F419F Chr.5: 1243680-1243680 on Build GRCh38
3135426 c.1162G>A p.V388I Chr.5: 1243585-1243585 on Build GRCh38
3608070 c.1198C>T p.P400S Chr.5: 1243621-1243621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185631475 Chr.5: 1243854-1243854 on Build GRCh38
rs753427597 Chr.5: 1243583-1243583 on Build GRCh38
rs371964521 Chr.5: 1243777-1243777 on Build GRCh38
rs760354145 Chr.5: 1243526-1243526 on Build GRCh38
rs948520092 Chr.5: 1243542-1243542 on Build GRCh38
rs142581804 Chr.5: 1243650-1243650 on Build GRCh38
rs751980651 Chr.5: 1243550-1243550 on Build GRCh38
rs181138957 Chr.5: 1243824-1243824 on Build GRCh38
rs372157012 Chr.5: 1243798-1243798 on Build GRCh38
rs752302912 Chr.5: 1243524-1243524 on Build GRCh38
rs371070285 Chr.5: 1243741-1243741 on Build GRCh38
rs754073687 Chr.5: 1243474-1243474 on Build GRCh38
rs925776606 Chr.5: 1243646-1243646 on Build GRCh38
rs201512302 Chr.5: 1243687-1243687 on Build GRCh38
rs1000921862 Chr.5: 1243546-1243546 on Build GRCh38
rs150034651 Chr.5: 1243573-1243573 on Build GRCh38
rs746920363 Chr.5: 1243694-1243694 on Build GRCh38
rs371771771 Chr.5: 1243600-1243600 on Build GRCh38
rs757641266 Chr.5: 1243568-1243569 on Build GRCh38
rs986332485 Chr.5: 1243475-1243475 on Build GRCh38
rs370746444 Chr.5: 1243521-1243521 on Build GRCh38
rs745545985 Chr.5: 1243723-1243723 on Build GRCh38
rs111236460 Chr.5: 1243551-1243551 on Build GRCh38
rs763225212 Chr.5: 1243810-1243810 on Build GRCh38
rs149391939 Chr.5: 1243716-1243716 on Build GRCh38
rs756656750 Chr.5: 1243670-1243670 on Build GRCh38
rs982225950 Chr.5: 1243900-1243900 on Build GRCh38
rs778524616 Chr.5: 1243614-1243614 on Build GRCh38
rs200639430 Chr.5: 1243535-1243535 on Build GRCh38
rs534829526 Chr.5: 1243562-1243562 on Build GRCh38
rs555658261 Chr.5: 1243705-1243705 on Build GRCh38
rs774138692 Chr.5: 1243514-1243514 on Build GRCh38
rs201305109 Chr.5: 1243522-1243522 on Build GRCh38
rs75434344 Chr.5: 1243735-1243735 on Build GRCh38
rs912452437 Chr.5: 1243567-1243567 on Build GRCh38
rs768735435 Chr.5: 1243695-1243695 on Build GRCh38
rs768414081 Chr.5: 1243812-1243812 on Build GRCh38
rs141226479 Chr.5: 1243625-1243625 on Build GRCh38
rs748920454 Chr.5: 1243778-1243778 on Build GRCh38
rs770840769 Chr.5: 1243511-1243511 on Build GRCh38
rs761033923 Chr.5: 1243792-1243792 on Build GRCh38
rs116761974 Chr.5: 1243601-1243601 on Build GRCh38
rs868575398 Chr.5: 1243867-1243867 on Build GRCh38
rs766711024 Chr.5: 1243811-1243811 on Build GRCh38
rs750642461 Chr.5: 1243744-1243744 on Build GRCh38
rs754802241 Chr.5: 1243765-1243765 on Build GRCh38
rs745591980 Chr.5: 1243686-1243686 on Build GRCh38
rs373783511 Chr.5: 1243728-1243728 on Build GRCh38
rs770624023 Chr.5: 1243564-1243564 on Build GRCh38
rs762267341 Chr.5: 1243796-1243796 on Build GRCh38
rs776252759 Chr.5: 1243632-1243632 on Build GRCh38
rs138053797 Chr.5: 1243645-1243645 on Build GRCh38
rs913937756 Chr.5: 1243820-1243820 on Build GRCh38
rs574913359 Chr.5: 1243618-1243618 on Build GRCh38
rs965264111 Chr.5: 1243661-1243661 on Build GRCh38
rs754142878 Chr.5: 1243509-1243510 on Build GRCh38
rs899421280 Chr.5: 1243870-1243870 on Build GRCh38
rs765299867 Chr.5: 1243742-1243742 on Build GRCh38
rs138520447 Chr.5: 1243891-1243891 on Build GRCh38
rs780033519 Chr.5: 1243691-1243691 on Build GRCh38
rs373175159 Chr.5: 1243543-1243543 on Build GRCh38
rs368760212 Chr.5: 1243717-1243717 on Build GRCh38
rs199865996 Chr.5: 1243568-1243568 on Build GRCh38
rs775794533 Chr.5: 1243785-1243785 on Build GRCh38
rs201446388 Chr.5: 1243701-1243701 on Build GRCh38
rs114952930 Chr.5: 1243633-1243633 on Build GRCh38
rs761123823 Chr.5: 1243720-1243720 on Build GRCh38
rs973036624 Chr.5: 1243502-1243502 on Build GRCh38
rs35948576 Chr.5: 1243752-1243752 on Build GRCh38
rs749443178 Chr.5: 1243613-1243613 on Build GRCh38
rs199801467 Chr.5: 1243644-1243644 on Build GRCh38
rs151263479 Chr.5: 1243563-1243563 on Build GRCh38
rs769268784 Chr.5: 1243711-1243711 on Build GRCh38
rs780535792 Chr.5: 1243544-1243544 on Build GRCh38
rs139307806 Chr.5: 1243569-1243569 on Build GRCh38
rs546803554 Chr.5: 1243793-1243793 on Build GRCh38
rs113079314 Chr.5: 1243879-1243879 on Build GRCh38
rs756176531 Chr.5: 1243611-1243611 on Build GRCh38
rs775100753 Chr.5: 1243581-1243581 on Build GRCh38
rs74581452 Chr.5: 1243593-1243593 on Build GRCh38
rs367900698 Chr.5: 1243599-1243599 on Build GRCh38
rs779067358 Chr.5: 1243611-1243611 on Build GRCh38
rs758741125 Chr.5: 1243746-1243746 on Build GRCh38
rs758479180 Chr.5: 1243748-1243749 on Build GRCh38
rs529556547 Chr.5: 1243488-1243488 on Build GRCh38
rs768249532 Chr.5: 1243630-1243630 on Build GRCh38
rs943389970 Chr.5: 1243855-1243855 on Build GRCh38
rs35661976 Chr.5: 1243584-1243584 on Build GRCh38
rs79457861 Chr.5: 1243536-1243536 on Build GRCh38
rs377325729 Chr.5: 1243681-1243681 on Build GRCh38
rs147728672 Chr.5: 1243582-1243582 on Build GRCh38
rs143776496 Chr.5: 1243776-1243776 on Build GRCh38
rs1026499420 Chr.5: 1243912-1243912 on Build GRCh38
rs147961181 Chr.5: 1243592-1243592 on Build GRCh38
rs772068090 Chr.5: 1243621-1243621 on Build GRCh38
rs781203007 Chr.5: 1243553-1243553 on Build GRCh38
rs370591425 Chr.5: 1243671-1243671 on Build GRCh38
rs746964531 Chr.5: 1243804-1243807 on Build GRCh38
rs568177053 Chr.5: 1243819-1243819 on Build GRCh38
rs550462743 Chr.5: 1243845-1243845 on Build GRCh38
rs908390656 Chr.5: 1243886-1243886 on Build GRCh38
rs750552565 Chr.5: 1243808-1243808 on Build GRCh38
rs377615252 Chr.5: 1243751-1243751 on Build GRCh38
rs775884526 Chr.5: 1243718-1243718 on Build GRCh38
rs753201844 Chr.5: 1243664-1243664 on Build GRCh38
rs374489811 Chr.5: 1243707-1243707 on Build GRCh38
rs748262389 Chr.5: 1243557-1243557 on Build GRCh38
rs945500754 Chr.5: 1243833-1243833 on Build GRCh38
rs910269365 Chr.5: 1243493-1243493 on Build GRCh38
rs149337877 Chr.5: 1243758-1243758 on Build GRCh38
rs973130185 Chr.5: 1243598-1243598 on Build GRCh38
rs199723497 Chr.5: 1243653-1243653 on Build GRCh38
rs375378208 Chr.5: 1243779-1243779 on Build GRCh38
rs767932255 Chr.5: 1243658-1243658 on Build GRCh38
rs146896608 Chr.5: 1243585-1243585 on Build GRCh38
rs4994840 Chr.5: 1243732-1243732 on Build GRCh38
rs746113206 Chr.5: 1243718-1243719 on Build GRCh38
rs551021545 Chr.5: 1243505-1243505 on Build GRCh38
rs372900404 Chr.5: 1243676-1243676 on Build GRCh38
rs141229418 Chr.5: 1243493-1243494 on Build GRCh38
rs908601746 Chr.5: 1243697-1243697 on Build GRCh38
rs576827891 Chr.5: 1243472-1243472 on Build GRCh38
rs111592068 Chr.5: 1243823-1243823 on Build GRCh38
rs148220007 Chr.5: 1243612-1243612 on Build GRCh38
rs979972205 Chr.5: 1243791-1243791 on Build GRCh38
rs1028241112 Chr.5: 1243863-1243863 on Build GRCh38
rs781198609 Chr.5: 1243775-1243775 on Build GRCh38
rs1030606866 Chr.5: 1243463-1243463 on Build GRCh38
rs755129926 Chr.5: 1243494-1243494 on Build GRCh38
rs780052917 Chr.5: 1243760-1243761 on Build GRCh38
rs757382342 Chr.5: 1243680-1243680 on Build GRCh38

View Full Product Details