Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10254006-10254502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT5
Entrez Gene ID
22948
Gene Name
chaperonin containing TCP1 subunit 5
Gene Aliases
CCT-epsilon, CCTE, HEL-S-69, PNAS-102, TCP-1-epsilon
UniGene
Hs.1600
Gene Chromosome Location
Chr.5: 10249921-10266412 on Build GRCh38
Transcripts
NM_001306153.1 NM_001306154.1 NM_001306155.1 NM_001306156.1 NM_012073.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6863508 c.111T>G p.H37Q Chr.5: 10254150-10254150 on Build GRCh38
4791503 c.133G>A p.A45T Chr.5: 10254172-10254172 on Build GRCh38
3776093 c.150A>C p.T50T Chr.5: 10254189-10254189 on Build GRCh38
1742949 c.159A>G p.G53G Chr.5: 10254198-10254198 on Build GRCh38
1059304 c.113T>C p.I38T Chr.5: 10254152-10254152 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201641048 Chr.5: 10254139-10254139 on Build GRCh38
rs200807677 Chr.5: 10254224-10254224 on Build GRCh38
rs540664122 Chr.5: 10254462-10254462 on Build GRCh38
rs935593997 Chr.5: 10254079-10254079 on Build GRCh38
rs199849658 Chr.5: 10254255-10254255 on Build GRCh38
rs957120124 Chr.5: 10254478-10254478 on Build GRCh38
rs943842372 Chr.5: 10254298-10254298 on Build GRCh38
rs745860397 Chr.5: 10254199-10254199 on Build GRCh38
rs781397784 Chr.5: 10254121-10254121 on Build GRCh38
rs897035142 Chr.5: 10254321-10254321 on Build GRCh38
rs773556943 Chr.5: 10254213-10254213 on Build GRCh38
rs1012231005 Chr.5: 10254466-10254466 on Build GRCh38
rs369064257 Chr.5: 10254223-10254223 on Build GRCh38
rs758147072 Chr.5: 10254040-10254040 on Build GRCh38
rs779761046 Chr.5: 10254117-10254117 on Build GRCh38
rs556294617 Chr.5: 10254252-10254252 on Build GRCh38
rs869142760 Chr.5: 10254408-10254409 on Build GRCh38
rs758769591 Chr.5: 10254095-10254095 on Build GRCh38
rs931218312 Chr.5: 10254310-10254310 on Build GRCh38
rs57187607 Chr.5: 10254409-10254409 on Build GRCh38
rs1032254297 Chr.5: 10254409-10254409 on Build GRCh38
rs527286294 Chr.5: 10254371-10254371 on Build GRCh38
rs771901324 Chr.5: 10254202-10254202 on Build GRCh38
rs544661811 Chr.5: 10254327-10254327 on Build GRCh38
rs912628844 Chr.5: 10254241-10254241 on Build GRCh38
rs748160823 Chr.5: 10254124-10254124 on Build GRCh38
rs1040850940 Chr.5: 10254307-10254307 on Build GRCh38
rs774203439 Chr.5: 10254050-10254051 on Build GRCh38
rs1027346918 Chr.5: 10254388-10254388 on Build GRCh38
rs888091635 Chr.5: 10254406-10254407 on Build GRCh38
rs986865707 Chr.5: 10254240-10254240 on Build GRCh38
rs1037236407 Chr.5: 10254264-10254264 on Build GRCh38
rs999286656 Chr.5: 10254389-10254389 on Build GRCh38
rs770210482 Chr.5: 10254136-10254137 on Build GRCh38
rs763214622 Chr.5: 10254246-10254247 on Build GRCh38
rs1054999657 Chr.5: 10254347-10254347 on Build GRCh38
rs755408043 Chr.5: 10254115-10254115 on Build GRCh38
rs1041756595 Chr.5: 10254390-10254390 on Build GRCh38
rs932669924 Chr.5: 10254157-10254157 on Build GRCh38
rs1012345238 Chr.5: 10254412-10254412 on Build GRCh38
rs902237773 Chr.5: 10254387-10254387 on Build GRCh38
rs1023355730 Chr.5: 10254413-10254413 on Build GRCh38
rs752159897 Chr.5: 10254107-10254107 on Build GRCh38
rs982284343 Chr.5: 10254034-10254034 on Build GRCh38
rs564640942 Chr.5: 10254348-10254348 on Build GRCh38
rs142638550 Chr.5: 10254156-10254156 on Build GRCh38
rs767014834 Chr.5: 10254102-10254102 on Build GRCh38
rs370908686 Chr.5: 10254251-10254251 on Build GRCh38
rs879081858 Chr.5: 10254086-10254086 on Build GRCh38
rs746733570 Chr.5: 10254137-10254137 on Build GRCh38
rs970457917 Chr.5: 10254416-10254416 on Build GRCh38
rs766889699 Chr.5: 10254227-10254227 on Build GRCh38
rs777997985 Chr.5: 10254257-10254257 on Build GRCh38
rs751965723 Chr.5: 10254239-10254239 on Build GRCh38
rs1046060677 Chr.5: 10254385-10254385 on Build GRCh38
rs1050508166 Chr.5: 10254236-10254236 on Build GRCh38
rs762170913 Chr.5: 10254119-10254119 on Build GRCh38
rs898328854 Chr.5: 10254296-10254296 on Build GRCh38
rs766482931 Chr.5: 10254251-10254252 on Build GRCh38
rs775126012 Chr.5: 10254204-10254204 on Build GRCh38
rs754615590 Chr.5: 10254473-10254473 on Build GRCh38
rs770668105 Chr.5: 10254178-10254178 on Build GRCh38
rs778660679 Chr.5: 10254181-10254181 on Build GRCh38
rs1003333706 Chr.5: 10254419-10254421 on Build GRCh38
rs760145054 Chr.5: 10254245-10254245 on Build GRCh38
rs760046237 Chr.5: 10254216-10254216 on Build GRCh38

View Full Product Details