Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 31299497-31300040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH6
Entrez Gene ID
1004
Gene Name
cadherin 6
Gene Aliases
CAD6, KCAD
UniGene
Hs.124776
Gene Chromosome Location
Chr.5: 31193655-31329146 on Build GRCh38
Transcripts
NM_004932.3 XM_011513921.2 XM_017008910.1 XM_017008911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3615319 c.740G>A p.G247E Chr.5: 31299560-31299560 on Build GRCh38
70120 c.727G>A p.G243S Chr.5: 31299547-31299547 on Build GRCh38
482704 c.788A>T p.D263V Chr.5: 31299608-31299608 on Build GRCh38
1695527 c.806C>T p.P269L Chr.5: 31299626-31299626 on Build GRCh38
3615320 c.750G>A p.G250G Chr.5: 31299570-31299570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529891401 Chr.5: 31299527-31299527 on Build GRCh38
rs571023915 Chr.5: 31299659-31299659 on Build GRCh38
rs904794767 Chr.5: 31299732-31299732 on Build GRCh38
rs764989718 Chr.5: 31299957-31299957 on Build GRCh38
rs552235433 Chr.5: 31299803-31299803 on Build GRCh38
rs971892592 Chr.5: 31299966-31299966 on Build GRCh38
rs375949044 Chr.5: 31299570-31299570 on Build GRCh38
rs770016474 Chr.5: 31299664-31299664 on Build GRCh38
rs753390985 Chr.5: 31299606-31299606 on Build GRCh38
rs909455896 Chr.5: 31299914-31299914 on Build GRCh38
rs999832945 Chr.5: 31299774-31299774 on Build GRCh38
rs772550867 Chr.5: 31299666-31299666 on Build GRCh38
rs763196951 Chr.5: 31299537-31299537 on Build GRCh38
rs998808279 Chr.5: 31299624-31299624 on Build GRCh38
rs927285145 Chr.5: 31300001-31300001 on Build GRCh38
rs770944665 Chr.5: 31299523-31299523 on Build GRCh38
rs199901263 Chr.5: 31299620-31299620 on Build GRCh38
rs768323258 Chr.5: 31300000-31300000 on Build GRCh38
rs910351429 Chr.5: 31299668-31299668 on Build GRCh38
rs373866360 Chr.5: 31299663-31299663 on Build GRCh38
rs749485405 Chr.5: 31299681-31299681 on Build GRCh38
rs755804928 Chr.5: 31299585-31299585 on Build GRCh38
rs1018728657 Chr.5: 31299925-31299925 on Build GRCh38
rs755530248 Chr.5: 31299646-31299646 on Build GRCh38
rs776523565 Chr.5: 31299623-31299624 on Build GRCh38
rs529288494 Chr.5: 31299819-31299819 on Build GRCh38
rs938234077 Chr.5: 31299932-31299932 on Build GRCh38
rs774413104 Chr.5: 31299682-31299682 on Build GRCh38
rs532137627 Chr.5: 31299784-31299784 on Build GRCh38
rs1041974434 Chr.5: 31299725-31299725 on Build GRCh38
rs867578516 Chr.5: 31299525-31299525 on Build GRCh38
rs1014398832 Chr.5: 31299552-31299552 on Build GRCh38
rs745438339 Chr.5: 31299638-31299638 on Build GRCh38
rs138288285 Chr.5: 31299580-31299580 on Build GRCh38
rs756992324 Chr.5: 31299607-31299607 on Build GRCh38
rs1032928602 Chr.5: 31299627-31299627 on Build GRCh38
rs773652381 Chr.5: 31299679-31299679 on Build GRCh38
rs1027502469 Chr.5: 31299967-31299967 on Build GRCh38
rs141433876 Chr.5: 31299540-31299540 on Build GRCh38
rs888832107 Chr.5: 31299878-31299878 on Build GRCh38
rs1009059058 Chr.5: 31299885-31299885 on Build GRCh38
rs150406388 Chr.5: 31299546-31299546 on Build GRCh38
rs763953119 Chr.5: 31299586-31299586 on Build GRCh38
rs879555536 Chr.5: 31299993-31299993 on Build GRCh38
rs984101450 Chr.5: 31299988-31299988 on Build GRCh38
rs937393347 Chr.5: 31300003-31300003 on Build GRCh38
rs188602764 Chr.5: 31299828-31299828 on Build GRCh38
rs759433625 Chr.5: 31299553-31299553 on Build GRCh38
rs776063914 Chr.5: 31299734-31299734 on Build GRCh38
rs957219383 Chr.5: 31299639-31299639 on Build GRCh38
rs895980075 Chr.5: 31299642-31299642 on Build GRCh38
rs746667713 Chr.5: 31299823-31299823 on Build GRCh38
rs951973797 Chr.5: 31299971-31299971 on Build GRCh38
rs896610487 Chr.5: 31299963-31299963 on Build GRCh38

View Full Product Details