Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 32354566-32355111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFR
Entrez Gene ID
51663
Gene Name
zinc finger RNA binding protein
Gene Aliases
SPG71, ZFR1
UniGene
Hs.435231
Gene Chromosome Location
Chr.5: 32354350-32444738 on Build GRCh38
Transcripts
NM_016107.3

SNP Detail

SNP ID Chromosome Location
rs890194326 Chr.5: 32354722-32354722 on Build GRCh38
rs989378328 Chr.5: 32354611-32354611 on Build GRCh38
rs576668360 Chr.5: 32354618-32354619 on Build GRCh38
rs574839129 Chr.5: 32354771-32354771 on Build GRCh38
rs1051373594 Chr.5: 32355082-32355082 on Build GRCh38
rs766564169 Chr.5: 32354957-32354957 on Build GRCh38
rs534916797 Chr.5: 32354635-32354635 on Build GRCh38
rs750131610 Chr.5: 32354857-32354857 on Build GRCh38
rs112864921 Chr.5: 32354641-32354641 on Build GRCh38
rs913359348 Chr.5: 32354615-32354619 on Build GRCh38
rs980086384 Chr.5: 32354841-32354841 on Build GRCh38
rs554909788 Chr.5: 32354770-32354770 on Build GRCh38
rs143085858 Chr.5: 32354666-32354666 on Build GRCh38
rs764301146 Chr.5: 32354730-32354730 on Build GRCh38
rs768889023 Chr.5: 32354629-32354629 on Build GRCh38
rs932267076 Chr.5: 32355050-32355050 on Build GRCh38
rs758162976 Chr.5: 32354928-32354928 on Build GRCh38
rs192882845 Chr.5: 32354926-32354926 on Build GRCh38
rs1015091933 Chr.5: 32354757-32354757 on Build GRCh38
rs1051403740 Chr.5: 32354688-32354688 on Build GRCh38
rs960110030 Chr.5: 32354889-32354889 on Build GRCh38
rs56046182 Chr.5: 32354623-32354623 on Build GRCh38
rs968890424 Chr.5: 32354956-32354956 on Build GRCh38
rs913319538 Chr.5: 32354940-32354940 on Build GRCh38
rs555956487 Chr.5: 32355057-32355057 on Build GRCh38
rs534700306 Chr.5: 32354734-32354734 on Build GRCh38
rs566126173 Chr.5: 32354687-32354687 on Build GRCh38
rs1035729460 Chr.5: 32354886-32354886 on Build GRCh38
rs931628903 Chr.5: 32354674-32354674 on Build GRCh38
rs922172098 Chr.5: 32354972-32354972 on Build GRCh38
rs373926238 Chr.5: 32354694-32354694 on Build GRCh38
rs898966312 Chr.5: 32354760-32354760 on Build GRCh38
rs552883070 Chr.5: 32354682-32354682 on Build GRCh38
rs969318254 Chr.5: 32354802-32354802 on Build GRCh38

View Full Product Details