Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 32363904-32364403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFR
Entrez Gene ID
51663
Gene Name
zinc finger RNA binding protein
Gene Aliases
SPG71, ZFR1
UniGene
Hs.435231
Gene Chromosome Location
Chr.5: 32354350-32444738 on Build GRCh38
Transcripts
NM_016107.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3661787 c.2882C>T p.P961L Chr.5: 32364229-32364229 on Build GRCh38
1437045 c.2970T>C p.P990P Chr.5: 32364023-32364023 on Build GRCh38
6250051 c.2880C>A p.S960R Chr.5: 32364231-32364231 on Build GRCh38
1437046 c.2913T>C p.V971V Chr.5: 32364198-32364198 on Build GRCh38
4485731 c.2968C>T p.P990S Chr.5: 32364025-32364025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375469281 Chr.5: 32364036-32364036 on Build GRCh38
rs563902966 Chr.5: 32364008-32364008 on Build GRCh38
rs7721791 Chr.5: 32364081-32364081 on Build GRCh38
rs150661230 Chr.5: 32364249-32364249 on Build GRCh38
rs774586995 Chr.5: 32364160-32364160 on Build GRCh38
rs759887884 Chr.5: 32364180-32364180 on Build GRCh38
rs372448245 Chr.5: 32364031-32364031 on Build GRCh38
rs376915428 Chr.5: 32364029-32364029 on Build GRCh38
rs955004835 Chr.5: 32364357-32364357 on Build GRCh38
rs543530321 Chr.5: 32364000-32364000 on Build GRCh38
rs547262180 Chr.5: 32364067-32364067 on Build GRCh38
rs757648349 Chr.5: 32364126-32364135 on Build GRCh38
rs1009814892 Chr.5: 32364173-32364173 on Build GRCh38
rs763408972 Chr.5: 32364010-32364010 on Build GRCh38
rs186122587 Chr.5: 32364189-32364189 on Build GRCh38
rs779736725 Chr.5: 32364120-32364120 on Build GRCh38
rs749786578 Chr.5: 32364150-32364150 on Build GRCh38
rs746709163 Chr.5: 32364326-32364326 on Build GRCh38
rs1053530496 Chr.5: 32364333-32364333 on Build GRCh38
rs367873889 Chr.5: 32363973-32363973 on Build GRCh38
rs991501602 Chr.5: 32364155-32364155 on Build GRCh38
rs760949082 Chr.5: 32364198-32364198 on Build GRCh38
rs776101027 Chr.5: 32364148-32364148 on Build GRCh38
rs371003733 Chr.5: 32363963-32363963 on Build GRCh38
rs967925166 Chr.5: 32364224-32364224 on Build GRCh38
rs746655952 Chr.5: 32364124-32364124 on Build GRCh38
rs764290897 Chr.5: 32364216-32364216 on Build GRCh38
rs774984560 Chr.5: 32363959-32363959 on Build GRCh38
rs750312372 Chr.5: 32364266-32364266 on Build GRCh38
rs200618639 Chr.5: 32363976-32363976 on Build GRCh38
rs191031844 Chr.5: 32364325-32364325 on Build GRCh38
rs142124719 Chr.5: 32364083-32364083 on Build GRCh38
rs901057028 Chr.5: 32364367-32364367 on Build GRCh38
rs996528914 Chr.5: 32364238-32364238 on Build GRCh38
rs768138742 Chr.5: 32363974-32363974 on Build GRCh38
rs568077424 Chr.5: 32364108-32364108 on Build GRCh38
rs771713689 Chr.5: 32363939-32363939 on Build GRCh38
rs148538401 Chr.5: 32364242-32364242 on Build GRCh38
rs745608850 Chr.5: 32364089-32364089 on Build GRCh38
rs758254326 Chr.5: 32364297-32364297 on Build GRCh38
rs918144214 Chr.5: 32364156-32364157 on Build GRCh38
rs977691424 Chr.5: 32364210-32364210 on Build GRCh38
rs201002022 Chr.5: 32364132-32364132 on Build GRCh38
rs574361293 Chr.5: 32364275-32364275 on Build GRCh38
rs1045009052 Chr.5: 32364094-32364094 on Build GRCh38
rs548196626 Chr.5: 32364086-32364086 on Build GRCh38
rs202186346 Chr.5: 32364060-32364060 on Build GRCh38
rs936455649 Chr.5: 32364298-32364298 on Build GRCh38
rs367948960 Chr.5: 32364030-32364030 on Build GRCh38
rs767849865 Chr.5: 32363975-32363975 on Build GRCh38
rs768676427 Chr.5: 32364106-32364106 on Build GRCh38
rs779463055 Chr.5: 32364326-32364327 on Build GRCh38
rs889644045 Chr.5: 32364352-32364352 on Build GRCh38
rs1040997582 Chr.5: 32364360-32364360 on Build GRCh38
rs773771441 Chr.5: 32363995-32363995 on Build GRCh38
rs200215101 Chr.5: 32364026-32364026 on Build GRCh38
rs771722927 Chr.5: 32364116-32364116 on Build GRCh38
rs997490838 Chr.5: 32364051-32364051 on Build GRCh38
rs763490085 Chr.5: 32363996-32363996 on Build GRCh38
rs777997030 Chr.5: 32364068-32364068 on Build GRCh38
rs750868644 Chr.5: 32364355-32364355 on Build GRCh38
rs753913825 Chr.5: 32364228-32364228 on Build GRCh38

View Full Product Details