Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55232246-55232736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNO
Entrez Gene ID
10309
Gene Name
cyclin O
Gene Aliases
CCNU, CILD29, UDG2
UniGene
Hs.3041
Gene Chromosome Location
Chr.5: 55231152-55233717 on Build GRCh38
Transcripts
NM_021147.4 NR_125346.1 NR_125347.1 NR_125348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4402046 c.396C>T p.S132S Chr.5: 55232532-55232532 on Build GRCh38
738522 c.502A>T p.T168S Chr.5: 55232426-55232426 on Build GRCh38
7143237 c.447C>G p.L149L Chr.5: 55232481-55232481 on Build GRCh38
1068801 c.525C>A p.F175L Chr.5: 55232403-55232403 on Build GRCh38
4141999 c.433C>G p.R145G Chr.5: 55232495-55232495 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553049896 Chr.5: 55232553-55232553 on Build GRCh38
rs916873844 Chr.5: 55232307-55232307 on Build GRCh38
rs755937476 Chr.5: 55232437-55232437 on Build GRCh38
rs957579096 Chr.5: 55232707-55232707 on Build GRCh38
rs751000643 Chr.5: 55232427-55232427 on Build GRCh38
rs576286586 Chr.5: 55232571-55232571 on Build GRCh38
rs201505004 Chr.5: 55232589-55232589 on Build GRCh38
rs772531671 Chr.5: 55232587-55232587 on Build GRCh38
rs779079579 Chr.5: 55232386-55232386 on Build GRCh38
rs761359374 Chr.5: 55232543-55232543 on Build GRCh38
rs762886738 Chr.5: 55232335-55232336 on Build GRCh38
rs897613913 Chr.5: 55232288-55232288 on Build GRCh38
rs776757725 Chr.5: 55232595-55232595 on Build GRCh38
rs761762345 Chr.5: 55232469-55232469 on Build GRCh38
rs535119119 Chr.5: 55232446-55232446 on Build GRCh38
rs879101149 Chr.5: 55232309-55232309 on Build GRCh38
rs1056784047 Chr.5: 55232671-55232671 on Build GRCh38
rs587777503 Chr.5: 55232446-55232447 on Build GRCh38
rs368481974 Chr.5: 55232327-55232327 on Build GRCh38
rs766425941 Chr.5: 55232552-55232552 on Build GRCh38
rs201370811 Chr.5: 55232400-55232400 on Build GRCh38
rs776920818 Chr.5: 55232501-55232501 on Build GRCh38
rs992583778 Chr.5: 55232296-55232296 on Build GRCh38
rs771267355 Chr.5: 55232484-55232484 on Build GRCh38
rs774041124 Chr.5: 55232541-55232541 on Build GRCh38
rs763746503 Chr.5: 55232423-55232423 on Build GRCh38
rs748183136 Chr.5: 55232388-55232388 on Build GRCh38
rs769224375 Chr.5: 55232424-55232424 on Build GRCh38
rs374720682 Chr.5: 55232523-55232523 on Build GRCh38
rs894315363 Chr.5: 55232695-55232695 on Build GRCh38
rs780395694 Chr.5: 55232357-55232357 on Build GRCh38
rs771239201 Chr.5: 55232414-55232414 on Build GRCh38
rs1026116826 Chr.5: 55232644-55232644 on Build GRCh38
rs778868763 Chr.5: 55232425-55232425 on Build GRCh38
rs144371373 Chr.5: 55232698-55232698 on Build GRCh38
rs767005607 Chr.5: 55232546-55232546 on Build GRCh38
rs1048686695 Chr.5: 55232600-55232600 on Build GRCh38
rs779308069 Chr.5: 55232462-55232462 on Build GRCh38
rs958502668 Chr.5: 55232431-55232431 on Build GRCh38
rs199762121 Chr.5: 55232340-55232340 on Build GRCh38
rs994757037 Chr.5: 55232603-55232603 on Build GRCh38
rs368489305 Chr.5: 55232561-55232561 on Build GRCh38
rs767470748 Chr.5: 55232432-55232432 on Build GRCh38
rs970118116 Chr.5: 55232682-55232682 on Build GRCh38
rs529691602 Chr.5: 55232665-55232665 on Build GRCh38
rs766335987 Chr.5: 55232440-55232440 on Build GRCh38
rs910172212 Chr.5: 55232611-55232611 on Build GRCh38
rs1035517200 Chr.5: 55232404-55232404 on Build GRCh38
rs772590141 Chr.5: 55232397-55232397 on Build GRCh38
rs762648052 Chr.5: 55232422-55232422 on Build GRCh38
rs755915635 Chr.5: 55232351-55232351 on Build GRCh38
rs752394589 Chr.5: 55232312-55232312 on Build GRCh38
rs762414326 Chr.5: 55232313-55232313 on Build GRCh38
rs200976457 Chr.5: 55232569-55232569 on Build GRCh38
rs566922203 Chr.5: 55232714-55232715 on Build GRCh38
rs541131375 Chr.5: 55232649-55232649 on Build GRCh38
rs548514182 Chr.5: 55232283-55232283 on Build GRCh38
rs559076864 Chr.5: 55232653-55232653 on Build GRCh38
rs752527657 Chr.5: 55232503-55232503 on Build GRCh38
rs768844025 Chr.5: 55232368-55232368 on Build GRCh38
rs749589131 Chr.5: 55232358-55232358 on Build GRCh38
rs778310459 Chr.5: 55232466-55232466 on Build GRCh38
rs751483554 Chr.5: 55232326-55232326 on Build GRCh38
rs72749883 Chr.5: 55232560-55232560 on Build GRCh38
rs763656338 Chr.5: 55232319-55232319 on Build GRCh38
rs773613084 Chr.5: 55232399-55232399 on Build GRCh38
rs334892 Chr.5: 55232565-55232565 on Build GRCh38
rs926256890 Chr.5: 55232352-55232352 on Build GRCh38
rs570120073 Chr.5: 55232278-55232278 on Build GRCh38
rs770926618 Chr.5: 55232344-55232344 on Build GRCh38
rs747121961 Chr.5: 55232566-55232566 on Build GRCh38
rs750185563 Chr.5: 55232348-55232348 on Build GRCh38
rs530289717 Chr.5: 55232277-55232277 on Build GRCh38
rs143559138 Chr.5: 55232441-55232441 on Build GRCh38
rs748208556 Chr.5: 55232463-55232463 on Build GRCh38
rs537278082 Chr.5: 55232328-55232328 on Build GRCh38
rs758456393 Chr.5: 55232465-55232465 on Build GRCh38
rs764894902 Chr.5: 55232311-55232311 on Build GRCh38
rs371349055 Chr.5: 55232442-55232442 on Build GRCh38
rs771769193 Chr.5: 55232586-55232586 on Build GRCh38
rs773188175 Chr.5: 55232551-55232551 on Build GRCh38
rs532064458 Chr.5: 55232622-55232622 on Build GRCh38
rs573986503 Chr.5: 55232696-55232696 on Build GRCh38
rs1048656067 Chr.5: 55232274-55232274 on Build GRCh38
rs773349948 Chr.5: 55232311-55232313 on Build GRCh38
rs13169396 Chr.5: 55232447-55232447 on Build GRCh38
rs746232811 Chr.5: 55232503-55232503 on Build GRCh38
rs747816939 Chr.5: 55232576-55232576 on Build GRCh38
rs969770754 Chr.5: 55232336-55232336 on Build GRCh38
rs146796153 Chr.5: 55232525-55232525 on Build GRCh38
rs189447634 Chr.5: 55232669-55232669 on Build GRCh38
rs1051206782 Chr.5: 55232537-55232537 on Build GRCh38
rs775115664 Chr.5: 55232420-55232420 on Build GRCh38
rs199517268 Chr.5: 55232575-55232575 on Build GRCh38
rs750371928 Chr.5: 55232433-55232433 on Build GRCh38
rs926195687 Chr.5: 55232704-55232704 on Build GRCh38
rs569915958 Chr.5: 55232286-55232286 on Build GRCh38
rs747533134 Chr.5: 55232481-55232481 on Build GRCh38
rs761255457 Chr.5: 55232429-55232429 on Build GRCh38
rs1021501792 Chr.5: 55232486-55232486 on Build GRCh38
rs759515939 Chr.5: 55232414-55232417 on Build GRCh38
rs932915305 Chr.5: 55232273-55232273 on Build GRCh38
rs774393276 Chr.5: 55232389-55232390 on Build GRCh38

View Full Product Details