Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60644566-60645092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPDC1B
Entrez Gene ID
55789
Gene Name
DEP domain containing 1B
Gene Aliases
BRCC3, XTP1
UniGene
Hs.482233
Gene Chromosome Location
Chr.5: 60596912-60700166 on Build GRCh38
Transcripts
NM_001145208.1 NM_018369.2 XM_011543509.2 XM_011543510.2 XM_017009629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1069202 c.661G>A p.V221I Chr.5: 60644793-60644793 on Build GRCh38
738440 c.659A>T p.N220I Chr.5: 60644795-60644795 on Build GRCh38
4918180 c.636C>A p.V212V Chr.5: 60644818-60644818 on Build GRCh38
4904267 c.655C>T p.H219Y Chr.5: 60644799-60644799 on Build GRCh38
136482 c.605C>T p.S202F Chr.5: 60644849-60644849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545673848 Chr.5: 60644630-60644630 on Build GRCh38
rs963398823 Chr.5: 60644602-60644602 on Build GRCh38
rs757362322 Chr.5: 60644904-60644905 on Build GRCh38
rs779893453 Chr.5: 60644798-60644798 on Build GRCh38
rs753126020 Chr.5: 60644883-60644883 on Build GRCh38
rs756431781 Chr.5: 60644786-60644787 on Build GRCh38
rs769269437 Chr.5: 60644796-60644796 on Build GRCh38
rs749274404 Chr.5: 60644802-60644802 on Build GRCh38
rs890319687 Chr.5: 60644921-60644921 on Build GRCh38
rs183114104 Chr.5: 60644936-60644936 on Build GRCh38
rs745438799 Chr.5: 60644794-60644794 on Build GRCh38
rs1015276831 Chr.5: 60644955-60644955 on Build GRCh38
rs755338750 Chr.5: 60644733-60644733 on Build GRCh38
rs866802545 Chr.5: 60644985-60644985 on Build GRCh38
rs559836099 Chr.5: 60644657-60644657 on Build GRCh38
rs1022320234 Chr.5: 60644945-60644946 on Build GRCh38
rs762153345 Chr.5: 60644705-60644705 on Build GRCh38
rs755678171 Chr.5: 60644903-60644903 on Build GRCh38
rs958694243 Chr.5: 60644999-60644999 on Build GRCh38
rs753822246 Chr.5: 60644862-60644862 on Build GRCh38
rs757931727 Chr.5: 60644842-60644842 on Build GRCh38
rs768462930 Chr.5: 60644696-60644696 on Build GRCh38
rs758746895 Chr.5: 60644899-60644899 on Build GRCh38
rs201451342 Chr.5: 60644813-60644813 on Build GRCh38
rs112568063 Chr.5: 60644829-60644829 on Build GRCh38
rs994726175 Chr.5: 60644965-60644965 on Build GRCh38
rs200114483 Chr.5: 60644872-60644872 on Build GRCh38
rs990406134 Chr.5: 60644940-60644940 on Build GRCh38
rs190950359 Chr.5: 60644900-60644900 on Build GRCh38
rs142916456 Chr.5: 60644773-60644773 on Build GRCh38
rs973381930 Chr.5: 60644606-60644606 on Build GRCh38
rs760102134 Chr.5: 60644871-60644871 on Build GRCh38
rs779367812 Chr.5: 60644753-60644753 on Build GRCh38
rs750538428 Chr.5: 60644718-60644718 on Build GRCh38
rs760889560 Chr.5: 60644850-60644850 on Build GRCh38
rs542247916 Chr.5: 60644726-60644726 on Build GRCh38
rs576994140 Chr.5: 60644623-60644623 on Build GRCh38
rs141096982 Chr.5: 60644812-60644812 on Build GRCh38
rs753947164 Chr.5: 60644875-60644876 on Build GRCh38
rs575737262 Chr.5: 60644810-60644810 on Build GRCh38
rs760834825 Chr.5: 60644722-60644722 on Build GRCh38
rs528131102 Chr.5: 60644998-60644998 on Build GRCh38
rs766451194 Chr.5: 60644857-60644857 on Build GRCh38
rs946230518 Chr.5: 60644751-60644751 on Build GRCh38
rs944122201 Chr.5: 60645044-60645044 on Build GRCh38
rs764321128 Chr.5: 60644838-60644840 on Build GRCh38
rs200675292 Chr.5: 60644830-60644830 on Build GRCh38
rs894791935 Chr.5: 60644795-60644795 on Build GRCh38
rs185959881 Chr.5: 60645005-60645005 on Build GRCh38
rs773973011 Chr.5: 60644704-60644704 on Build GRCh38
rs992889237 Chr.5: 60644686-60644686 on Build GRCh38
rs369140199 Chr.5: 60644709-60644709 on Build GRCh38
rs111289794 Chr.5: 60644594-60644594 on Build GRCh38
rs993356729 Chr.5: 60645002-60645002 on Build GRCh38
rs2061252 Chr.5: 60644964-60644964 on Build GRCh38
rs368367512 Chr.5: 60644772-60644772 on Build GRCh38
rs917447464 Chr.5: 60644712-60644712 on Build GRCh38
rs780816165 Chr.5: 60644787-60644787 on Build GRCh38
rs6877561 Chr.5: 60644675-60644675 on Build GRCh38
rs753001310 Chr.5: 60644760-60644760 on Build GRCh38
rs151101076 Chr.5: 60644806-60644806 on Build GRCh38
rs1004707116 Chr.5: 60644926-60644926 on Build GRCh38

View Full Product Details