Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132542955-132543490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL5
Entrez Gene ID
3567
Gene Name
interleukin 5
Gene Aliases
EDF, IL-5, TRF
UniGene
Hs.2247
Gene Chromosome Location
Chr.5: 132539194-132556827 on Build GRCh38
Transcripts
NM_000879.2 XM_005271988.3 XM_011543373.2 XM_011543374.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6056042 c.5G>T p.R2M Chr.5: 132543474-132543474 on Build GRCh38
6168676 c.83C>A p.A28E Chr.5: 132543396-132543396 on Build GRCh38
5923695 c.73C>T p.P25S Chr.5: 132543406-132543406 on Build GRCh38
1060664 c.121C>T p.R41* Chr.5: 132543358-132543358 on Build GRCh38
7113789 c.48C>T p.Y16Y Chr.5: 132543431-132543431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755713715 Chr.5: 132543402-132543404 on Build GRCh38
rs753662432 Chr.5: 132543051-132543051 on Build GRCh38
rs747193100 Chr.5: 132543358-132543358 on Build GRCh38
rs1800474 Chr.5: 132543356-132543356 on Build GRCh38
rs139238685 Chr.5: 132543334-132543334 on Build GRCh38
rs746231797 Chr.5: 132543100-132543100 on Build GRCh38
rs185977955 Chr.5: 132543463-132543463 on Build GRCh38
rs768300703 Chr.5: 132543465-132543465 on Build GRCh38
rs780238566 Chr.5: 132543335-132543335 on Build GRCh38
rs909530658 Chr.5: 132543031-132543031 on Build GRCh38
rs897264262 Chr.5: 132543376-132543376 on Build GRCh38
rs779089112 Chr.5: 132543342-132543342 on Build GRCh38
rs34328342 Chr.5: 132543044-132543045 on Build GRCh38
rs181065673 Chr.5: 132543125-132543125 on Build GRCh38
rs368875825 Chr.5: 132543345-132543345 on Build GRCh38
rs566875704 Chr.5: 132543363-132543363 on Build GRCh38
rs764851944 Chr.5: 132543305-132543305 on Build GRCh38
rs893788025 Chr.5: 132543214-132543214 on Build GRCh38
rs1044218622 Chr.5: 132543169-132543169 on Build GRCh38
rs775355739 Chr.5: 132543360-132543360 on Build GRCh38
rs758195556 Chr.5: 132543311-132543311 on Build GRCh38
rs200785840 Chr.5: 132543069-132543069 on Build GRCh38
rs550846478 Chr.5: 132542993-132542993 on Build GRCh38
rs775126409 Chr.5: 132543373-132543373 on Build GRCh38
rs769576892 Chr.5: 132543450-132543450 on Build GRCh38
rs769846245 Chr.5: 132543249-132543249 on Build GRCh38
rs972399853 Chr.5: 132543372-132543372 on Build GRCh38
rs762568489 Chr.5: 132543379-132543379 on Build GRCh38
rs201524603 Chr.5: 132543430-132543430 on Build GRCh38
rs897211748 Chr.5: 132543063-132543063 on Build GRCh38
rs775296895 Chr.5: 132543451-132543451 on Build GRCh38
rs751285038 Chr.5: 132543391-132543391 on Build GRCh38
rs745640961 Chr.5: 132543445-132543445 on Build GRCh38
rs755574063 Chr.5: 132543428-132543428 on Build GRCh38
rs184698719 Chr.5: 132543150-132543150 on Build GRCh38
rs781719328 Chr.5: 132543093-132543093 on Build GRCh38
rs752420524 Chr.5: 132543088-132543088 on Build GRCh38
rs771028408 Chr.5: 132543441-132543441 on Build GRCh38
rs754523453 Chr.5: 132543068-132543068 on Build GRCh38
rs755418324 Chr.5: 132543339-132543339 on Build GRCh38
rs2069815 Chr.5: 132543318-132543318 on Build GRCh38
rs189973397 Chr.5: 132543282-132543282 on Build GRCh38
rs957904341 Chr.5: 132543153-132543153 on Build GRCh38
rs371004662 Chr.5: 132543338-132543338 on Build GRCh38
rs201297950 Chr.5: 132543381-132543381 on Build GRCh38
rs945116757 Chr.5: 132543207-132543207 on Build GRCh38
rs200264275 Chr.5: 132543456-132543456 on Build GRCh38
rs1009595663 Chr.5: 132543218-132543218 on Build GRCh38
rs754121375 Chr.5: 132543426-132543426 on Build GRCh38
rs112946922 Chr.5: 132543371-132543371 on Build GRCh38
rs781112205 Chr.5: 132543442-132543442 on Build GRCh38
rs566899580 Chr.5: 132543241-132543241 on Build GRCh38
rs770483682 Chr.5: 132543107-132543107 on Build GRCh38
rs199954909 Chr.5: 132543146-132543146 on Build GRCh38
rs202132888 Chr.5: 132543294-132543294 on Build GRCh38
rs766944689 Chr.5: 132543158-132543165 on Build GRCh38
rs1050043156 Chr.5: 132543337-132543337 on Build GRCh38
rs146633313 Chr.5: 132543431-132543431 on Build GRCh38
rs1014365012 Chr.5: 132543420-132543420 on Build GRCh38
rs901398216 Chr.5: 132543225-132543225 on Build GRCh38
rs748593806 Chr.5: 132543429-132543429 on Build GRCh38
rs770735768 Chr.5: 132543359-132543359 on Build GRCh38
rs772336303 Chr.5: 132543351-132543351 on Build GRCh38
rs140074297 Chr.5: 132543404-132543404 on Build GRCh38
rs539027225 Chr.5: 132543256-132543256 on Build GRCh38
rs1001939635 Chr.5: 132543460-132543460 on Build GRCh38
rs1021384840 Chr.5: 132543221-132543221 on Build GRCh38
rs374667414 Chr.5: 132543174-132543174 on Build GRCh38
rs150829213 Chr.5: 132543111-132543111 on Build GRCh38
rs2069822 Chr.5: 132543190-132543190 on Build GRCh38
rs751197081 Chr.5: 132543327-132543327 on Build GRCh38
rs752121509 Chr.5: 132543306-132543306 on Build GRCh38
rs752943234 Chr.5: 132543166-132543166 on Build GRCh38
rs34909832 Chr.5: 132543071-132543072 on Build GRCh38
rs766810951 Chr.5: 132543398-132543398 on Build GRCh38
rs778250389 Chr.5: 132543078-132543078 on Build GRCh38
rs149977915 Chr.5: 132543361-132543361 on Build GRCh38
rs759343155 Chr.5: 132543297-132543297 on Build GRCh38
rs760447335 Chr.5: 132543168-132543168 on Build GRCh38
rs201066544 Chr.5: 132543357-132543357 on Build GRCh38
rs370838683 Chr.5: 132543397-132543397 on Build GRCh38
rs776665689 Chr.5: 132543289-132543289 on Build GRCh38
rs1028442766 Chr.5: 132543303-132543303 on Build GRCh38
rs373235478 Chr.5: 132543033-132543033 on Build GRCh38
rs922313565 Chr.5: 132543224-132543224 on Build GRCh38
rs772248130 Chr.5: 132543156-132543156 on Build GRCh38
rs765991275 Chr.5: 132543441-132543441 on Build GRCh38
rs749963235 Chr.5: 132543286-132543295 on Build GRCh38
rs989233985 Chr.5: 132543165-132543165 on Build GRCh38
rs976840224 Chr.5: 132543216-132543216 on Build GRCh38

View Full Product Details