Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138445957-138446450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REEP2
Entrez Gene ID
51308
Gene Name
receptor accessory protein 2
Gene Aliases
C5orf19, SGC32445, SPG72, Yip2d
UniGene
Hs.416090
Gene Chromosome Location
Chr.5: 138439001-138446969 on Build GRCh38
Transcripts
NM_001271803.1 NM_016606.3 NR_073448.1 NR_073449.1

SNP Detail

SNP ID Chromosome Location
rs892952543 Chr.5: 138446320-138446320 on Build GRCh38
rs567743349 Chr.5: 138446030-138446030 on Build GRCh38
rs892157724 Chr.5: 138446140-138446140 on Build GRCh38
rs923991842 Chr.5: 138446267-138446267 on Build GRCh38
rs997600085 Chr.5: 138446171-138446171 on Build GRCh38
rs1004719118 Chr.5: 138446272-138446272 on Build GRCh38
rs748796941 Chr.5: 138446393-138446396 on Build GRCh38
rs746705617 Chr.5: 138446143-138446143 on Build GRCh38
rs943187251 Chr.5: 138446328-138446328 on Build GRCh38
rs770453005 Chr.5: 138446281-138446281 on Build GRCh38
rs559754711 Chr.5: 138446216-138446216 on Build GRCh38
rs909370700 Chr.5: 138446201-138446201 on Build GRCh38
rs997481588 Chr.5: 138446408-138446408 on Build GRCh38
rs866647967 Chr.5: 138446022-138446022 on Build GRCh38
rs547963293 Chr.5: 138446162-138446162 on Build GRCh38
rs1054273774 Chr.5: 138446139-138446139 on Build GRCh38
rs528903885 Chr.5: 138446148-138446148 on Build GRCh38
rs1038935477 Chr.5: 138446352-138446352 on Build GRCh38
rs918164870 Chr.5: 138446208-138446208 on Build GRCh38
rs575389338 Chr.5: 138446105-138446105 on Build GRCh38
rs552517619 Chr.5: 138445995-138445995 on Build GRCh38
rs1000999259 Chr.5: 138446137-138446137 on Build GRCh38
rs955683106 Chr.5: 138446011-138446011 on Build GRCh38
rs1015557082 Chr.5: 138446029-138446029 on Build GRCh38
rs147656297 Chr.5: 138446192-138446192 on Build GRCh38
rs991006674 Chr.5: 138446207-138446207 on Build GRCh38
rs867046978 Chr.5: 138446175-138446175 on Build GRCh38
rs564374449 Chr.5: 138446286-138446286 on Build GRCh38
rs909424456 Chr.5: 138446041-138446041 on Build GRCh38
rs947983458 Chr.5: 138446098-138446105 on Build GRCh38
rs748472169 Chr.5: 138446203-138446203 on Build GRCh38
rs1042781277 Chr.5: 138446222-138446222 on Build GRCh38
rs559846254 Chr.5: 138446040-138446040 on Build GRCh38
rs1026381804 Chr.5: 138446411-138446411 on Build GRCh38
rs1052155259 Chr.5: 138446120-138446120 on Build GRCh38
rs4835796 Chr.5: 138446426-138446426 on Build GRCh38
rs757847555 Chr.5: 138446048-138446048 on Build GRCh38
rs920535805 Chr.5: 138446053-138446053 on Build GRCh38
rs1055331962 Chr.5: 138446086-138446086 on Build GRCh38
rs563865197 Chr.5: 138446296-138446296 on Build GRCh38
rs950516859 Chr.5: 138446196-138446196 on Build GRCh38
rs984528834 Chr.5: 138446198-138446198 on Build GRCh38
rs35989890 Chr.5: 138446146-138446147 on Build GRCh38
rs902602518 Chr.5: 138446122-138446122 on Build GRCh38
rs901749894 Chr.5: 138446405-138446405 on Build GRCh38
rs939098433 Chr.5: 138446113-138446113 on Build GRCh38
rs773786147 Chr.5: 138446393-138446393 on Build GRCh38
rs182406448 Chr.5: 138446144-138446144 on Build GRCh38
rs927815623 Chr.5: 138446104-138446104 on Build GRCh38
rs962750417 Chr.5: 138446318-138446318 on Build GRCh38
rs530144954 Chr.5: 138446218-138446218 on Build GRCh38
rs186402775 Chr.5: 138446184-138446184 on Build GRCh38
rs938246746 Chr.5: 138446062-138446062 on Build GRCh38
rs917886847 Chr.5: 138446090-138446090 on Build GRCh38
rs535345279 Chr.5: 138446100-138446100 on Build GRCh38
rs893005084 Chr.5: 138446142-138446142 on Build GRCh38
rs4835795 Chr.5: 138446103-138446103 on Build GRCh38

View Full Product Details