Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140127049-140127549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGIP
Entrez Gene ID
492311
Gene Name
IgA inducing protein
Gene Aliases
C5orf53
UniGene
Hs.696360
Gene Chromosome Location
Chr.5: 140125936-140128806 on Build GRCh38
Transcripts
NM_001007189.1

SNP Detail

SNP ID Chromosome Location
rs112520221 Chr.5: 140127144-140127144 on Build GRCh38
rs1040173268 Chr.5: 140127373-140127373 on Build GRCh38
rs928173450 Chr.5: 140127291-140127291 on Build GRCh38
rs962406122 Chr.5: 140127169-140127169 on Build GRCh38
rs763670134 Chr.5: 140127330-140127330 on Build GRCh38
rs544948154 Chr.5: 140127125-140127125 on Build GRCh38
rs969938568 Chr.5: 140127225-140127225 on Build GRCh38
rs542799233 Chr.5: 140127364-140127364 on Build GRCh38
rs1019961913 Chr.5: 140127327-140127327 on Build GRCh38
rs879128085 Chr.5: 140127433-140127433 on Build GRCh38
rs1001398135 Chr.5: 140127274-140127274 on Build GRCh38
rs1013977657 Chr.5: 140127309-140127309 on Build GRCh38
rs929898601 Chr.5: 140127198-140127198 on Build GRCh38
rs554723566 Chr.5: 140127170-140127170 on Build GRCh38
rs778678613 Chr.5: 140127495-140127496 on Build GRCh38
rs1048270549 Chr.5: 140127246-140127246 on Build GRCh38
rs1056030457 Chr.5: 140127275-140127275 on Build GRCh38
rs770747327 Chr.5: 140127281-140127281 on Build GRCh38
rs1016861804 Chr.5: 140127100-140127100 on Build GRCh38
rs574207746 Chr.5: 140127448-140127448 on Build GRCh38
rs572285130 Chr.5: 140127191-140127191 on Build GRCh38
rs771910098 Chr.5: 140127163-140127164 on Build GRCh38
rs759175373 Chr.5: 140127465-140127465 on Build GRCh38
rs372162365 Chr.5: 140127207-140127207 on Build GRCh38
rs888347675 Chr.5: 140127258-140127258 on Build GRCh38
rs982315926 Chr.5: 140127263-140127263 on Build GRCh38
rs34224597 Chr.5: 140127320-140127321 on Build GRCh38
rs946747042 Chr.5: 140127369-140127369 on Build GRCh38
rs145736775 Chr.5: 140127174-140127174 on Build GRCh38
rs921374581 Chr.5: 140127403-140127403 on Build GRCh38
rs560974771 Chr.5: 140127367-140127367 on Build GRCh38

View Full Product Details