Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 142310743-142311241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRY4
Entrez Gene ID
81848
Gene Name
sprouty RTK signaling antagonist 4
Gene Aliases
HH17
UniGene
Hs.323308
Gene Chromosome Location
Chr.5: 142310427-142325055 on Build GRCh38
Transcripts
NM_001127496.1 NM_001293289.1 NM_001293290.1 NM_030964.3 XM_011537685.2 XM_017009910.1

SNP Detail

SNP ID Chromosome Location
rs777915578 Chr.5: 142311216-142311216 on Build GRCh38
rs999618559 Chr.5: 142311054-142311054 on Build GRCh38
rs991863108 Chr.5: 142311087-142311087 on Build GRCh38
rs140546932 Chr.5: 142310981-142310981 on Build GRCh38
rs1031037824 Chr.5: 142310920-142310920 on Build GRCh38
rs532868993 Chr.5: 142311019-142311019 on Build GRCh38
rs539519230 Chr.5: 142310941-142310941 on Build GRCh38
rs923700796 Chr.5: 142310840-142310840 on Build GRCh38
rs566884810 Chr.5: 142311021-142311021 on Build GRCh38
rs558333132 Chr.5: 142311064-142311064 on Build GRCh38
rs769852226 Chr.5: 142311215-142311215 on Build GRCh38
rs72801999 Chr.5: 142310798-142310798 on Build GRCh38
rs538025484 Chr.5: 142311063-142311063 on Build GRCh38
rs146590995 Chr.5: 142311126-142311126 on Build GRCh38
rs568993184 Chr.5: 142311030-142311030 on Build GRCh38
rs1010928990 Chr.5: 142311147-142311147 on Build GRCh38
rs145679094 Chr.5: 142311072-142311072 on Build GRCh38
rs532810547 Chr.5: 142310945-142310945 on Build GRCh38
rs149853026 Chr.5: 142310853-142310853 on Build GRCh38
rs113186785 Chr.5: 142311199-142311199 on Build GRCh38
rs546178807 Chr.5: 142310954-142310954 on Build GRCh38
rs72801997 Chr.5: 142310784-142310784 on Build GRCh38
rs933213517 Chr.5: 142310783-142310783 on Build GRCh38
rs375909403 Chr.5: 142311202-142311202 on Build GRCh38
rs530644704 Chr.5: 142310870-142310870 on Build GRCh38
rs1038248200 Chr.5: 142310903-142310903 on Build GRCh38
rs956920631 Chr.5: 142310943-142310943 on Build GRCh38
rs1037414974 Chr.5: 142310866-142310866 on Build GRCh38
rs1022399192 Chr.5: 142310951-142310951 on Build GRCh38
rs113022449 Chr.5: 142310785-142310786 on Build GRCh38
rs948007317 Chr.5: 142310897-142310897 on Build GRCh38
rs907834187 Chr.5: 142311165-142311165 on Build GRCh38
rs1004192738 Chr.5: 142310908-142310908 on Build GRCh38
rs781494430 Chr.5: 142311130-142311130 on Build GRCh38
rs987791395 Chr.5: 142311040-142311040 on Build GRCh38
rs574208325 Chr.5: 142310790-142310790 on Build GRCh38
rs115695299 Chr.5: 142311141-142311141 on Build GRCh38
rs559272751 Chr.5: 142310784-142310785 on Build GRCh38
rs145695366 Chr.5: 142310895-142310895 on Build GRCh38
rs906689514 Chr.5: 142311123-142311123 on Build GRCh38
rs907744849 Chr.5: 142311051-142311051 on Build GRCh38
rs3839201 Chr.5: 142311199-142311199 on Build GRCh38
rs1045187891 Chr.5: 142310893-142310893 on Build GRCh38
rs867564483 Chr.5: 142310919-142310919 on Build GRCh38
rs6874376 Chr.5: 142310959-142310959 on Build GRCh38
rs964150928 Chr.5: 142311185-142311185 on Build GRCh38
rs1022285948 Chr.5: 142311154-142311154 on Build GRCh38
rs397882959 Chr.5: 142311200-142311200 on Build GRCh38
rs368038214 Chr.5: 142311200-142311200 on Build GRCh38
rs113427374 Chr.5: 142310889-142310889 on Build GRCh38
rs920364539 Chr.5: 142311066-142311066 on Build GRCh38
rs554158619 Chr.5: 142311129-142311129 on Build GRCh38
rs1023464161 Chr.5: 142311122-142311122 on Build GRCh38
rs190918583 Chr.5: 142310980-142310980 on Build GRCh38
rs996924314 Chr.5: 142311203-142311203 on Build GRCh38
rs1045237281 Chr.5: 142311074-142311074 on Build GRCh38
rs1030628291 Chr.5: 142311056-142311056 on Build GRCh38
rs371568182 Chr.5: 142311201-142311201 on Build GRCh38
rs920619683 Chr.5: 142310934-142310934 on Build GRCh38
rs900661565 Chr.5: 142310997-142310997 on Build GRCh38
rs752616895 Chr.5: 142310944-142310946 on Build GRCh38

View Full Product Details