Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149981783-149982284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC26A2
Entrez Gene ID
1836
Gene Name
solute carrier family 26 member 2
Gene Aliases
D5S1708, DTD, DTDST, EDM4, MST153, MSTP157
UniGene
Hs.302738
Gene Chromosome Location
Chr.5: 149960729-149987400 on Build GRCh38
Transcripts
NM_000112.3 XM_017009191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1634061 c.2206C>A p.L736I Chr.5: 149981799-149981799 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs932757555 Chr.5: 149981926-149981926 on Build GRCh38
rs748275663 Chr.5: 149981822-149981822 on Build GRCh38
rs969153746 Chr.5: 149982052-149982052 on Build GRCh38
rs560524828 Chr.5: 149982081-149982081 on Build GRCh38
rs529112280 Chr.5: 149981882-149981882 on Build GRCh38
rs998171839 Chr.5: 149981834-149981834 on Build GRCh38
rs932656670 Chr.5: 149982213-149982213 on Build GRCh38
rs1001883031 Chr.5: 149982061-149982061 on Build GRCh38
rs552470458 Chr.5: 149982089-149982089 on Build GRCh38
rs775182119 Chr.5: 149981838-149981838 on Build GRCh38
rs934808344 Chr.5: 149982247-149982247 on Build GRCh38
rs529744947 Chr.5: 149982017-149982017 on Build GRCh38
rs377375559 Chr.5: 149981821-149981821 on Build GRCh38
rs886060228 Chr.5: 149982077-149982077 on Build GRCh38
rs570796999 Chr.5: 149982113-149982113 on Build GRCh38
rs757369599 Chr.5: 149982110-149982110 on Build GRCh38
rs769986063 Chr.5: 149981825-149981825 on Build GRCh38
rs1051193183 Chr.5: 149981938-149981938 on Build GRCh38
rs999027543 Chr.5: 149981959-149981959 on Build GRCh38
rs930336319 Chr.5: 149982072-149982072 on Build GRCh38
rs893413322 Chr.5: 149982005-149982005 on Build GRCh38
rs1000234877 Chr.5: 149982223-149982223 on Build GRCh38
rs143847430 Chr.5: 149981896-149981896 on Build GRCh38
rs891274631 Chr.5: 149981945-149981945 on Build GRCh38
rs1033105432 Chr.5: 149981965-149981965 on Build GRCh38
rs562629009 Chr.5: 149981948-149981948 on Build GRCh38
rs535585042 Chr.5: 149982241-149982241 on Build GRCh38
rs763101767 Chr.5: 149981831-149981831 on Build GRCh38
rs191884433 Chr.5: 149982143-149982143 on Build GRCh38
rs886060226 Chr.5: 149981866-149981866 on Build GRCh38
rs115977282 Chr.5: 149982256-149982256 on Build GRCh38
rs770082455 Chr.5: 149981995-149981995 on Build GRCh38
rs766854098 Chr.5: 149981823-149981826 on Build GRCh38
rs956059188 Chr.5: 149981973-149981973 on Build GRCh38
rs1051142063 Chr.5: 149982238-149982238 on Build GRCh38
rs775062734 Chr.5: 149981828-149981830 on Build GRCh38
rs1050012102 Chr.5: 149982206-149982206 on Build GRCh38
rs115383424 Chr.5: 149982018-149982018 on Build GRCh38
rs1034732914 Chr.5: 149982070-149982070 on Build GRCh38
rs954088228 Chr.5: 149981868-149981868 on Build GRCh38
rs1028298398 Chr.5: 149981866-149981866 on Build GRCh38
rs760414785 Chr.5: 149981846-149981846 on Build GRCh38
rs891909766 Chr.5: 149982249-149982249 on Build GRCh38
rs934205916 Chr.5: 149982157-149982157 on Build GRCh38
rs568578594 Chr.5: 149982221-149982221 on Build GRCh38
rs907658315 Chr.5: 149982109-149982109 on Build GRCh38
rs775637237 Chr.5: 149982033-149982033 on Build GRCh38
rs140641532 Chr.5: 149982207-149982207 on Build GRCh38
rs552562764 Chr.5: 149982125-149982125 on Build GRCh38
rs771121032 Chr.5: 149981833-149981833 on Build GRCh38
rs146843578 Chr.5: 149982083-149982083 on Build GRCh38
rs773546058 Chr.5: 149981826-149981826 on Build GRCh38
rs886060227 Chr.5: 149982029-149982029 on Build GRCh38
rs761224496 Chr.5: 149981807-149981814 on Build GRCh38
rs370711127 Chr.5: 149982202-149982202 on Build GRCh38
rs753464677 Chr.5: 149982034-149982035 on Build GRCh38
rs1024164474 Chr.5: 149982006-149982006 on Build GRCh38
rs61732052 Chr.5: 149981813-149981813 on Build GRCh38
rs1038813518 Chr.5: 149981859-149981859 on Build GRCh38
rs926914921 Chr.5: 149982038-149982038 on Build GRCh38
rs960493031 Chr.5: 149982092-149982092 on Build GRCh38
rs747357127 Chr.5: 149981811-149981811 on Build GRCh38
rs1022405083 Chr.5: 149982029-149982029 on Build GRCh38

View Full Product Details