Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150057206-150057665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF1R
Entrez Gene ID
1436
Gene Name
colony stimulating factor 1 receptor
Gene Aliases
C-FMS, CD115, CSF-1R, CSFR, FIM2, FMS, HDLS, M-CSF-R
UniGene
Hs.586219
Gene Chromosome Location
Chr.5: 150053291-150113372 on Build GRCh38
Transcripts
NM_001288705.1 NM_005211.3 NR_109969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5967327 c.2292_2319+8del p.? Chr.5: 150057287-150057314 on Build GRCh38
6933688 c.2291A>C p.Q764P Chr.5: 150057315-150057315 on Build GRCh38
5608166 c.2133G>A p.R711R Chr.5: 150057592-150057592 on Build GRCh38
5019283 c.2239G>A p.G747R Chr.5: 150057367-150057367 on Build GRCh38
1202374 c.2211C>A p.F737L Chr.5: 150057514-150057514 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199915312 Chr.5: 150057508-150057508 on Build GRCh38
rs887139619 Chr.5: 150057522-150057522 on Build GRCh38
rs777325801 Chr.5: 150057580-150057580 on Build GRCh38
rs375119067 Chr.5: 150057239-150057239 on Build GRCh38
rs777951907 Chr.5: 150057491-150057491 on Build GRCh38
rs144976075 Chr.5: 150057362-150057362 on Build GRCh38
rs760200293 Chr.5: 150057605-150057605 on Build GRCh38
rs751713758 Chr.5: 150057377-150057382 on Build GRCh38
rs764658321 Chr.5: 150057409-150057409 on Build GRCh38
rs368898219 Chr.5: 150057237-150057237 on Build GRCh38
rs376570011 Chr.5: 150057559-150057559 on Build GRCh38
rs577596858 Chr.5: 150057229-150057229 on Build GRCh38
rs143167751 Chr.5: 150057517-150057517 on Build GRCh38
rs762540571 Chr.5: 150057416-150057416 on Build GRCh38
rs755412466 Chr.5: 150057363-150057363 on Build GRCh38
rs369879651 Chr.5: 150057373-150057373 on Build GRCh38
rs768790029 Chr.5: 150057260-150057260 on Build GRCh38
rs374818714 Chr.5: 150057402-150057402 on Build GRCh38
rs959559690 Chr.5: 150057270-150057270 on Build GRCh38
rs574305529 Chr.5: 150057420-150057420 on Build GRCh38
rs769685450 Chr.5: 150057578-150057578 on Build GRCh38
rs772597939 Chr.5: 150057617-150057617 on Build GRCh38
rs748747560 Chr.5: 150057590-150057590 on Build GRCh38
rs746764799 Chr.5: 150057510-150057510 on Build GRCh38
rs768704008 Chr.5: 150057515-150057515 on Build GRCh38
rs751851291 Chr.5: 150057457-150057457 on Build GRCh38
rs748111236 Chr.5: 150057576-150057576 on Build GRCh38
rs746033718 Chr.5: 150057572-150057572 on Build GRCh38
rs769312420 Chr.5: 150057519-150057519 on Build GRCh38
rs775511069 Chr.5: 150057335-150057335 on Build GRCh38
rs281860271 Chr.5: 150057298-150057298 on Build GRCh38
rs200830545 Chr.5: 150057400-150057400 on Build GRCh38
rs749554100 Chr.5: 150057497-150057497 on Build GRCh38
rs753281770 Chr.5: 150057543-150057543 on Build GRCh38
rs370577638 Chr.5: 150057640-150057640 on Build GRCh38
rs372068592 Chr.5: 150057349-150057349 on Build GRCh38
rs781385572 Chr.5: 150057364-150057364 on Build GRCh38
rs761500096 Chr.5: 150057347-150057347 on Build GRCh38
rs764054281 Chr.5: 150057247-150057248 on Build GRCh38
rs376651450 Chr.5: 150057353-150057353 on Build GRCh38
rs780417451 Chr.5: 150057634-150057635 on Build GRCh38
rs780914237 Chr.5: 150057575-150057575 on Build GRCh38
rs777889423 Chr.5: 150057520-150057520 on Build GRCh38
rs774848316 Chr.5: 150057533-150057533 on Build GRCh38
rs690016566 Chr.5: 150057312-150057312 on Build GRCh38
rs968512218 Chr.5: 150057556-150057556 on Build GRCh38
rs761311600 Chr.5: 150057625-150057625 on Build GRCh38
rs371017407 Chr.5: 150057560-150057560 on Build GRCh38
rs749367958 Chr.5: 150057592-150057592 on Build GRCh38
rs373083937 Chr.5: 150057361-150057361 on Build GRCh38
rs749466218 Chr.5: 150057368-150057368 on Build GRCh38
rs771928787 Chr.5: 150057334-150057334 on Build GRCh38
rs747406297 Chr.5: 150057249-150057249 on Build GRCh38
rs1042396588 Chr.5: 150057553-150057553 on Build GRCh38
rs748228488 Chr.5: 150057366-150057366 on Build GRCh38
rs41355444 Chr.5: 150057367-150057367 on Build GRCh38
rs216138 Chr.5: 150057627-150057627 on Build GRCh38
rs1021348545 Chr.5: 150057228-150057228 on Build GRCh38
rs781080752 Chr.5: 150057290-150057290 on Build GRCh38
rs373100109 Chr.5: 150057238-150057238 on Build GRCh38
rs781457547 Chr.5: 150057569-150057570 on Build GRCh38
rs772862480 Chr.5: 150057411-150057411 on Build GRCh38
rs898751485 Chr.5: 150057473-150057473 on Build GRCh38
rs568840375 Chr.5: 150057476-150057476 on Build GRCh38
rs1041149647 Chr.5: 150057539-150057539 on Build GRCh38
rs281860270 Chr.5: 150057309-150057309 on Build GRCh38
rs759826953 Chr.5: 150057535-150057535 on Build GRCh38
rs763690501 Chr.5: 150057338-150057338 on Build GRCh38
rs542316497 Chr.5: 150057337-150057337 on Build GRCh38
rs756649081 Chr.5: 150057548-150057548 on Build GRCh38
rs775967780 Chr.5: 150057622-150057622 on Build GRCh38
rs1031529577 Chr.5: 150057565-150057565 on Build GRCh38
rs199640967 Chr.5: 150057299-150057299 on Build GRCh38
rs532265671 Chr.5: 150057417-150057417 on Build GRCh38
rs374630808 Chr.5: 150057642-150057642 on Build GRCh38
rs767909181 Chr.5: 150057471-150057471 on Build GRCh38
rs775108440 Chr.5: 150057383-150057383 on Build GRCh38
rs753256015 Chr.5: 150057343-150057343 on Build GRCh38
rs1021454741 Chr.5: 150057551-150057551 on Build GRCh38
rs757402879 Chr.5: 150057524-150057524 on Build GRCh38
rs372508167 Chr.5: 150057401-150057401 on Build GRCh38
rs750645188 Chr.5: 150057611-150057612 on Build GRCh38
rs373164720 Chr.5: 150057601-150057601 on Build GRCh38
rs975624605 Chr.5: 150057274-150057274 on Build GRCh38
rs756411343 Chr.5: 150057627-150057629 on Build GRCh38
rs780603197 Chr.5: 150057240-150057240 on Build GRCh38
rs774348652 Chr.5: 150057307-150057307 on Build GRCh38
rs377690615 Chr.5: 150057611-150057611 on Build GRCh38
rs41287096 Chr.5: 150057227-150057227 on Build GRCh38
rs778996649 Chr.5: 150057568-150057568 on Build GRCh38
rs560352241 Chr.5: 150057348-150057348 on Build GRCh38
rs759403025 Chr.5: 150057323-150057323 on Build GRCh38
rs145711151 Chr.5: 150057236-150057236 on Build GRCh38
rs754740710 Chr.5: 150057574-150057574 on Build GRCh38
rs931586898 Chr.5: 150057478-150057478 on Build GRCh38
rs748019030 Chr.5: 150057518-150057518 on Build GRCh38
rs142978503 Chr.5: 150057558-150057558 on Build GRCh38
rs772656669 Chr.5: 150057589-150057589 on Build GRCh38
rs374820700 Chr.5: 150057632-150057632 on Build GRCh38
rs373427460 Chr.5: 150057527-150057527 on Build GRCh38
rs747007270 Chr.5: 150057387-150057387 on Build GRCh38
rs200193476 Chr.5: 150057468-150057468 on Build GRCh38
rs139061724 Chr.5: 150057540-150057540 on Build GRCh38
rs773889293 Chr.5: 150057602-150057602 on Build GRCh38
rs758764883 Chr.5: 150057351-150057351 on Build GRCh38
rs899883054 Chr.5: 150057547-150057547 on Build GRCh38
rs915620614 Chr.5: 150057432-150057432 on Build GRCh38
rs754016717 Chr.5: 150057630-150057630 on Build GRCh38
rs767100188 Chr.5: 150057481-150057481 on Build GRCh38

View Full Product Details