Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 156647374-156647873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGCD
Entrez Gene ID
6444
Gene Name
sarcoglycan delta
Gene Aliases
35DAG, CMD1L, DAGD, SG-delta, SGCDP, SGD
UniGene
Hs.387207
Gene Chromosome Location
Chr.5: 155728636-156767788 on Build GRCh38
Transcripts
NM_000337.5 NM_001128209.1 NM_172244.2 XM_005265966.4 XM_005265967.2 XM_011534621.2 XM_017009723.1 XM_017009724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4552729 c.568G>A p.E190K Chr.5: 156647529-156647529 on Build GRCh38
3613015 c.522C>T p.F174F Chr.5: 156647483-156647483 on Build GRCh38
5413863 c.524C>T p.P175L Chr.5: 156647485-156647485 on Build GRCh38
4552728 c.568G>A p.E190K Chr.5: 156647529-156647529 on Build GRCh38
6169845 c.559C>A p.P187T Chr.5: 156647520-156647520 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931206627 Chr.5: 156647755-156647755 on Build GRCh38
rs115312312 Chr.5: 156647697-156647697 on Build GRCh38
rs187924293 Chr.5: 156647766-156647766 on Build GRCh38
rs547385035 Chr.5: 156647414-156647414 on Build GRCh38
rs778724633 Chr.5: 156647481-156647481 on Build GRCh38
rs141437738 Chr.5: 156647645-156647651 on Build GRCh38
rs576593385 Chr.5: 156647741-156647741 on Build GRCh38
rs896886297 Chr.5: 156647685-156647685 on Build GRCh38
rs918011162 Chr.5: 156647740-156647740 on Build GRCh38
rs759981619 Chr.5: 156647423-156647423 on Build GRCh38
rs769250018 Chr.5: 156647520-156647520 on Build GRCh38
rs753952200 Chr.5: 156647477-156647477 on Build GRCh38
rs889587203 Chr.5: 156647795-156647795 on Build GRCh38
rs748900550 Chr.5: 156647747-156647747 on Build GRCh38
rs1049686171 Chr.5: 156647785-156647785 on Build GRCh38
rs772427118 Chr.5: 156647769-156647769 on Build GRCh38
rs376209679 Chr.5: 156647559-156647559 on Build GRCh38
rs896157702 Chr.5: 156647709-156647709 on Build GRCh38
rs923276685 Chr.5: 156647613-156647613 on Build GRCh38
rs757279786 Chr.5: 156647494-156647494 on Build GRCh38
rs760523974 Chr.5: 156647816-156647816 on Build GRCh38
rs773886933 Chr.5: 156647421-156647421 on Build GRCh38
rs1005711488 Chr.5: 156647417-156647417 on Build GRCh38
rs748854138 Chr.5: 156647580-156647580 on Build GRCh38
rs754243570 Chr.5: 156647588-156647589 on Build GRCh38
rs200176911 Chr.5: 156647648-156647648 on Build GRCh38
rs554934712 Chr.5: 156647629-156647629 on Build GRCh38
rs967944466 Chr.5: 156647526-156647526 on Build GRCh38
rs919967605 Chr.5: 156647753-156647753 on Build GRCh38
rs574766030 Chr.5: 156647634-156647634 on Build GRCh38
rs545887965 Chr.5: 156647712-156647712 on Build GRCh38
rs538002169 Chr.5: 156647527-156647527 on Build GRCh38
rs923265218 Chr.5: 156647846-156647846 on Build GRCh38
rs780862246 Chr.5: 156647534-156647534 on Build GRCh38
rs368838376 Chr.5: 156647471-156647471 on Build GRCh38
rs876658001 Chr.5: 156647467-156647467 on Build GRCh38
rs879244683 Chr.5: 156647661-156647661 on Build GRCh38
rs886044432 Chr.5: 156647542-156647542 on Build GRCh38
rs1050997677 Chr.5: 156647412-156647412 on Build GRCh38
rs940013005 Chr.5: 156647445-156647445 on Build GRCh38
rs32055 Chr.5: 156647703-156647703 on Build GRCh38
rs1040730132 Chr.5: 156647810-156647810 on Build GRCh38
rs1023848898 Chr.5: 156647506-156647506 on Build GRCh38
rs992212573 Chr.5: 156647736-156647736 on Build GRCh38
rs747907940 Chr.5: 156647537-156647537 on Build GRCh38
rs116258549 Chr.5: 156647733-156647733 on Build GRCh38
rs570898631 Chr.5: 156647422-156647422 on Build GRCh38
rs542004220 Chr.5: 156647815-156647815 on Build GRCh38
rs1021328124 Chr.5: 156647635-156647635 on Build GRCh38
rs201963763 Chr.5: 156647647-156647647 on Build GRCh38
rs932236177 Chr.5: 156647401-156647401 on Build GRCh38
rs10071079 Chr.5: 156647468-156647468 on Build GRCh38
rs1019335531 Chr.5: 156647570-156647570 on Build GRCh38
rs140311698 Chr.5: 156647647-156647648 on Build GRCh38
rs764237334 Chr.5: 156647472-156647472 on Build GRCh38
rs896813117 Chr.5: 156647444-156647444 on Build GRCh38
rs968694226 Chr.5: 156647674-156647674 on Build GRCh38
rs533820377 Chr.5: 156647691-156647691 on Build GRCh38
rs200451694 Chr.5: 156647504-156647504 on Build GRCh38
rs868554008 Chr.5: 156647469-156647469 on Build GRCh38
rs773735436 Chr.5: 156647811-156647811 on Build GRCh38
rs770530590 Chr.5: 156647582-156647582 on Build GRCh38
rs527714222 Chr.5: 156647411-156647411 on Build GRCh38
rs755861001 Chr.5: 156647543-156647543 on Build GRCh38
rs751421543 Chr.5: 156647508-156647508 on Build GRCh38
rs769459793 Chr.5: 156647644-156647645 on Build GRCh38
rs976169291 Chr.5: 156647845-156647845 on Build GRCh38
rs372967785 Chr.5: 156647453-156647453 on Build GRCh38
rs943881556 Chr.5: 156647798-156647798 on Build GRCh38
rs3832352 Chr.5: 156647647-156647653 on Build GRCh38
rs935422875 Chr.5: 156647571-156647571 on Build GRCh38

View Full Product Details