Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 160598680-160599186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP10B
Entrez Gene ID
23120
Gene Name
ATPase phospholipid transporting 10B (putative)
Gene Aliases
ATPVB
UniGene
Hs.109358
Gene Chromosome Location
Chr.5: 160563120-160929411 on Build GRCh38
Transcripts
NM_025153.2 XM_006714833.2 XM_011534468.2 XM_011534469.1 XM_011534472.2 XM_017009252.1 XM_017009253.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4601633 c.3498C>A p.D1166E Chr.5: 160598836-160598836 on Build GRCh38
1065223 c.3483C>A p.V1161V Chr.5: 160598851-160598851 on Build GRCh38
1065222 c.3507C>A p.I1169I Chr.5: 160598827-160598827 on Build GRCh38
3247576 c.3369C>T p.Y1123Y Chr.5: 160598965-160598965 on Build GRCh38
3613468 c.3408C>T p.F1136F Chr.5: 160598926-160598926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368682194 Chr.5: 160598878-160598878 on Build GRCh38
rs759285990 Chr.5: 160598894-160598894 on Build GRCh38
rs370433512 Chr.5: 160598953-160598953 on Build GRCh38
rs770052169 Chr.5: 160599157-160599157 on Build GRCh38
rs747392154 Chr.5: 160598861-160598861 on Build GRCh38
rs999340796 Chr.5: 160598744-160598744 on Build GRCh38
rs56811519 Chr.5: 160599120-160599120 on Build GRCh38
rs374867390 Chr.5: 160598722-160598722 on Build GRCh38
rs61734662 Chr.5: 160598892-160598892 on Build GRCh38
rs748330253 Chr.5: 160599055-160599055 on Build GRCh38
rs375493659 Chr.5: 160598819-160598819 on Build GRCh38
rs537902467 Chr.5: 160599088-160599088 on Build GRCh38
rs766188751 Chr.5: 160598890-160598890 on Build GRCh38
rs995379082 Chr.5: 160598860-160598860 on Build GRCh38
rs377651496 Chr.5: 160599031-160599031 on Build GRCh38
rs17480493 Chr.5: 160599018-160599018 on Build GRCh38
rs376724032 Chr.5: 160599001-160599001 on Build GRCh38
rs759252566 Chr.5: 160599003-160599003 on Build GRCh38
rs770432631 Chr.5: 160598987-160598987 on Build GRCh38
rs140530527 Chr.5: 160599063-160599064 on Build GRCh38
rs751319442 Chr.5: 160598799-160598799 on Build GRCh38
rs891012831 Chr.5: 160598814-160598814 on Build GRCh38
rs761298581 Chr.5: 160598785-160598785 on Build GRCh38
rs372351351 Chr.5: 160598812-160598812 on Build GRCh38
rs989675762 Chr.5: 160598836-160598836 on Build GRCh38
rs1018243247 Chr.5: 160598822-160598822 on Build GRCh38
rs774123754 Chr.5: 160598934-160598936 on Build GRCh38
rs767464886 Chr.5: 160598897-160598897 on Build GRCh38
rs373444145 Chr.5: 160598999-160598999 on Build GRCh38
rs772883953 Chr.5: 160598887-160598887 on Build GRCh38
rs772794149 Chr.5: 160598988-160598988 on Build GRCh38
rs758605751 Chr.5: 160598964-160598964 on Build GRCh38
rs186650804 Chr.5: 160599117-160599117 on Build GRCh38
rs746048369 Chr.5: 160598842-160598842 on Build GRCh38
rs567281421 Chr.5: 160598965-160598965 on Build GRCh38
rs748870369 Chr.5: 160598996-160598996 on Build GRCh38
rs1050709348 Chr.5: 160599006-160599006 on Build GRCh38
rs995722080 Chr.5: 160599080-160599080 on Build GRCh38
rs777046244 Chr.5: 160598867-160598867 on Build GRCh38
rs768201680 Chr.5: 160598866-160598866 on Build GRCh38
rs764077152 Chr.5: 160599017-160599017 on Build GRCh38
rs749381947 Chr.5: 160598740-160598740 on Build GRCh38
rs772371139 Chr.5: 160598756-160598756 on Build GRCh38
rs753919903 Chr.5: 160598811-160598811 on Build GRCh38
rs778897678 Chr.5: 160598924-160598924 on Build GRCh38
rs980984147 Chr.5: 160598947-160598947 on Build GRCh38
rs893655617 Chr.5: 160599093-160599093 on Build GRCh38
rs761177277 Chr.5: 160598759-160598759 on Build GRCh38
rs191261996 Chr.5: 160599143-160599143 on Build GRCh38
rs925566830 Chr.5: 160598952-160598952 on Build GRCh38
rs144973457 Chr.5: 160598974-160598974 on Build GRCh38
rs756076021 Chr.5: 160598915-160598915 on Build GRCh38
rs774592448 Chr.5: 160598747-160598747 on Build GRCh38
rs766241787 Chr.5: 160598798-160598798 on Build GRCh38
rs772285071 Chr.5: 160598843-160598843 on Build GRCh38
rs542967515 Chr.5: 160599129-160599129 on Build GRCh38
rs771178091 Chr.5: 160598742-160598742 on Build GRCh38
rs762631498 Chr.5: 160598889-160598889 on Build GRCh38
rs748565686 Chr.5: 160598986-160598986 on Build GRCh38
rs776191749 Chr.5: 160598779-160598779 on Build GRCh38
rs373302217 Chr.5: 160598882-160598882 on Build GRCh38
rs139029862 Chr.5: 160599027-160599027 on Build GRCh38
rs762719323 Chr.5: 160598794-160598794 on Build GRCh38
rs769058029 Chr.5: 160598776-160598776 on Build GRCh38
rs746208164 Chr.5: 160598749-160598749 on Build GRCh38
rs912943192 Chr.5: 160598982-160598982 on Build GRCh38
rs747125105 Chr.5: 160598901-160598901 on Build GRCh38
rs183826088 Chr.5: 160598787-160598787 on Build GRCh38
rs781054316 Chr.5: 160598803-160598803 on Build GRCh38
rs752526269 Chr.5: 160598907-160598907 on Build GRCh38
rs762916325 Chr.5: 160599141-160599143 on Build GRCh38
rs373405927 Chr.5: 160598859-160598859 on Build GRCh38
rs201047636 Chr.5: 160599010-160599010 on Build GRCh38
rs768952031 Chr.5: 160598978-160598978 on Build GRCh38
rs969402760 Chr.5: 160598900-160598900 on Build GRCh38
rs538813679 Chr.5: 160598829-160598829 on Build GRCh38
rs1025485128 Chr.5: 160599086-160599086 on Build GRCh38
rs571435566 Chr.5: 160598757-160598757 on Build GRCh38
rs371345553 Chr.5: 160599016-160599016 on Build GRCh38
rs1010600437 Chr.5: 160599101-160599102 on Build GRCh38
rs781638558 Chr.5: 160598984-160598984 on Build GRCh38
rs558443588 Chr.5: 160599068-160599068 on Build GRCh38
rs368078734 Chr.5: 160598791-160598791 on Build GRCh38
rs920794273 Chr.5: 160598903-160598903 on Build GRCh38
rs575456051 Chr.5: 160599084-160599084 on Build GRCh38
rs887153689 Chr.5: 160599105-160599105 on Build GRCh38
rs767175036 Chr.5: 160599012-160599012 on Build GRCh38
rs754926803 Chr.5: 160598801-160598801 on Build GRCh38
rs988347126 Chr.5: 160598979-160598979 on Build GRCh38
rs377316993 Chr.5: 160598866-160598866 on Build GRCh38
rs369899160 Chr.5: 160598840-160598840 on Build GRCh38
rs775357935 Chr.5: 160599013-160599013 on Build GRCh38
rs775863902 Chr.5: 160598758-160598758 on Build GRCh38

View Full Product Details